Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30112299-30112878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD3
Entrez Gene ID
79153
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
GDE7
UniGene
Hs.289015
Gene Chromosome Location
Chr.16: 30104810-30113557 on Build GRCh38
Transcripts
NM_024307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670379 c.418C>A p.Q140K Chr.16: 30112371-30112371 on Build GRCh38
4917481 c.250G>A p.V84M Chr.16: 30112726-30112726 on Build GRCh38
6670380 c.275G>A p.R92H Chr.16: 30112701-30112701 on Build GRCh38
969779 c.466G>A p.E156K Chr.16: 30112323-30112323 on Build GRCh38
3794787 c.420G>T p.Q140H Chr.16: 30112369-30112369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770112694 Chr.16: 30112451-30112451 on Build GRCh38
rs200678682 Chr.16: 30112397-30112397 on Build GRCh38
rs779696097 Chr.16: 30112522-30112522 on Build GRCh38
rs374835162 Chr.16: 30112398-30112398 on Build GRCh38
rs561889339 Chr.16: 30112506-30112506 on Build GRCh38
rs749615608 Chr.16: 30112678-30112678 on Build GRCh38
rs906775871 Chr.16: 30112476-30112476 on Build GRCh38
rs143649081 Chr.16: 30112467-30112467 on Build GRCh38
rs748529299 Chr.16: 30112450-30112450 on Build GRCh38
rs746705329 Chr.16: 30112528-30112528 on Build GRCh38
rs375485050 Chr.16: 30112751-30112751 on Build GRCh38
rs76435425 Chr.16: 30112388-30112388 on Build GRCh38
rs367918036 Chr.16: 30112701-30112701 on Build GRCh38
rs1018816054 Chr.16: 30112694-30112694 on Build GRCh38
rs771446176 Chr.16: 30112466-30112466 on Build GRCh38
rs775629140 Chr.16: 30112474-30112474 on Build GRCh38
rs201916465 Chr.16: 30112505-30112505 on Build GRCh38
rs898823207 Chr.16: 30112389-30112390 on Build GRCh38
rs61764619 Chr.16: 30112743-30112743 on Build GRCh38
rs768792433 Chr.16: 30112708-30112708 on Build GRCh38
rs762792576 Chr.16: 30112829-30112829 on Build GRCh38
rs769710666 Chr.16: 30112502-30112508 on Build GRCh38
rs928667830 Chr.16: 30112792-30112810 on Build GRCh38
rs774855444 Chr.16: 30112813-30112814 on Build GRCh38
rs773620006 Chr.16: 30112455-30112455 on Build GRCh38
rs776763054 Chr.16: 30112345-30112352 on Build GRCh38
rs950144814 Chr.16: 30112638-30112638 on Build GRCh38
rs780570506 Chr.16: 30112799-30112799 on Build GRCh38
rs752449266 Chr.16: 30112776-30112776 on Build GRCh38
rs201068732 Chr.16: 30112520-30112520 on Build GRCh38
rs886210424 Chr.16: 30112441-30112441 on Build GRCh38
rs773160281 Chr.16: 30112826-30112826 on Build GRCh38
rs771455266 Chr.16: 30112562-30112562 on Build GRCh38
rs199833751 Chr.16: 30112537-30112537 on Build GRCh38
rs776313331 Chr.16: 30112619-30112619 on Build GRCh38
rs767055474 Chr.16: 30112807-30112807 on Build GRCh38
rs758404610 Chr.16: 30112656-30112656 on Build GRCh38
rs1004708839 Chr.16: 30112675-30112675 on Build GRCh38
rs763866245 Chr.16: 30112494-30112494 on Build GRCh38
rs370168697 Chr.16: 30112411-30112411 on Build GRCh38
rs372903211 Chr.16: 30112439-30112439 on Build GRCh38
rs138427877 Chr.16: 30112695-30112695 on Build GRCh38
rs1033199644 Chr.16: 30112853-30112853 on Build GRCh38
rs751057196 Chr.16: 30112660-30112660 on Build GRCh38
rs1055263935 Chr.16: 30112800-30112800 on Build GRCh38
rs367999022 Chr.16: 30112782-30112782 on Build GRCh38
rs770115803 Chr.16: 30112820-30112820 on Build GRCh38
rs192236532 Chr.16: 30112775-30112775 on Build GRCh38
rs754609869 Chr.16: 30112661-30112661 on Build GRCh38
rs749612150 Chr.16: 30112560-30112560 on Build GRCh38
rs139302544 Chr.16: 30112345-30112345 on Build GRCh38
rs748838280 Chr.16: 30112324-30112324 on Build GRCh38
rs752244188 Chr.16: 30112833-30112853 on Build GRCh38
rs767727591 Chr.16: 30112471-30112471 on Build GRCh38
rs747062560 Chr.16: 30112703-30112703 on Build GRCh38
rs202089119 Chr.16: 30112623-30112623 on Build GRCh38
rs761629427 Chr.16: 30112628-30112628 on Build GRCh38
rs201511312 Chr.16: 30112744-30112744 on Build GRCh38
rs769777350 Chr.16: 30112734-30112734 on Build GRCh38
rs376867572 Chr.16: 30112593-30112593 on Build GRCh38
rs930633968 Chr.16: 30112831-30112831 on Build GRCh38
rs763349663 Chr.16: 30112585-30112585 on Build GRCh38
rs143388174 Chr.16: 30112325-30112325 on Build GRCh38
rs879242982 Chr.16: 30112561-30112561 on Build GRCh38
rs750330096 Chr.16: 30112650-30112650 on Build GRCh38
rs965291638 Chr.16: 30112493-30112493 on Build GRCh38
rs764359103 Chr.16: 30112606-30112606 on Build GRCh38
rs761529339 Chr.16: 30112726-30112726 on Build GRCh38
rs773384722 Chr.16: 30112581-30112581 on Build GRCh38
rs745958173 Chr.16: 30112574-30112574 on Build GRCh38
rs778611106 Chr.16: 30112431-30112431 on Build GRCh38
rs945809323 Chr.16: 30112760-30112760 on Build GRCh38
rs147449044 Chr.16: 30112401-30112401 on Build GRCh38
rs371568180 Chr.16: 30112512-30112512 on Build GRCh38
rs775547129 Chr.16: 30112600-30112600 on Build GRCh38
rs572381199 Chr.16: 30112389-30112389 on Build GRCh38
rs569651014 Chr.16: 30112323-30112323 on Build GRCh38
rs775485096 Chr.16: 30112702-30112702 on Build GRCh38
rs777781970 Chr.16: 30112400-30112400 on Build GRCh38
rs1033258564 Chr.16: 30112581-30112581 on Build GRCh38
rs781121631 Chr.16: 30112818-30112818 on Build GRCh38
rs746443339 Chr.16: 30112698-30112698 on Build GRCh38
rs748405421 Chr.16: 30112826-30112827 on Build GRCh38
rs751057816 Chr.16: 30112747-30112747 on Build GRCh38
rs921636694 Chr.16: 30112378-30112378 on Build GRCh38
rs780854305 Chr.16: 30112674-30112674 on Build GRCh38
rs766435548 Chr.16: 30112377-30112377 on Build GRCh38
rs373928234 Chr.16: 30112320-30112320 on Build GRCh38
rs911785911 Chr.16: 30112327-30112327 on Build GRCh38
rs763409065 Chr.16: 30112464-30112464 on Build GRCh38
rs773531646 Chr.16: 30112369-30112369 on Build GRCh38
rs74746402 Chr.16: 30112625-30112625 on Build GRCh38
rs886507963 Chr.16: 30112845-30112845 on Build GRCh38
rs779814542 Chr.16: 30112448-30112448 on Build GRCh38
rs199955545 Chr.16: 30112399-30112399 on Build GRCh38
rs58842311 Chr.16: 30112813-30112813 on Build GRCh38
rs770777307 Chr.16: 30112838-30112838 on Build GRCh38
rs199660924 Chr.16: 30112367-30112367 on Build GRCh38
rs1009454383 Chr.16: 30112456-30112456 on Build GRCh38
rs975021184 Chr.16: 30112541-30112541 on Build GRCh38
rs766026177 Chr.16: 30112657-30112657 on Build GRCh38
rs375857248 Chr.16: 30112633-30112633 on Build GRCh38
rs776846892 Chr.16: 30112709-30112709 on Build GRCh38
rs760850563 Chr.16: 30112603-30112603 on Build GRCh38
rs758578117 Chr.16: 30112629-30112629 on Build GRCh38
rs758930580 Chr.16: 30112783-30112783 on Build GRCh38
rs372092356 Chr.16: 30112410-30112410 on Build GRCh38
rs755761103 Chr.16: 30112779-30112779 on Build GRCh38
rs535489009 Chr.16: 30112362-30112362 on Build GRCh38
rs549898215 Chr.16: 30112780-30112780 on Build GRCh38
rs748413197 Chr.16: 30112388-30112403 on Build GRCh38
rs760941351 Chr.16: 30112478-30112478 on Build GRCh38
rs774770045 Chr.16: 30112573-30112573 on Build GRCh38
rs771873871 Chr.16: 30112445-30112445 on Build GRCh38
rs759914644 Chr.16: 30112815-30112816 on Build GRCh38
rs763502423 Chr.16: 30112370-30112370 on Build GRCh38
rs762221083 Chr.16: 30112363-30112363 on Build GRCh38
rs772199432 Chr.16: 30112586-30112586 on Build GRCh38

View Full Product Details