Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87407194-87407786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
UniGene
Hs.156231
Gene Chromosome Location
Chr.16: 87406246-87492948 on Build GRCh38
Transcripts
NM_015144.2

SNP Detail

SNP ID Chromosome Location
rs1050863 Chr.16: 87407289-87407289 on Build GRCh38
rs116337491 Chr.16: 87407682-87407682 on Build GRCh38
rs926925353 Chr.16: 87407239-87407239 on Build GRCh38
rs1012700925 Chr.16: 87407269-87407269 on Build GRCh38
rs759564863 Chr.16: 87407351-87407351 on Build GRCh38
rs143930775 Chr.16: 87407681-87407681 on Build GRCh38
rs536137568 Chr.16: 87407293-87407293 on Build GRCh38
rs1015237787 Chr.16: 87407316-87407316 on Build GRCh38
rs938640263 Chr.16: 87407493-87407493 on Build GRCh38
rs1016068602 Chr.16: 87407477-87407479 on Build GRCh38
rs989605087 Chr.16: 87407384-87407384 on Build GRCh38
rs536280895 Chr.16: 87407438-87407438 on Build GRCh38
rs945520764 Chr.16: 87407306-87407306 on Build GRCh38
rs940583728 Chr.16: 87407691-87407691 on Build GRCh38
rs1005224085 Chr.16: 87407470-87407470 on Build GRCh38
rs992814779 Chr.16: 87407280-87407280 on Build GRCh38
rs898998266 Chr.16: 87407729-87407729 on Build GRCh38
rs899009718 Chr.16: 87407493-87407494 on Build GRCh38
rs1015269017 Chr.16: 87407679-87407679 on Build GRCh38
rs1034046127 Chr.16: 87407735-87407735 on Build GRCh38
rs894296716 Chr.16: 87407235-87407235 on Build GRCh38
rs574173917 Chr.16: 87407597-87407598 on Build GRCh38
rs774304839 Chr.16: 87407309-87407309 on Build GRCh38
rs1001353211 Chr.16: 87407732-87407732 on Build GRCh38
rs922040210 Chr.16: 87407650-87407650 on Build GRCh38
rs752791376 Chr.16: 87407497-87407497 on Build GRCh38
rs933298786 Chr.16: 87407669-87407669 on Build GRCh38
rs992532134 Chr.16: 87407551-87407551 on Build GRCh38
rs34068017 Chr.16: 87407444-87407445 on Build GRCh38
rs886380541 Chr.16: 87407463-87407463 on Build GRCh38
rs906505478 Chr.16: 87407461-87407461 on Build GRCh38
rs966959089 Chr.16: 87407600-87407600 on Build GRCh38
rs965624234 Chr.16: 87407325-87407325 on Build GRCh38
rs895548416 Chr.16: 87407753-87407754 on Build GRCh38
rs1037582672 Chr.16: 87407702-87407702 on Build GRCh38
rs575004453 Chr.16: 87407420-87407420 on Build GRCh38
rs925456556 Chr.16: 87407341-87407341 on Build GRCh38
rs977002380 Chr.16: 87407339-87407339 on Build GRCh38
rs544044821 Chr.16: 87407651-87407651 on Build GRCh38
rs936861691 Chr.16: 87407631-87407631 on Build GRCh38
rs932800412 Chr.16: 87407372-87407372 on Build GRCh38
rs533984917 Chr.16: 87407626-87407626 on Build GRCh38
rs2241617 Chr.16: 87407442-87407442 on Build GRCh38
rs1034098529 Chr.16: 87407508-87407509 on Build GRCh38
rs138371721 Chr.16: 87407629-87407629 on Build GRCh38
rs1057418243 Chr.16: 87407502-87407502 on Build GRCh38
rs573016889 Chr.16: 87407449-87407449 on Build GRCh38
rs2241616 Chr.16: 87407511-87407511 on Build GRCh38
rs865820471 Chr.16: 87407410-87407410 on Build GRCh38
rs1001198280 Chr.16: 87407495-87407495 on Build GRCh38
rs1039140610 Chr.16: 87407387-87407387 on Build GRCh38
rs959688787 Chr.16: 87407515-87407517 on Build GRCh38
rs974815627 Chr.16: 87407649-87407649 on Build GRCh38
rs1027136474 Chr.16: 87407271-87407271 on Build GRCh38
rs764322339 Chr.16: 87407641-87407641 on Build GRCh38
rs556831853 Chr.16: 87407348-87407348 on Build GRCh38

View Full Product Details