Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43925269-43925770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF45
Entrez Gene ID
7596
Gene Name
zinc finger protein 45
Gene Aliases
KOX5, ZNF13
UniGene
Hs.381285
Gene Chromosome Location
Chr.19: 43912624-43935278 on Build GRCh38
Transcripts
NM_003425.3 XM_011527267.1 XM_011527269.1 XM_011527271.1 XM_011527273.1 XM_017027217.1 XM_017027218.1 XM_017027219.1 XM_017027220.1 XM_017027221.1 XM_017027222.1 XM_017027223.1 XM_017027224.1 XM_017027225.1 XM_017027226.1 XM_017027227.1 XM_017027228.1 XM_017027229.1 XM_017027230.1

SNP Detail

SNP ID Chromosome Location
rs1018895883 Chr.19: 43925367-43925367 on Build GRCh38
rs1022754647 Chr.19: 43925395-43925395 on Build GRCh38
rs994517342 Chr.19: 43925306-43925306 on Build GRCh38
rs989526001 Chr.19: 43925443-43925443 on Build GRCh38
rs73036776 Chr.19: 43925417-43925417 on Build GRCh38
rs444350 Chr.19: 43925317-43925317 on Build GRCh38
rs966094265 Chr.19: 43925475-43925475 on Build GRCh38
rs945432091 Chr.19: 43925450-43925451 on Build GRCh38
rs151076660 Chr.19: 43925445-43925445 on Build GRCh38
rs906807664 Chr.19: 43925476-43925476 on Build GRCh38
rs150255714 Chr.19: 43925714-43925714 on Build GRCh38
rs528141167 Chr.19: 43925561-43925561 on Build GRCh38
rs557599764 Chr.19: 43925622-43925625 on Build GRCh38
rs752250740 Chr.19: 43925599-43925599 on Build GRCh38
rs371709848 Chr.19: 43925563-43925571 on Build GRCh38
rs1019040086 Chr.19: 43925459-43925459 on Build GRCh38
rs561901565 Chr.19: 43925654-43925654 on Build GRCh38
rs114798521 Chr.19: 43925655-43925655 on Build GRCh38
rs369412892 Chr.19: 43925571-43925571 on Build GRCh38
rs551231810 Chr.19: 43925562-43925567 on Build GRCh38
rs937053960 Chr.19: 43925716-43925716 on Build GRCh38
rs932733478 Chr.19: 43925433-43925433 on Build GRCh38
rs62116568 Chr.19: 43925326-43925326 on Build GRCh38
rs777463234 Chr.19: 43925603-43925603 on Build GRCh38
rs112513663 Chr.19: 43925606-43925606 on Build GRCh38
rs984723111 Chr.19: 43925640-43925640 on Build GRCh38
rs765248400 Chr.19: 43925517-43925517 on Build GRCh38
rs757002408 Chr.19: 43925670-43925670 on Build GRCh38
rs145713184 Chr.19: 43925325-43925325 on Build GRCh38
rs571104059 Chr.19: 43925735-43925735 on Build GRCh38
rs200984090 Chr.19: 43925564-43925571 on Build GRCh38
rs895283648 Chr.19: 43925562-43925570 on Build GRCh38
rs1011081617 Chr.19: 43925348-43925348 on Build GRCh38
rs868175822 Chr.19: 43925493-43925493 on Build GRCh38
rs371365650 Chr.19: 43925324-43925324 on Build GRCh38
rs966339705 Chr.19: 43925406-43925406 on Build GRCh38
rs895407729 Chr.19: 43925327-43925327 on Build GRCh38
rs940475029 Chr.19: 43925296-43925296 on Build GRCh38
rs769808225 Chr.19: 43925409-43925409 on Build GRCh38
rs939574129 Chr.19: 43925522-43925522 on Build GRCh38
rs907557387 Chr.19: 43925650-43925650 on Build GRCh38
rs199804753 Chr.19: 43925559-43925565 on Build GRCh38
rs444247 Chr.19: 43925410-43925410 on Build GRCh38
rs553474422 Chr.19: 43925569-43925571 on Build GRCh38
rs757889517 Chr.19: 43925600-43925600 on Build GRCh38
rs894915971 Chr.19: 43925299-43925299 on Build GRCh38
rs866501722 Chr.19: 43925729-43925729 on Build GRCh38
rs957230649 Chr.19: 43925739-43925739 on Build GRCh38
rs975033892 Chr.19: 43925703-43925703 on Build GRCh38
rs761716526 Chr.19: 43925323-43925323 on Build GRCh38
rs978608734 Chr.19: 43925432-43925432 on Build GRCh38

View Full Product Details