Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41357679-41358118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B9D2
Entrez Gene ID
80776
Gene Name
B9 protein domain 2
Gene Aliases
ICIS-1, MKS10, MKSR2
UniGene
Hs.567596
Gene Chromosome Location
Chr.19: 41354417-41364540 on Build GRCh38
Transcripts
NM_030578.3 XM_011527349.2 XM_011527350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078507 c.115G>A p.V39M Chr.19: 41357996-41357996 on Build GRCh38
7416572 c.94G>A p.A32T Chr.19: 41358017-41358017 on Build GRCh38
7137432 c.150G>C p.Q50H Chr.19: 41357961-41357961 on Build GRCh38
5936430 c.145C>T p.P49S Chr.19: 41357966-41357966 on Build GRCh38
439590 c.202A>G p.K68E Chr.19: 41357909-41357909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368434905 Chr.19: 41357997-41357997 on Build GRCh38
rs762649671 Chr.19: 41357853-41357853 on Build GRCh38
rs766906829 Chr.19: 41357900-41357900 on Build GRCh38
rs776771096 Chr.19: 41357875-41357875 on Build GRCh38
rs150023579 Chr.19: 41357980-41357980 on Build GRCh38
rs972379533 Chr.19: 41357975-41357975 on Build GRCh38
rs375676828 Chr.19: 41357927-41357927 on Build GRCh38
rs1050505012 Chr.19: 41358020-41358020 on Build GRCh38
rs370560381 Chr.19: 41357992-41357992 on Build GRCh38
rs541603457 Chr.19: 41357790-41357790 on Build GRCh38
rs1005811224 Chr.19: 41357841-41357841 on Build GRCh38
rs1050929719 Chr.19: 41357806-41357806 on Build GRCh38
rs751469905 Chr.19: 41358019-41358019 on Build GRCh38
rs751082879 Chr.19: 41357953-41357954 on Build GRCh38
rs756761176 Chr.19: 41357847-41357847 on Build GRCh38
rs754889797 Chr.19: 41358025-41358025 on Build GRCh38
rs994123927 Chr.19: 41357857-41357857 on Build GRCh38
rs781151351 Chr.19: 41358027-41358027 on Build GRCh38
rs139520227 Chr.19: 41357970-41357970 on Build GRCh38
rs886054457 Chr.19: 41358002-41358002 on Build GRCh38
rs757908270 Chr.19: 41357948-41357948 on Build GRCh38
rs757863670 Chr.19: 41358004-41358004 on Build GRCh38
rs769974706 Chr.19: 41357964-41357964 on Build GRCh38
rs761105034 Chr.19: 41358066-41358066 on Build GRCh38
rs901887502 Chr.19: 41357749-41357749 on Build GRCh38
rs201182397 Chr.19: 41357844-41357844 on Build GRCh38
rs755963627 Chr.19: 41358030-41358030 on Build GRCh38
rs114133150 Chr.19: 41358065-41358065 on Build GRCh38
rs763870135 Chr.19: 41357852-41357853 on Build GRCh38
rs765029105 Chr.19: 41357944-41357944 on Build GRCh38
rs142424836 Chr.19: 41358018-41358018 on Build GRCh38
rs34088631 Chr.19: 41357916-41357916 on Build GRCh38
rs772884480 Chr.19: 41358073-41358073 on Build GRCh38
rs368393895 Chr.19: 41357960-41357960 on Build GRCh38
rs772020545 Chr.19: 41357853-41357853 on Build GRCh38
rs761158280 Chr.19: 41357996-41357996 on Build GRCh38
rs746928334 Chr.19: 41357954-41357954 on Build GRCh38
rs935611541 Chr.19: 41357729-41357729 on Build GRCh38
rs757133492 Chr.19: 41357966-41357966 on Build GRCh38
rs867282292 Chr.19: 41357986-41357986 on Build GRCh38
rs904549746 Chr.19: 41357855-41357855 on Build GRCh38
rs73931466 Chr.19: 41358089-41358089 on Build GRCh38
rs372716136 Chr.19: 41357984-41357984 on Build GRCh38
rs745743114 Chr.19: 41357850-41357850 on Build GRCh38
rs770930532 Chr.19: 41357971-41357971 on Build GRCh38
rs140901774 Chr.19: 41357928-41357928 on Build GRCh38
rs890576595 Chr.19: 41357807-41357807 on Build GRCh38
rs564188704 Chr.19: 41357735-41357735 on Build GRCh38
rs185832302 Chr.19: 41357730-41357730 on Build GRCh38
rs78999030 Chr.19: 41357796-41357796 on Build GRCh38
rs577400270 Chr.19: 41357783-41357783 on Build GRCh38
rs773256220 Chr.19: 41357893-41357893 on Build GRCh38
rs780023869 Chr.19: 41357951-41357951 on Build GRCh38
rs374618635 Chr.19: 41358053-41358053 on Build GRCh38
rs777993645 Chr.19: 41358032-41358032 on Build GRCh38
rs746810819 Chr.19: 41357863-41357863 on Build GRCh38
rs375582943 Chr.19: 41357933-41357933 on Build GRCh38
rs924184336 Chr.19: 41357724-41357724 on Build GRCh38
rs370955358 Chr.19: 41357876-41357876 on Build GRCh38
rs749915355 Chr.19: 41357947-41357947 on Build GRCh38
rs769818136 Chr.19: 41357883-41357883 on Build GRCh38
rs371483684 Chr.19: 41358037-41358037 on Build GRCh38
rs191089800 Chr.19: 41358079-41358079 on Build GRCh38
rs778595061 Chr.19: 41357849-41357849 on Build GRCh38
rs1053043709 Chr.19: 41358080-41358080 on Build GRCh38
rs867075613 Chr.19: 41357717-41357717 on Build GRCh38
rs773375953 Chr.19: 41357965-41357965 on Build GRCh38
rs11668109 Chr.19: 41357872-41357872 on Build GRCh38
rs1000123002 Chr.19: 41357866-41357866 on Build GRCh38
rs747933592 Chr.19: 41358029-41358029 on Build GRCh38
rs946875358 Chr.19: 41357716-41357716 on Build GRCh38
rs779893483 Chr.19: 41357858-41357858 on Build GRCh38
rs745849111 Chr.19: 41358043-41358043 on Build GRCh38
rs769152584 Chr.19: 41358070-41358070 on Build GRCh38
rs756990381 Chr.19: 41357940-41357940 on Build GRCh38
rs774854310 Chr.19: 41357979-41357979 on Build GRCh38
rs1050943988 Chr.19: 41357792-41357792 on Build GRCh38
rs747988749 Chr.19: 41357961-41357961 on Build GRCh38
rs772675261 Chr.19: 41358064-41358064 on Build GRCh38
rs778881892 Chr.19: 41358050-41358050 on Build GRCh38

View Full Product Details