Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48577741-48578314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM3
Entrez Gene ID
5935
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Gene Aliases
IS1-RNPL, RNPL
UniGene
Hs.301404
Gene Chromosome Location
Chr.X: 48574353-48581165 on Build GRCh38
Transcripts
NM_006743.4

SNP Detail

SNP ID Chromosome Location
rs782444094 Chr.X: 48578238-48578238 on Build GRCh38
rs139329149 Chr.X: 48578275-48578275 on Build GRCh38
rs41307347 Chr.X: 48578005-48578005 on Build GRCh38
rs894472178 Chr.X: 48577932-48577932 on Build GRCh38
rs920151794 Chr.X: 48578107-48578107 on Build GRCh38
rs1022207784 Chr.X: 48577967-48577967 on Build GRCh38
rs7065384 Chr.X: 48578022-48578022 on Build GRCh38
rs978304775 Chr.X: 48578000-48578000 on Build GRCh38
rs188646118 Chr.X: 48577884-48577884 on Build GRCh38
rs8931 Chr.X: 48578119-48578119 on Build GRCh38
rs374726233 Chr.X: 48578013-48578013 on Build GRCh38
rs3200611 Chr.X: 48578038-48578038 on Build GRCh38
rs928828385 Chr.X: 48578226-48578226 on Build GRCh38
rs782759357 Chr.X: 48578117-48578117 on Build GRCh38
rs1047241596 Chr.X: 48577872-48577872 on Build GRCh38
rs781815728 Chr.X: 48578012-48578012 on Build GRCh38
rs782528762 Chr.X: 48578150-48578150 on Build GRCh38
rs1034318317 Chr.X: 48577898-48577898 on Build GRCh38
rs782044857 Chr.X: 48577979-48577979 on Build GRCh38
rs782177260 Chr.X: 48578204-48578204 on Build GRCh38
rs35340315 Chr.X: 48577995-48577996 on Build GRCh38
rs930124912 Chr.X: 48577802-48577802 on Build GRCh38
rs1002962968 Chr.X: 48577895-48577895 on Build GRCh38
rs181435733 Chr.X: 48577954-48577954 on Build GRCh38
rs112276526 Chr.X: 48578285-48578285 on Build GRCh38
rs1136985 Chr.X: 48578058-48578058 on Build GRCh38
rs782670858 Chr.X: 48578177-48578177 on Build GRCh38
rs782134647 Chr.X: 48577857-48577857 on Build GRCh38
rs938883453 Chr.X: 48578237-48578237 on Build GRCh38

View Full Product Details