Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100597340-100597889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNMD
Entrez Gene ID
64102
Gene Name
tenomodulin
Gene Aliases
BRICD4, CHM1L, TEM
UniGene
Hs.132957
Gene Chromosome Location
Chr.X: 100584793-100599885 on Build GRCh38
Transcripts
NM_022144.2

SNP Detail

SNP ID Chromosome Location
rs771584316 Chr.X: 100597525-100597525 on Build GRCh38
rs771443528 Chr.X: 100597750-100597750 on Build GRCh38
rs866286351 Chr.X: 100597516-100597516 on Build GRCh38
rs1034694292 Chr.X: 100597762-100597762 on Build GRCh38
rs757698917 Chr.X: 100597671-100597671 on Build GRCh38
rs767540371 Chr.X: 100597573-100597573 on Build GRCh38
rs912774272 Chr.X: 100597402-100597402 on Build GRCh38
rs755288765 Chr.X: 100597601-100597601 on Build GRCh38
rs758382654 Chr.X: 100597629-100597629 on Build GRCh38
rs1026040465 Chr.X: 100597641-100597641 on Build GRCh38
rs765377253 Chr.X: 100597620-100597620 on Build GRCh38
rs747423279 Chr.X: 100597494-100597494 on Build GRCh38
rs140614111 Chr.X: 100597598-100597598 on Build GRCh38
rs768062613 Chr.X: 100597528-100597528 on Build GRCh38
rs902283402 Chr.X: 100597667-100597667 on Build GRCh38
rs750312969 Chr.X: 100597575-100597575 on Build GRCh38
rs747715772 Chr.X: 100597749-100597749 on Build GRCh38
rs766097519 Chr.X: 100597577-100597577 on Build GRCh38
rs746219722 Chr.X: 100597685-100597685 on Build GRCh38
rs942625155 Chr.X: 100597376-100597376 on Build GRCh38
rs778443610 Chr.X: 100597636-100597636 on Build GRCh38
rs942962203 Chr.X: 100597472-100597472 on Build GRCh38
rs768421798 Chr.X: 100597705-100597705 on Build GRCh38
rs760118309 Chr.X: 100597693-100597693 on Build GRCh38
rs190332194 Chr.X: 100597623-100597623 on Build GRCh38
rs776583016 Chr.X: 100597707-100597707 on Build GRCh38
rs951545143 Chr.X: 100597757-100597757 on Build GRCh38
rs960607999 Chr.X: 100597837-100597837 on Build GRCh38
rs771369045 Chr.X: 100597501-100597501 on Build GRCh38
rs747476447 Chr.X: 100597645-100597645 on Build GRCh38
rs775309826 Chr.X: 100597518-100597518 on Build GRCh38
rs370976073 Chr.X: 100597481-100597481 on Build GRCh38
rs1039562328 Chr.X: 100597822-100597822 on Build GRCh38
rs999153564 Chr.X: 100597730-100597730 on Build GRCh38
rs113058075 Chr.X: 100597816-100597817 on Build GRCh38
rs536993921 Chr.X: 100597457-100597457 on Build GRCh38
rs781640897 Chr.X: 100597680-100597680 on Build GRCh38
rs760496189 Chr.X: 100597576-100597576 on Build GRCh38
rs772706571 Chr.X: 100597479-100597479 on Build GRCh38
rs748166762 Chr.X: 100597462-100597462 on Build GRCh38
rs760570279 Chr.X: 100597552-100597552 on Build GRCh38
rs763597004 Chr.X: 100597758-100597758 on Build GRCh38
rs1038730495 Chr.X: 100597535-100597535 on Build GRCh38
rs773967502 Chr.X: 100597549-100597549 on Build GRCh38
rs112898376 Chr.X: 100597412-100597412 on Build GRCh38

View Full Product Details