Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70259381-70259974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RY4
Entrez Gene ID
5030
Gene Name
pyrimidinergic receptor P2Y4
Gene Aliases
NRU, P2P, P2Y4, UNR
UniGene
Hs.673854
Gene Chromosome Location
Chr.X: 70258166-70259804 on Build GRCh38
Transcripts
NM_002565.3

SNP Detail

SNP ID Chromosome Location
rs757415130 Chr.X: 70259631-70259631 on Build GRCh38
rs748759540 Chr.X: 70259739-70259739 on Build GRCh38
rs531425052 Chr.X: 70259743-70259743 on Build GRCh38
rs953845182 Chr.X: 70259770-70259770 on Build GRCh38
rs201387920 Chr.X: 70259650-70259650 on Build GRCh38
rs745586529 Chr.X: 70259628-70259628 on Build GRCh38
rs757546796 Chr.X: 70259467-70259467 on Build GRCh38
rs974816656 Chr.X: 70259708-70259708 on Build GRCh38
rs1036224801 Chr.X: 70259878-70259878 on Build GRCh38
rs202151113 Chr.X: 70259428-70259428 on Build GRCh38
rs141499158 Chr.X: 70259472-70259472 on Build GRCh38
rs912479924 Chr.X: 70259852-70259853 on Build GRCh38
rs201294501 Chr.X: 70259585-70259585 on Build GRCh38
rs768085584 Chr.X: 70259756-70259756 on Build GRCh38
rs753718610 Chr.X: 70259416-70259416 on Build GRCh38
rs375799352 Chr.X: 70259706-70259706 on Build GRCh38
rs765017183 Chr.X: 70259588-70259588 on Build GRCh38
rs776082448 Chr.X: 70259520-70259520 on Build GRCh38
rs781633256 Chr.X: 70259578-70259578 on Build GRCh38
rs760225809 Chr.X: 70259565-70259565 on Build GRCh38
rs139178534 Chr.X: 70259566-70259566 on Build GRCh38
rs771669510 Chr.X: 70259551-70259551 on Build GRCh38
rs1011468922 Chr.X: 70259707-70259707 on Build GRCh38
rs192398503 Chr.X: 70259757-70259757 on Build GRCh38
rs754843343 Chr.X: 70259429-70259429 on Build GRCh38
rs1025171062 Chr.X: 70259611-70259611 on Build GRCh38
rs746309988 Chr.X: 70259499-70259499 on Build GRCh38
rs775134638 Chr.X: 70259675-70259675 on Build GRCh38
rs758865613 Chr.X: 70259412-70259412 on Build GRCh38
rs758402356 Chr.X: 70259438-70259438 on Build GRCh38
rs747148895 Chr.X: 70259466-70259466 on Build GRCh38
rs945093931 Chr.X: 70259913-70259913 on Build GRCh38
rs772873619 Chr.X: 70259558-70259558 on Build GRCh38
rs998911054 Chr.X: 70259796-70259796 on Build GRCh38
rs781534707 Chr.X: 70259639-70259639 on Build GRCh38
rs916198667 Chr.X: 70259478-70259478 on Build GRCh38
rs146193354 Chr.X: 70259524-70259524 on Build GRCh38
rs766042959 Chr.X: 70259415-70259415 on Build GRCh38
rs371902337 Chr.X: 70259523-70259523 on Build GRCh38
rs760408937 Chr.X: 70259410-70259410 on Build GRCh38
rs986739390 Chr.X: 70259787-70259787 on Build GRCh38
rs769303347 Chr.X: 70259645-70259645 on Build GRCh38
rs886467196 Chr.X: 70259624-70259624 on Build GRCh38
rs752641496 Chr.X: 70259437-70259437 on Build GRCh38
rs919444236 Chr.X: 70259552-70259552 on Build GRCh38
rs758352422 Chr.X: 70259605-70259605 on Build GRCh38
rs772935339 Chr.X: 70259407-70259407 on Build GRCh38
rs771441510 Chr.X: 70259927-70259927 on Build GRCh38
rs745423223 Chr.X: 70259862-70259862 on Build GRCh38
rs960061038 Chr.X: 70259894-70259894 on Build GRCh38
rs752588394 Chr.X: 70259601-70259601 on Build GRCh38
rs746092057 Chr.X: 70259640-70259640 on Build GRCh38
rs770410425 Chr.X: 70259515-70259515 on Build GRCh38
rs764135040 Chr.X: 70259627-70259627 on Build GRCh38
rs765229817 Chr.X: 70259431-70259431 on Build GRCh38
rs764012588 Chr.X: 70259633-70259633 on Build GRCh38
rs991926575 Chr.X: 70259925-70259925 on Build GRCh38
rs138327932 Chr.X: 70259444-70259444 on Build GRCh38

View Full Product Details