Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75871895-75872490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPEF2
Entrez Gene ID
5470
Gene Name
protein phosphatase with EF-hand domain 2
Gene Aliases
PPP7CB
UniGene
Hs.290873
Gene Chromosome Location
Chr.4: 75859872-75902528 on Build GRCh38
Transcripts
NM_006239.2 XM_011532039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151923 c.1600G>A p.V534M Chr.4: 75872074-75872074 on Build GRCh38
4159185 c.1558G>T p.G520W Chr.4: 75872116-75872116 on Build GRCh38
5401555 c.1638C>T p.T546T Chr.4: 75872036-75872036 on Build GRCh38
1057586 c.1548C>T p.G516G Chr.4: 75872126-75872126 on Build GRCh38
4880523 c.1583C>A p.A528D Chr.4: 75872091-75872091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914608456 Chr.4: 75872290-75872291 on Build GRCh38
rs376588577 Chr.4: 75872136-75872136 on Build GRCh38
rs781482501 Chr.4: 75872091-75872091 on Build GRCh38
rs775420243 Chr.4: 75872298-75872299 on Build GRCh38
rs746158105 Chr.4: 75871996-75871996 on Build GRCh38
rs746471265 Chr.4: 75872194-75872194 on Build GRCh38
rs773089610 Chr.4: 75872163-75872163 on Build GRCh38
rs747603998 Chr.4: 75871979-75871979 on Build GRCh38
rs905783010 Chr.4: 75871930-75871930 on Build GRCh38
rs995697736 Chr.4: 75872449-75872449 on Build GRCh38
rs770766160 Chr.4: 75872167-75872167 on Build GRCh38
rs950140862 Chr.4: 75872289-75872289 on Build GRCh38
rs568226988 Chr.4: 75872448-75872448 on Build GRCh38
rs779453312 Chr.4: 75872066-75872067 on Build GRCh38
rs778586794 Chr.4: 75872141-75872141 on Build GRCh38
rs1048990812 Chr.4: 75872354-75872354 on Build GRCh38
rs560991167 Chr.4: 75871920-75871920 on Build GRCh38
rs375466728 Chr.4: 75872271-75872271 on Build GRCh38
rs142351049 Chr.4: 75871927-75871927 on Build GRCh38
rs373861132 Chr.4: 75872186-75872186 on Build GRCh38
rs954209 Chr.4: 75872005-75872005 on Build GRCh38
rs769488980 Chr.4: 75872009-75872009 on Build GRCh38
rs1001481405 Chr.4: 75872337-75872337 on Build GRCh38
rs1014674599 Chr.4: 75872031-75872031 on Build GRCh38
rs936162690 Chr.4: 75872223-75872223 on Build GRCh38
rs772053083 Chr.4: 75872156-75872156 on Build GRCh38
rs151263751 Chr.4: 75872429-75872429 on Build GRCh38
rs201392251 Chr.4: 75872164-75872164 on Build GRCh38
rs956339791 Chr.4: 75872221-75872221 on Build GRCh38
rs544413220 Chr.4: 75872018-75872018 on Build GRCh38
rs1028629175 Chr.4: 75872390-75872390 on Build GRCh38
rs560189375 Chr.4: 75872244-75872244 on Build GRCh38
rs951845906 Chr.4: 75871973-75871973 on Build GRCh38
rs370391289 Chr.4: 75872379-75872379 on Build GRCh38
rs1004712387 Chr.4: 75872365-75872365 on Build GRCh38
rs151082105 Chr.4: 75872042-75872042 on Build GRCh38
rs776382922 Chr.4: 75872170-75872170 on Build GRCh38
rs758317524 Chr.4: 75871988-75871988 on Build GRCh38
rs905837190 Chr.4: 75872328-75872328 on Build GRCh38
rs780540735 Chr.4: 75872115-75872115 on Build GRCh38
rs779446288 Chr.4: 75872147-75872147 on Build GRCh38
rs756601730 Chr.4: 75872113-75872113 on Build GRCh38
rs997651882 Chr.4: 75872389-75872389 on Build GRCh38
rs773968098 Chr.4: 75872024-75872024 on Build GRCh38
rs186321394 Chr.4: 75872155-75872155 on Build GRCh38
rs181358727 Chr.4: 75871954-75871954 on Build GRCh38
rs748988869 Chr.4: 75872135-75872135 on Build GRCh38
rs201202030 Chr.4: 75872064-75872064 on Build GRCh38
rs932444119 Chr.4: 75872348-75872348 on Build GRCh38
rs1036693279 Chr.4: 75872423-75872423 on Build GRCh38
rs898716418 Chr.4: 75871980-75871980 on Build GRCh38
rs750935388 Chr.4: 75872100-75872100 on Build GRCh38
rs764255505 Chr.4: 75872188-75872188 on Build GRCh38
rs752960549 Chr.4: 75871958-75871958 on Build GRCh38
rs772466896 Chr.4: 75872291-75872298 on Build GRCh38
rs763143738 Chr.4: 75872074-75872074 on Build GRCh38
rs766158650 Chr.4: 75872059-75872059 on Build GRCh38
rs1015136095 Chr.4: 75872378-75872378 on Build GRCh38
rs751660553 Chr.4: 75872211-75872211 on Build GRCh38
rs552279420 Chr.4: 75872282-75872282 on Build GRCh38
rs760322160 Chr.4: 75872043-75872043 on Build GRCh38
rs771854329 Chr.4: 75872036-75872036 on Build GRCh38
rs1024279498 Chr.4: 75871928-75871928 on Build GRCh38
rs760461880 Chr.4: 75871984-75871984 on Build GRCh38
rs939772807 Chr.4: 75872401-75872401 on Build GRCh38
rs570733593 Chr.4: 75872310-75872310 on Build GRCh38
rs757808446 Chr.4: 75872079-75872079 on Build GRCh38
rs913325028 Chr.4: 75872288-75872288 on Build GRCh38
rs764105291 Chr.4: 75871986-75871986 on Build GRCh38
rs1001431325 Chr.4: 75871949-75871949 on Build GRCh38
rs1394916 Chr.4: 75872075-75872075 on Build GRCh38
rs376120897 Chr.4: 75872109-75872109 on Build GRCh38
rs985697506 Chr.4: 75872450-75872450 on Build GRCh38
rs779075316 Chr.4: 75872291-75872294 on Build GRCh38
rs369831198 Chr.4: 75872139-75872139 on Build GRCh38
rs374184516 Chr.4: 75872035-75872035 on Build GRCh38
rs201465969 Chr.4: 75871987-75871988 on Build GRCh38
rs781646011 Chr.4: 75871987-75871987 on Build GRCh38
rs372081041 Chr.4: 75872077-75872077 on Build GRCh38
rs887647112 Chr.4: 75872363-75872363 on Build GRCh38

View Full Product Details