Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34606421-34607001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf106
Entrez Gene ID
64771
Gene Name
chromosome 6 open reading frame 106
Gene Aliases
FP852, dJ391O22.4
UniGene
Hs.643498
Gene Chromosome Location
Chr.6: 34587280-34696850 on Build GRCh38
Transcripts
NM_022758.5 NM_024294.3 XM_005249298.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
742266 c.754C>T p.P252S Chr.6: 34606662-34606662 on Build GRCh38
109285 c.699C>T p.S233S Chr.6: 34606717-34606717 on Build GRCh38
5870445 c.818C>T p.P273L Chr.6: 34606598-34606598 on Build GRCh38
7286539 c.689C>A p.P230H Chr.6: 34606727-34606727 on Build GRCh38
1621552 c.809A>G p.N270S Chr.6: 34606607-34606607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377504022 Chr.6: 34606918-34606918 on Build GRCh38
rs759751988 Chr.6: 34606650-34606651 on Build GRCh38
rs893665544 Chr.6: 34606689-34606689 on Build GRCh38
rs765705386 Chr.6: 34606919-34606920 on Build GRCh38
rs368491252 Chr.6: 34606829-34606829 on Build GRCh38
rs747918125 Chr.6: 34606683-34606683 on Build GRCh38
rs943712259 Chr.6: 34606455-34606455 on Build GRCh38
rs750056993 Chr.6: 34606716-34606716 on Build GRCh38
rs749306924 Chr.6: 34606542-34606542 on Build GRCh38
rs548568403 Chr.6: 34606975-34606975 on Build GRCh38
rs768763183 Chr.6: 34606932-34606932 on Build GRCh38
rs773437919 Chr.6: 34606523-34606523 on Build GRCh38
rs771227849 Chr.6: 34606579-34606579 on Build GRCh38
rs79514997 Chr.6: 34606524-34606524 on Build GRCh38
rs764543291 Chr.6: 34606604-34606604 on Build GRCh38
rs762160757 Chr.6: 34606707-34606707 on Build GRCh38
rs751143477 Chr.6: 34606504-34606504 on Build GRCh38
rs777005775 Chr.6: 34606580-34606580 on Build GRCh38
rs747303458 Chr.6: 34606944-34606944 on Build GRCh38
rs749151218 Chr.6: 34606669-34606669 on Build GRCh38
rs375098975 Chr.6: 34606863-34606863 on Build GRCh38
rs936404154 Chr.6: 34606470-34606470 on Build GRCh38
rs772397191 Chr.6: 34606805-34606805 on Build GRCh38
rs146730430 Chr.6: 34606588-34606588 on Build GRCh38
rs113840191 Chr.6: 34606481-34606481 on Build GRCh38
rs1054037716 Chr.6: 34606518-34606518 on Build GRCh38
rs747914055 Chr.6: 34606509-34606510 on Build GRCh38
rs765476917 Chr.6: 34606594-34606594 on Build GRCh38
rs757579186 Chr.6: 34606628-34606628 on Build GRCh38
rs149349132 Chr.6: 34606874-34606874 on Build GRCh38
rs145682744 Chr.6: 34606882-34606882 on Build GRCh38
rs774911852 Chr.6: 34606765-34606765 on Build GRCh38
rs745328279 Chr.6: 34606513-34606513 on Build GRCh38
rs148908492 Chr.6: 34606734-34606734 on Build GRCh38
rs751344052 Chr.6: 34606912-34606912 on Build GRCh38
rs369149849 Chr.6: 34606706-34606706 on Build GRCh38
rs1039366289 Chr.6: 34606456-34606456 on Build GRCh38
rs920799409 Chr.6: 34606465-34606465 on Build GRCh38
rs370439242 Chr.6: 34606935-34606935 on Build GRCh38
rs149675520 Chr.6: 34606646-34606646 on Build GRCh38
rs763801825 Chr.6: 34606651-34606671 on Build GRCh38
rs766048503 Chr.6: 34606721-34606721 on Build GRCh38
rs200230008 Chr.6: 34606671-34606671 on Build GRCh38
rs373122080 Chr.6: 34606728-34606728 on Build GRCh38
rs771263498 Chr.6: 34606945-34606945 on Build GRCh38
rs772140476 Chr.6: 34606653-34606653 on Build GRCh38
rs754458960 Chr.6: 34606724-34606724 on Build GRCh38
rs746147397 Chr.6: 34606692-34606692 on Build GRCh38
rs1046197069 Chr.6: 34606969-34606969 on Build GRCh38
rs769969729 Chr.6: 34606699-34606699 on Build GRCh38
rs146278037 Chr.6: 34606717-34606717 on Build GRCh38
rs774265101 Chr.6: 34606631-34606631 on Build GRCh38
rs761011110 Chr.6: 34606569-34606570 on Build GRCh38
rs139342047 Chr.6: 34606596-34606596 on Build GRCh38
rs199754841 Chr.6: 34606687-34606687 on Build GRCh38
rs777757738 Chr.6: 34606744-34606744 on Build GRCh38
rs151076011 Chr.6: 34606711-34606711 on Build GRCh38
rs758250953 Chr.6: 34606737-34606737 on Build GRCh38
rs1008614618 Chr.6: 34606799-34606799 on Build GRCh38
rs545603862 Chr.6: 34606751-34606751 on Build GRCh38
rs574960451 Chr.6: 34606957-34606958 on Build GRCh38
rs753338316 Chr.6: 34606723-34606723 on Build GRCh38
rs374964078 Chr.6: 34606702-34606702 on Build GRCh38
rs961703039 Chr.6: 34606813-34606813 on Build GRCh38
rs144085981 Chr.6: 34606705-34606705 on Build GRCh38
rs936421643 Chr.6: 34606916-34606916 on Build GRCh38
rs778902606 Chr.6: 34606934-34606934 on Build GRCh38
rs778736991 Chr.6: 34606677-34606677 on Build GRCh38
rs74903498 Chr.6: 34606757-34606757 on Build GRCh38
rs746054761 Chr.6: 34606747-34606747 on Build GRCh38
rs765996037 Chr.6: 34606811-34606811 on Build GRCh38
rs751906052 Chr.6: 34606625-34606625 on Build GRCh38
rs950146141 Chr.6: 34606966-34606966 on Build GRCh38
rs753433730 Chr.6: 34606828-34606828 on Build GRCh38
rs372401388 Chr.6: 34606511-34606511 on Build GRCh38
rs746009090 Chr.6: 34606639-34606639 on Build GRCh38
rs200981086 Chr.6: 34606760-34606760 on Build GRCh38
rs571387827 Chr.6: 34606466-34606466 on Build GRCh38
rs779590612 Chr.6: 34606651-34606651 on Build GRCh38
rs117366766 Chr.6: 34606528-34606528 on Build GRCh38
rs573391171 Chr.6: 34606560-34606560 on Build GRCh38
rs771996552 Chr.6: 34606684-34606684 on Build GRCh38
rs762359158 Chr.6: 34606795-34606795 on Build GRCh38
rs201498474 Chr.6: 34606568-34606568 on Build GRCh38
rs376293203 Chr.6: 34606810-34606810 on Build GRCh38
rs371793801 Chr.6: 34606872-34606872 on Build GRCh38
rs950115187 Chr.6: 34606546-34606546 on Build GRCh38
rs760398267 Chr.6: 34606784-34606784 on Build GRCh38
rs780156128 Chr.6: 34606753-34606753 on Build GRCh38
rs147879404 Chr.6: 34606718-34606718 on Build GRCh38
rs756897665 Chr.6: 34606510-34606510 on Build GRCh38
rs749535340 Chr.6: 34606926-34606926 on Build GRCh38
rs771780698 Chr.6: 34606517-34606517 on Build GRCh38
rs776475035 Chr.6: 34606943-34606943 on Build GRCh38
rs188849106 Chr.6: 34606700-34606700 on Build GRCh38
rs771325811 Chr.6: 34606580-34606583 on Build GRCh38
rs777878238 Chr.6: 34606903-34606903 on Build GRCh38
rs145448520 Chr.6: 34606685-34606685 on Build GRCh38
rs202096693 Chr.6: 34606561-34606561 on Build GRCh38
rs763561912 Chr.6: 34606599-34606599 on Build GRCh38

View Full Product Details