Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41930011-41930610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BYSL
Entrez Gene ID
705
Gene Name
bystin like
Gene Aliases
BYSTIN
UniGene
Hs.106880
Gene Chromosome Location
Chr.6: 41921227-41933046 on Build GRCh38
Transcripts
NM_004053.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741920 c.487G>C p.E163Q Chr.6: 41930187-41930187 on Build GRCh38
3627292 c.535C>T p.P179S Chr.6: 41930235-41930235 on Build GRCh38
3874046 c.510G>A p.S170S Chr.6: 41930210-41930210 on Build GRCh38
6106594 c.502G>T p.E168* Chr.6: 41930202-41930202 on Build GRCh38
3078933 c.564C>T p.V188V Chr.6: 41930264-41930264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373001397 Chr.6: 41930379-41930379 on Build GRCh38
rs757774434 Chr.6: 41930083-41930083 on Build GRCh38
rs150414779 Chr.6: 41930262-41930262 on Build GRCh38
rs1032513768 Chr.6: 41930497-41930497 on Build GRCh38
rs746015637 Chr.6: 41930093-41930093 on Build GRCh38
rs749342889 Chr.6: 41930209-41930209 on Build GRCh38
rs751926056 Chr.6: 41930157-41930157 on Build GRCh38
rs200946471 Chr.6: 41930252-41930252 on Build GRCh38
rs775113265 Chr.6: 41930261-41930261 on Build GRCh38
rs765781821 Chr.6: 41930134-41930134 on Build GRCh38
rs764938283 Chr.6: 41930258-41930258 on Build GRCh38
rs918291589 Chr.6: 41930365-41930365 on Build GRCh38
rs777741505 Chr.6: 41930588-41930588 on Build GRCh38
rs146819393 Chr.6: 41930239-41930239 on Build GRCh38
rs2479726 Chr.6: 41930358-41930358 on Build GRCh38
rs374026969 Chr.6: 41930043-41930043 on Build GRCh38
rs753100203 Chr.6: 41930234-41930234 on Build GRCh38
rs1001409058 Chr.6: 41930495-41930495 on Build GRCh38
rs757614914 Chr.6: 41930165-41930165 on Build GRCh38
rs563830254 Chr.6: 41930197-41930197 on Build GRCh38
rs964588553 Chr.6: 41930068-41930068 on Build GRCh38
rs771976157 Chr.6: 41930238-41930238 on Build GRCh38
rs768886100 Chr.6: 41930119-41930119 on Build GRCh38
rs140565706 Chr.6: 41930256-41930256 on Build GRCh38
rs767840364 Chr.6: 41930263-41930263 on Build GRCh38
rs375354481 Chr.6: 41930236-41930236 on Build GRCh38
rs1033043594 Chr.6: 41930120-41930120 on Build GRCh38
rs760288519 Chr.6: 41930133-41930133 on Build GRCh38
rs1041221678 Chr.6: 41930407-41930407 on Build GRCh38
rs890139095 Chr.6: 41930404-41930404 on Build GRCh38
rs779333015 Chr.6: 41930302-41930302 on Build GRCh38
rs759142222 Chr.6: 41930139-41930139 on Build GRCh38
rs750910730 Chr.6: 41930184-41930184 on Build GRCh38
rs182672732 Chr.6: 41930456-41930456 on Build GRCh38
rs867583845 Chr.6: 41930342-41930342 on Build GRCh38
rs755164999 Chr.6: 41930211-41930211 on Build GRCh38
rs780032351 Chr.6: 41930205-41930205 on Build GRCh38
rs138690250 Chr.6: 41930034-41930041 on Build GRCh38
rs761638483 Chr.6: 41930123-41930123 on Build GRCh38
rs1048038800 Chr.6: 41930374-41930374 on Build GRCh38
rs376820867 Chr.6: 41930143-41930143 on Build GRCh38
rs781261246 Chr.6: 41930265-41930265 on Build GRCh38
rs781257070 Chr.6: 41930266-41930266 on Build GRCh38
rs781614872 Chr.6: 41930086-41930086 on Build GRCh38
rs374629563 Chr.6: 41930324-41930336 on Build GRCh38
rs756605230 Chr.6: 41930267-41930267 on Build GRCh38
rs536897576 Chr.6: 41930526-41930526 on Build GRCh38
rs774305785 Chr.6: 41930122-41930122 on Build GRCh38
rs201698568 Chr.6: 41930320-41930332 on Build GRCh38
rs917937442 Chr.6: 41930175-41930175 on Build GRCh38
rs1007033333 Chr.6: 41930041-41930042 on Build GRCh38
rs972640662 Chr.6: 41930360-41930360 on Build GRCh38
rs143503892 Chr.6: 41930366-41930368 on Build GRCh38
rs1007301358 Chr.6: 41930434-41930434 on Build GRCh38
rs758902882 Chr.6: 41930302-41930303 on Build GRCh38
rs1018423673 Chr.6: 41930054-41930054 on Build GRCh38
rs182941010 Chr.6: 41930247-41930247 on Build GRCh38
rs753863237 Chr.6: 41930292-41930292 on Build GRCh38
rs772137406 Chr.6: 41930125-41930125 on Build GRCh38
rs749443531 Chr.6: 41930110-41930110 on Build GRCh38
rs373527407 Chr.6: 41930031-41930032 on Build GRCh38
rs185774115 Chr.6: 41930355-41930355 on Build GRCh38
rs758307999 Chr.6: 41930585-41930585 on Build GRCh38
rs899991493 Chr.6: 41930459-41930459 on Build GRCh38
rs919132255 Chr.6: 41930291-41930291 on Build GRCh38
rs768073170 Chr.6: 41930181-41930181 on Build GRCh38
rs755375386 Chr.6: 41930098-41930099 on Build GRCh38
rs769752113 Chr.6: 41930094-41930094 on Build GRCh38
rs756261056 Chr.6: 41930187-41930187 on Build GRCh38
rs751888848 Chr.6: 41930047-41930047 on Build GRCh38
rs755156077 Chr.6: 41930147-41930149 on Build GRCh38
rs779206176 Chr.6: 41930216-41930216 on Build GRCh38
rs1053125793 Chr.6: 41930570-41930570 on Build GRCh38
rs748357358 Chr.6: 41930314-41930314 on Build GRCh38
rs748347690 Chr.6: 41930275-41930275 on Build GRCh38
rs780374384 Chr.6: 41930102-41930102 on Build GRCh38
rs369710061 Chr.6: 41930293-41930293 on Build GRCh38
rs764811902 Chr.6: 41930151-41930151 on Build GRCh38
rs201331703 Chr.6: 41930307-41930307 on Build GRCh38
rs956985875 Chr.6: 41930124-41930124 on Build GRCh38
rs370150530 Chr.6: 41930551-41930551 on Build GRCh38
rs942441721 Chr.6: 41930405-41930405 on Build GRCh38
rs773275294 Chr.6: 41930131-41930131 on Build GRCh38
rs199673156 Chr.6: 41930220-41930220 on Build GRCh38
rs372324748 Chr.6: 41930321-41930321 on Build GRCh38
rs766847529 Chr.6: 41930286-41930286 on Build GRCh38
rs750764154 Chr.6: 41930264-41930264 on Build GRCh38

View Full Product Details