Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87283344-87283793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB7
Entrez Gene ID
375519
Gene Name
gap junction protein beta 7
Gene Aliases
CX25, bA136M9.1, connexin25
UniGene
Hs.146727
Gene Chromosome Location
Chr.6: 87282978-87329278 on Build GRCh38
Transcripts
NM_198568.2 XM_005248716.2

SNP Detail

SNP ID Chromosome Location
rs1042887323 Chr.6: 87283398-87283398 on Build GRCh38
rs1052498249 Chr.6: 87283697-87283697 on Build GRCh38
rs748418871 Chr.6: 87283571-87283571 on Build GRCh38
rs892476916 Chr.6: 87283440-87283440 on Build GRCh38
rs934457899 Chr.6: 87283689-87283689 on Build GRCh38
rs1007815014 Chr.6: 87283757-87283757 on Build GRCh38
rs1021063834 Chr.6: 87283527-87283527 on Build GRCh38
rs904321378 Chr.6: 87283419-87283419 on Build GRCh38
rs879892958 Chr.6: 87283645-87283645 on Build GRCh38
rs770972097 Chr.6: 87283383-87283383 on Build GRCh38
rs933361592 Chr.6: 87283720-87283720 on Build GRCh38
rs370812734 Chr.6: 87283540-87283540 on Build GRCh38
rs953629993 Chr.6: 87283529-87283529 on Build GRCh38
rs940106204 Chr.6: 87283681-87283681 on Build GRCh38
rs759117243 Chr.6: 87283583-87283583 on Build GRCh38
rs187461820 Chr.6: 87283638-87283638 on Build GRCh38
rs1041098503 Chr.6: 87283703-87283703 on Build GRCh38
rs773139798 Chr.6: 87283367-87283367 on Build GRCh38
rs1028721777 Chr.6: 87283766-87283766 on Build GRCh38
rs1051709205 Chr.6: 87283729-87283729 on Build GRCh38
rs925827435 Chr.6: 87283655-87283655 on Build GRCh38
rs773776481 Chr.6: 87283424-87283424 on Build GRCh38
rs1016905633 Chr.6: 87283765-87283765 on Build GRCh38
rs558266101 Chr.6: 87283546-87283546 on Build GRCh38
rs555955431 Chr.6: 87283636-87283636 on Build GRCh38
rs146029475 Chr.6: 87283584-87283584 on Build GRCh38
rs2240461 Chr.6: 87283372-87283372 on Build GRCh38
rs559150202 Chr.6: 87283665-87283665 on Build GRCh38
rs550703777 Chr.6: 87283470-87283470 on Build GRCh38
rs1000960012 Chr.6: 87283420-87283420 on Build GRCh38
rs889082986 Chr.6: 87283750-87283750 on Build GRCh38
rs139313778 Chr.6: 87283438-87283438 on Build GRCh38
rs927621944 Chr.6: 87283555-87283555 on Build GRCh38
rs959724885 Chr.6: 87283594-87283594 on Build GRCh38
rs142617101 Chr.6: 87283534-87283534 on Build GRCh38
rs374132464 Chr.6: 87283379-87283379 on Build GRCh38
rs376133277 Chr.6: 87283649-87283649 on Build GRCh38
rs997457543 Chr.6: 87283453-87283453 on Build GRCh38
rs113036753 Chr.6: 87283634-87283634 on Build GRCh38
rs979046942 Chr.6: 87283654-87283654 on Build GRCh38
rs1052444948 Chr.6: 87283436-87283436 on Build GRCh38
rs544890606 Chr.6: 87283718-87283718 on Build GRCh38
rs566831138 Chr.6: 87283547-87283547 on Build GRCh38

View Full Product Details