Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98117629-98118227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POP1
Entrez Gene ID
10940
Gene Name
POP1 homolog, ribonuclease P/MRP subunit
Gene Aliases
-
UniGene
Hs.252828
Gene Chromosome Location
Chr.8: 98117293-98159841 on Build GRCh38
Transcripts
NM_001145860.1 NM_001145861.1 NM_015029.2 XM_011516801.2

SNP Detail

SNP ID Chromosome Location
rs979190568 Chr.8: 98118204-98118204 on Build GRCh38
rs372101629 Chr.8: 98117792-98117795 on Build GRCh38
rs538910081 Chr.8: 98117977-98117978 on Build GRCh38
rs1016898193 Chr.8: 98117724-98117724 on Build GRCh38
rs202120933 Chr.8: 98118045-98118046 on Build GRCh38
rs187402660 Chr.8: 98117736-98117736 on Build GRCh38
rs535271844 Chr.8: 98117651-98117651 on Build GRCh38
rs1045292922 Chr.8: 98118019-98118019 on Build GRCh38
rs150466709 Chr.8: 98118165-98118165 on Build GRCh38
rs569953391 Chr.8: 98117706-98117706 on Build GRCh38
rs565839708 Chr.8: 98118127-98118127 on Build GRCh38
rs113320288 Chr.8: 98117789-98117792 on Build GRCh38
rs1017979989 Chr.8: 98118175-98118175 on Build GRCh38
rs760059326 Chr.8: 98117881-98117881 on Build GRCh38
rs553660211 Chr.8: 98117690-98117690 on Build GRCh38
rs1000344019 Chr.8: 98118097-98118097 on Build GRCh38
rs891366163 Chr.8: 98118152-98118152 on Build GRCh38
rs117361122 Chr.8: 98117927-98117927 on Build GRCh38
rs985105424 Chr.8: 98117769-98117769 on Build GRCh38
rs528415634 Chr.8: 98118129-98118129 on Build GRCh38
rs1014745239 Chr.8: 98117663-98117663 on Build GRCh38
rs182800776 Chr.8: 98117698-98117698 on Build GRCh38
rs558670727 Chr.8: 98117836-98117836 on Build GRCh38
rs58749995 Chr.8: 98118035-98118035 on Build GRCh38
rs1018448662 Chr.8: 98117792-98117792 on Build GRCh38
rs76697107 Chr.8: 98117988-98117988 on Build GRCh38
rs962645998 Chr.8: 98117745-98117745 on Build GRCh38
rs57047356 Chr.8: 98118137-98118137 on Build GRCh38
rs556700206 Chr.8: 98118203-98118203 on Build GRCh38
rs1001575364 Chr.8: 98117721-98117721 on Build GRCh38
rs529685464 Chr.8: 98117678-98117678 on Build GRCh38
rs950187815 Chr.8: 98118049-98118049 on Build GRCh38
rs1041813722 Chr.8: 98118055-98118055 on Build GRCh38
rs1052330258 Chr.8: 98117662-98117662 on Build GRCh38
rs904989877 Chr.8: 98118038-98118038 on Build GRCh38
rs574652394 Chr.8: 98117976-98117976 on Build GRCh38
rs1018862816 Chr.8: 98118184-98118184 on Build GRCh38
rs545820700 Chr.8: 98118090-98118090 on Build GRCh38
rs979167049 Chr.8: 98117877-98117877 on Build GRCh38
rs909758979 Chr.8: 98117973-98117973 on Build GRCh38
rs988086246 Chr.8: 98117956-98117956 on Build GRCh38
rs774679015 Chr.8: 98117746-98117746 on Build GRCh38
rs74951928 Chr.8: 98118046-98118046 on Build GRCh38
rs976452714 Chr.8: 98117885-98117885 on Build GRCh38
rs554758409 Chr.8: 98117948-98117948 on Build GRCh38
rs964221284 Chr.8: 98118192-98118192 on Build GRCh38
rs868622275 Chr.8: 98117870-98117870 on Build GRCh38
rs142126341 Chr.8: 98117670-98117670 on Build GRCh38
rs192260633 Chr.8: 98118088-98118088 on Build GRCh38
rs866074613 Chr.8: 98117901-98117901 on Build GRCh38
rs937346145 Chr.8: 98117980-98117980 on Build GRCh38
rs572439064 Chr.8: 98117868-98117868 on Build GRCh38
rs796944928 Chr.8: 98118045-98118046 on Build GRCh38
rs766986865 Chr.8: 98117891-98117891 on Build GRCh38
rs987374324 Chr.8: 98117827-98117827 on Build GRCh38
rs903366533 Chr.8: 98118082-98118082 on Build GRCh38
rs757651998 Chr.8: 98117788-98117789 on Build GRCh38
rs905940677 Chr.8: 98117711-98117711 on Build GRCh38
rs965191968 Chr.8: 98117866-98117866 on Build GRCh38
rs760470855 Chr.8: 98117943-98117943 on Build GRCh38
rs559066695 Chr.8: 98118128-98118128 on Build GRCh38
rs1033265467 Chr.8: 98118126-98118126 on Build GRCh38
rs5893455 Chr.8: 98118046-98118046 on Build GRCh38
rs964143921 Chr.8: 98117847-98117847 on Build GRCh38
rs763384974 Chr.8: 98117692-98117692 on Build GRCh38
rs376451024 Chr.8: 98118080-98118080 on Build GRCh38

View Full Product Details