Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101676228-101676688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW4
Entrez Gene ID
6468
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
DAC, FBW4, FBWD4, SHFM3, SHSF3
UniGene
Hs.500822
Gene Chromosome Location
Chr.10: 101610664-101695295 on Build GRCh38
Transcripts
NM_001323541.1 NM_022039.3 NR_136613.1 XM_005270053.2 XM_017016570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011145 c.291G>A p.V97V Chr.10: 101676406-101676406 on Build GRCh38
4684081 c.286C>T p.R96* Chr.10: 101676411-101676411 on Build GRCh38
4937322 c.310A>G p.R104G Chr.10: 101676387-101676387 on Build GRCh38
6662448 c.326G>A p.R109Q Chr.10: 101676371-101676371 on Build GRCh38
1602886 c.261-10C>G p.? Chr.10: 101676446-101676446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748178921 Chr.10: 101676478-101676478 on Build GRCh38
rs936746492 Chr.10: 101676512-101676512 on Build GRCh38
rs916578155 Chr.10: 101676587-101676587 on Build GRCh38
rs760956768 Chr.10: 101676388-101676388 on Build GRCh38
rs772128782 Chr.10: 101676480-101676480 on Build GRCh38
rs1006587631 Chr.10: 101676391-101676391 on Build GRCh38
rs774229163 Chr.10: 101676471-101676471 on Build GRCh38
rs768762737 Chr.10: 101676345-101676345 on Build GRCh38
rs963779441 Chr.10: 101676547-101676547 on Build GRCh38
rs774390490 Chr.10: 101676371-101676371 on Build GRCh38
rs749731095 Chr.10: 101676294-101676294 on Build GRCh38
rs765792909 Chr.10: 101676411-101676411 on Build GRCh38
rs780858814 Chr.10: 101676287-101676287 on Build GRCh38
rs925401129 Chr.10: 101676643-101676643 on Build GRCh38
rs555859926 Chr.10: 101676265-101676265 on Build GRCh38
rs11191082 Chr.10: 101676386-101676386 on Build GRCh38
rs773359787 Chr.10: 101676488-101676488 on Build GRCh38
rs766850913 Chr.10: 101676395-101676395 on Build GRCh38
rs533828971 Chr.10: 101676638-101676638 on Build GRCh38
rs892436866 Chr.10: 101676276-101676277 on Build GRCh38
rs56866043 Chr.10: 101676394-101676394 on Build GRCh38
rs11191084 Chr.10: 101676662-101676662 on Build GRCh38
rs755969920 Chr.10: 101676448-101676448 on Build GRCh38
rs755522422 Chr.10: 101676410-101676410 on Build GRCh38
rs749218179 Chr.10: 101676313-101676313 on Build GRCh38
rs761901213 Chr.10: 101676372-101676372 on Build GRCh38
rs149431155 Chr.10: 101676618-101676618 on Build GRCh38
rs187669687 Chr.10: 101676461-101676461 on Build GRCh38
rs149070050 Chr.10: 101676377-101676377 on Build GRCh38
rs749271242 Chr.10: 101676463-101676463 on Build GRCh38
rs768601295 Chr.10: 101676466-101676466 on Build GRCh38
rs751732677 Chr.10: 101676659-101676659 on Build GRCh38
rs949640532 Chr.10: 101676588-101676588 on Build GRCh38
rs1013575776 Chr.10: 101676323-101676323 on Build GRCh38
rs1046611895 Chr.10: 101676666-101676666 on Build GRCh38
rs529802357 Chr.10: 101676420-101676420 on Build GRCh38
rs569273181 Chr.10: 101676414-101676414 on Build GRCh38
rs769574767 Chr.10: 101676296-101676296 on Build GRCh38
rs773413422 Chr.10: 101676378-101676378 on Build GRCh38
rs143070839 Chr.10: 101676407-101676407 on Build GRCh38
rs745521092 Chr.10: 101676441-101676441 on Build GRCh38
rs1055083865 Chr.10: 101676585-101676585 on Build GRCh38
rs377638888 Chr.10: 101676464-101676464 on Build GRCh38
rs913064612 Chr.10: 101676453-101676453 on Build GRCh38

View Full Product Details