Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119015454-119015980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
UniGene
Hs.534613
Gene Chromosome Location
Chr.11: 118998067-119015793 on Build GRCh38
Transcripts
NM_198489.2 NR_104049.1 NR_104050.1 NR_104051.1 XM_011542799.1 XM_011542800.2 XM_017017652.1 NM_001028.2

Gene Symbol
RPS25
Entrez Gene ID
6230
Gene Name
ribosomal protein S25
Gene Aliases
S25
UniGene
Hs.512676
Gene Chromosome Location
Chr.11: 119015712-119018347 on Build GRCh38
Transcripts
NM_198489.2 NR_104049.1 NR_104050.1 NR_104051.1 XM_011542799.1 XM_011542800.2 XM_017017652.1 NM_001028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769409 c.284-4delC p.? Chr.11: 119015943-119015943 on Build GRCh38
7310837 c.338C>T p.T113I Chr.11: 119015885-119015885 on Build GRCh38
4453741 c.341A>C p.K114T Chr.11: 119015882-119015882 on Build GRCh38
4722684 c.365G>A p.G122D Chr.11: 119015858-119015858 on Build GRCh38
923695 c.352G>A p.A118T Chr.11: 119015871-119015871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371801166 Chr.11: 119015884-119015884 on Build GRCh38
rs1050888930 Chr.11: 119015711-119015711 on Build GRCh38
rs782367365 Chr.11: 119015591-119015591 on Build GRCh38
rs782658942 Chr.11: 119015632-119015632 on Build GRCh38
rs782061640 Chr.11: 119015601-119015601 on Build GRCh38
rs782685783 Chr.11: 119015606-119015606 on Build GRCh38
rs782804672 Chr.11: 119015851-119015851 on Build GRCh38
rs964250237 Chr.11: 119015653-119015653 on Build GRCh38
rs782359276 Chr.11: 119015943-119015943 on Build GRCh38
rs1000121364 Chr.11: 119015770-119015770 on Build GRCh38
rs781968974 Chr.11: 119015953-119015953 on Build GRCh38
rs782258873 Chr.11: 119015636-119015636 on Build GRCh38
rs920434572 Chr.11: 119015805-119015805 on Build GRCh38
rs377257030 Chr.11: 119015801-119015801 on Build GRCh38
rs781868957 Chr.11: 119015541-119015545 on Build GRCh38
rs375501027 Chr.11: 119015942-119015942 on Build GRCh38
rs782014341 Chr.11: 119015521-119015529 on Build GRCh38
rs781897531 Chr.11: 119015608-119015608 on Build GRCh38
rs753517323 Chr.11: 119015715-119015725 on Build GRCh38
rs782403790 Chr.11: 119015806-119015806 on Build GRCh38
rs782223204 Chr.11: 119015568-119015568 on Build GRCh38
rs781934290 Chr.11: 119015572-119015572 on Build GRCh38
rs200262306 Chr.11: 119015561-119015561 on Build GRCh38
rs781858092 Chr.11: 119015620-119015620 on Build GRCh38
rs1019857705 Chr.11: 119015615-119015615 on Build GRCh38
rs577898720 Chr.11: 119015700-119015705 on Build GRCh38
rs183037053 Chr.11: 119015870-119015870 on Build GRCh38
rs782043528 Chr.11: 119015512-119015512 on Build GRCh38
rs782677187 Chr.11: 119015557-119015557 on Build GRCh38
rs782454550 Chr.11: 119015579-119015580 on Build GRCh38
rs76904653 Chr.11: 119015691-119015691 on Build GRCh38
rs201052753 Chr.11: 119015516-119015516 on Build GRCh38
rs559515271 Chr.11: 119015783-119015783 on Build GRCh38
rs7484157 Chr.11: 119015887-119015887 on Build GRCh38
rs781793748 Chr.11: 119015625-119015625 on Build GRCh38
rs782531464 Chr.11: 119015547-119015548 on Build GRCh38
rs782694430 Chr.11: 119015568-119015570 on Build GRCh38
rs782651177 Chr.11: 119015911-119015911 on Build GRCh38
rs187605382 Chr.11: 119015914-119015914 on Build GRCh38
rs1054402817 Chr.11: 119015772-119015772 on Build GRCh38
rs142393808 Chr.11: 119015848-119015848 on Build GRCh38
rs782745378 Chr.11: 119015818-119015818 on Build GRCh38
rs200333473 Chr.11: 119015944-119015944 on Build GRCh38
rs537697599 Chr.11: 119015722-119015722 on Build GRCh38
rs375684853 Chr.11: 119015597-119015597 on Build GRCh38
rs7131534 Chr.11: 119015946-119015946 on Build GRCh38
rs369100949 Chr.11: 119015513-119015513 on Build GRCh38
rs186379534 Chr.11: 119015569-119015569 on Build GRCh38
rs782209141 Chr.11: 119015815-119015816 on Build GRCh38
rs962333617 Chr.11: 119015689-119015689 on Build GRCh38
rs782221471 Chr.11: 119015646-119015646 on Build GRCh38
rs782566323 Chr.11: 119015628-119015628 on Build GRCh38
rs944929793 Chr.11: 119015736-119015736 on Build GRCh38
rs782013262 Chr.11: 119015588-119015588 on Build GRCh38
rs533907143 Chr.11: 119015666-119015666 on Build GRCh38
rs564581029 Chr.11: 119015484-119015484 on Build GRCh38
rs202084293 Chr.11: 119015527-119015527 on Build GRCh38
rs918311988 Chr.11: 119015696-119015696 on Build GRCh38
rs900768911 Chr.11: 119015762-119015763 on Build GRCh38
rs782609881 Chr.11: 119015604-119015605 on Build GRCh38
rs781926608 Chr.11: 119015803-119015804 on Build GRCh38
rs782485533 Chr.11: 119015700-119015700 on Build GRCh38
rs371246256 Chr.11: 119015828-119015828 on Build GRCh38
rs200460989 Chr.11: 119015949-119015949 on Build GRCh38
rs763460226 Chr.11: 119015665-119015665 on Build GRCh38
rs996009597 Chr.11: 119015578-119015578 on Build GRCh38
rs782779231 Chr.11: 119015641-119015641 on Build GRCh38
rs782443849 Chr.11: 119015544-119015544 on Build GRCh38
rs782741485 Chr.11: 119015737-119015737 on Build GRCh38
rs554160175 Chr.11: 119015782-119015782 on Build GRCh38
rs782212077 Chr.11: 119015954-119015954 on Build GRCh38
rs782647993 Chr.11: 119015650-119015650 on Build GRCh38
rs1021764523 Chr.11: 119015683-119015683 on Build GRCh38
rs1047631067 Chr.11: 119015850-119015850 on Build GRCh38
rs756832048 Chr.11: 119015727-119015727 on Build GRCh38
rs782623092 Chr.11: 119015642-119015642 on Build GRCh38
rs568817363 Chr.11: 119015564-119015564 on Build GRCh38
rs782748677 Chr.11: 119015537-119015537 on Build GRCh38
rs189649668 Chr.11: 119015586-119015586 on Build GRCh38
rs151116131 Chr.11: 119015651-119015652 on Build GRCh38
rs570519238 Chr.11: 119015713-119015713 on Build GRCh38
rs782780002 Chr.11: 119015611-119015611 on Build GRCh38
rs977580439 Chr.11: 119015706-119015706 on Build GRCh38
rs782239203 Chr.11: 119015612-119015613 on Build GRCh38
rs760280051 Chr.11: 119015657-119015657 on Build GRCh38
rs139203713 Chr.11: 119015648-119015649 on Build GRCh38
rs782485079 Chr.11: 119015622-119015622 on Build GRCh38
rs962634201 Chr.11: 119015881-119015881 on Build GRCh38
rs1034647326 Chr.11: 119015871-119015871 on Build GRCh38
rs782124473 Chr.11: 119015589-119015589 on Build GRCh38
rs782605404 Chr.11: 119015624-119015624 on Build GRCh38
rs782284515 Chr.11: 119015799-119015800 on Build GRCh38
rs782143753 Chr.11: 119015610-119015610 on Build GRCh38
rs782750088 Chr.11: 119015523-119015523 on Build GRCh38
rs544033859 Chr.11: 119015933-119015933 on Build GRCh38
rs1037523791 Chr.11: 119015491-119015491 on Build GRCh38
rs782482576 Chr.11: 119015528-119015528 on Build GRCh38
rs555895596 Chr.11: 119015740-119015740 on Build GRCh38
rs149825323 Chr.11: 119015893-119015893 on Build GRCh38
rs929116967 Chr.11: 119015833-119015833 on Build GRCh38
rs782367240 Chr.11: 119015951-119015951 on Build GRCh38
rs144776371 Chr.11: 119015775-119015775 on Build GRCh38
rs782116795 Chr.11: 119015817-119015817 on Build GRCh38
rs782461548 Chr.11: 119015790-119015790 on Build GRCh38
rs377583343 Chr.11: 119015539-119015539 on Build GRCh38
rs781914740 Chr.11: 119015800-119015800 on Build GRCh38
rs781962522 Chr.11: 119015476-119015476 on Build GRCh38
rs376310021 Chr.11: 119015503-119015503 on Build GRCh38
rs781792161 Chr.11: 119015576-119015577 on Build GRCh38
rs181791793 Chr.11: 119015566-119015566 on Build GRCh38
rs781821493 Chr.11: 119015505-119015507 on Build GRCh38
rs574402256 Chr.11: 119015752-119015752 on Build GRCh38
rs782333917 Chr.11: 119015794-119015794 on Build GRCh38
rs782292570 Chr.11: 119015575-119015575 on Build GRCh38
rs992496073 Chr.11: 119015773-119015773 on Build GRCh38
rs371828691 Chr.11: 119015603-119015603 on Build GRCh38

View Full Product Details