Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92831817-92832279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL5
Entrez Gene ID
6125
Gene Name
ribosomal protein L5
Gene Aliases
DBA6, L5, MSTP030, PPP1R135
UniGene
Hs.532359
Gene Chromosome Location
Chr.1: 92832037-92841924 on Build GRCh38
Transcripts
NM_000969.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5751392 c.3+3G>CG p.? Chr.1: 92832120-92832120 on Build GRCh38
4880548 c.2T>C p.M1T Chr.1: 92832116-92832116 on Build GRCh38
398532 c.3+1G>A p.? Chr.1: 92832118-92832118 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752282235 Chr.1: 92832041-92832041 on Build GRCh38
rs374750719 Chr.1: 92832084-92832084 on Build GRCh38
rs376524432 Chr.1: 92832103-92832103 on Build GRCh38
rs200437092 Chr.1: 92832057-92832057 on Build GRCh38
rs372092314 Chr.1: 92832101-92832101 on Build GRCh38
rs954141839 Chr.1: 92831912-92831912 on Build GRCh38
rs1019596498 Chr.1: 92831966-92831966 on Build GRCh38
rs755133627 Chr.1: 92832100-92832100 on Build GRCh38
rs886046558 Chr.1: 92832113-92832113 on Build GRCh38
rs573276148 Chr.1: 92831856-92831856 on Build GRCh38
rs184590739 Chr.1: 92832000-92832000 on Build GRCh38
rs752798871 Chr.1: 92832155-92832156 on Build GRCh38
rs927080395 Chr.1: 92832032-92832032 on Build GRCh38
rs116168890 Chr.1: 92832060-92832060 on Build GRCh38
rs368663464 Chr.1: 92832053-92832053 on Build GRCh38
rs767175723 Chr.1: 92831852-92831854 on Build GRCh38
rs954912810 Chr.1: 92832030-92832030 on Build GRCh38
rs937099898 Chr.1: 92832033-92832033 on Build GRCh38
rs759783048 Chr.1: 92832163-92832163 on Build GRCh38
rs746539975 Chr.1: 92832083-92832083 on Build GRCh38
rs372984365 Chr.1: 92832050-92832050 on Build GRCh38
rs958164665 Chr.1: 92832062-92832062 on Build GRCh38
rs763019487 Chr.1: 92832093-92832093 on Build GRCh38
rs759522508 Chr.1: 92832074-92832097 on Build GRCh38
rs138710531 Chr.1: 92832117-92832117 on Build GRCh38
rs936703983 Chr.1: 92831881-92831881 on Build GRCh38
rs376208311 Chr.1: 92832069-92832069 on Build GRCh38
rs748646858 Chr.1: 92832065-92832065 on Build GRCh38
rs375759804 Chr.1: 92832242-92832242 on Build GRCh38
rs889933305 Chr.1: 92832056-92832056 on Build GRCh38
rs199523145 Chr.1: 92832109-92832109 on Build GRCh38
rs1034277591 Chr.1: 92832204-92832204 on Build GRCh38
rs902783306 Chr.1: 92832046-92832046 on Build GRCh38
rs372196117 Chr.1: 92832081-92832081 on Build GRCh38
rs535694254 Chr.1: 92832071-92832071 on Build GRCh38
rs1055731216 Chr.1: 92831931-92831931 on Build GRCh38
rs145634330 Chr.1: 92832073-92832073 on Build GRCh38
rs562811182 Chr.1: 92832020-92832020 on Build GRCh38
rs745448466 Chr.1: 92832088-92832088 on Build GRCh38
rs529127733 Chr.1: 92831927-92831927 on Build GRCh38
rs372080902 Chr.1: 92832077-92832077 on Build GRCh38
rs767046909 Chr.1: 92831908-92831908 on Build GRCh38
rs1001445033 Chr.1: 92831874-92831874 on Build GRCh38
rs376070413 Chr.1: 92832067-92832067 on Build GRCh38
rs533207942 Chr.1: 92832039-92832039 on Build GRCh38
rs967561587 Chr.1: 92832234-92832234 on Build GRCh38
rs770926296 Chr.1: 92832157-92832157 on Build GRCh38
rs1003097504 Chr.1: 92831917-92831917 on Build GRCh38
rs1006135937 Chr.1: 92832051-92832051 on Build GRCh38
rs143326916 Chr.1: 92832063-92832063 on Build GRCh38
rs893545382 Chr.1: 92832245-92832245 on Build GRCh38
rs759157675 Chr.1: 92832248-92832248 on Build GRCh38
rs201525257 Chr.1: 92832104-92832104 on Build GRCh38
rs965346239 Chr.1: 92831975-92831975 on Build GRCh38
rs1056901437 Chr.1: 92832037-92832037 on Build GRCh38
rs945454563 Chr.1: 92831898-92831898 on Build GRCh38
rs374853816 Chr.1: 92832206-92832206 on Build GRCh38
rs993255566 Chr.1: 92832002-92832002 on Build GRCh38
rs918188409 Chr.1: 92832241-92832256 on Build GRCh38
rs201295012 Chr.1: 92832097-92832097 on Build GRCh38
rs553391857 Chr.1: 92831997-92831997 on Build GRCh38
rs753789228 Chr.1: 92832164-92832164 on Build GRCh38
rs113487315 Chr.1: 92832119-92832119 on Build GRCh38
rs774218096 Chr.1: 92832138-92832138 on Build GRCh38
rs1053694400 Chr.1: 92831887-92831887 on Build GRCh38
rs754042893 Chr.1: 92832072-92832072 on Build GRCh38
rs538267855 Chr.1: 92832152-92832152 on Build GRCh38
rs577876567 Chr.1: 92832176-92832176 on Build GRCh38
rs1001095608 Chr.1: 92832052-92832052 on Build GRCh38
rs200628272 Chr.1: 92832120-92832120 on Build GRCh38
rs573475582 Chr.1: 92832003-92832003 on Build GRCh38
rs376414614 Chr.1: 92832128-92832128 on Build GRCh38
rs749147586 Chr.1: 92832133-92832133 on Build GRCh38
rs775379177 Chr.1: 92832091-92832091 on Build GRCh38
rs763982728 Chr.1: 92832094-92832094 on Build GRCh38
rs756162066 Chr.1: 92832157-92832158 on Build GRCh38
rs756191753 Chr.1: 92832070-92832070 on Build GRCh38
rs758757902 Chr.1: 92832102-92832102 on Build GRCh38
rs370494227 Chr.1: 92832068-92832068 on Build GRCh38
rs761739588 Chr.1: 92832153-92832153 on Build GRCh38
rs147349722 Chr.1: 92832215-92832215 on Build GRCh38
rs769726665 Chr.1: 92832090-92832090 on Build GRCh38
rs367703317 Chr.1: 92831996-92831996 on Build GRCh38
rs902731779 Chr.1: 92831867-92831867 on Build GRCh38
rs929204674 Chr.1: 92832211-92832211 on Build GRCh38
rs562146660 Chr.1: 92831909-92831909 on Build GRCh38
rs992679960 Chr.1: 92832212-92832212 on Build GRCh38
rs991371988 Chr.1: 92832074-92832074 on Build GRCh38
rs772537960 Chr.1: 92832066-92832066 on Build GRCh38
rs910766530 Chr.1: 92831868-92831868 on Build GRCh38
rs142722855 Chr.1: 92832187-92832187 on Build GRCh38
rs191909916 Chr.1: 92832159-92832159 on Build GRCh38
rs1046495339 Chr.1: 92832043-92832043 on Build GRCh38
rs931204478 Chr.1: 92832004-92832004 on Build GRCh38
rs200130961 Chr.1: 92832129-92832129 on Build GRCh38
rs758851102 Chr.1: 92832078-92832078 on Build GRCh38
rs755878194 Chr.1: 92832061-92832061 on Build GRCh38
rs377193970 Chr.1: 92832172-92832172 on Build GRCh38
rs189107197 Chr.1: 92832130-92832130 on Build GRCh38
rs551112484 Chr.1: 92832045-92832045 on Build GRCh38
rs560167530 Chr.1: 92832192-92832192 on Build GRCh38
rs914025684 Chr.1: 92831895-92831895 on Build GRCh38
rs115830437 Chr.1: 92831865-92831865 on Build GRCh38
rs768489177 Chr.1: 92832135-92832135 on Build GRCh38
rs752291928 Chr.1: 92831923-92831923 on Build GRCh38
rs527330312 Chr.1: 92832199-92832199 on Build GRCh38
rs754061218 Chr.1: 92832098-92832098 on Build GRCh38
rs893997782 Chr.1: 92832113-92832113 on Build GRCh38
rs1005914628 Chr.1: 92831941-92831941 on Build GRCh38
rs972708406 Chr.1: 92831994-92831994 on Build GRCh38
rs774327587 Chr.1: 92832096-92832096 on Build GRCh38
rs767318302 Chr.1: 92832100-92832101 on Build GRCh38
rs958574832 Chr.1: 92832207-92832207 on Build GRCh38
rs1030470562 Chr.1: 92832017-92832017 on Build GRCh38
rs925648871 Chr.1: 92832252-92832252 on Build GRCh38
rs985496331 Chr.1: 92831938-92831938 on Build GRCh38
rs569209548 Chr.1: 92832075-92832075 on Build GRCh38
rs28365860 Chr.1: 92831846-92831847 on Build GRCh38

View Full Product Details