Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70281073-70281664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYOX1
Entrez Gene ID
51449
Gene Name
prenylcysteine oxidase 1
Gene Aliases
PCL1
UniGene
Hs.567502
Gene Chromosome Location
Chr.2: 70258099-70281185 on Build GRCh38
Transcripts
NM_016297.3 NM_001317165.1 NM_001317166.1 NM_001317167.1 NM_001317168.1 NM_001317169.1 NM_001317171.1 NM_003096.3

Gene Symbol
SNRPG
Entrez Gene ID
6637
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
SMG, Sm-G
UniGene
Hs.465167
Gene Chromosome Location
Chr.2: 70281362-70293771 on Build GRCh38
Transcripts
NM_016297.3 NM_001317165.1 NM_001317166.1 NM_001317167.1 NM_001317168.1 NM_001317169.1 NM_001317171.1 NM_003096.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337985 c.223C>T p.R75* Chr.2: 70281642-70281642 on Build GRCh38
3426635 c.201C>A p.I67I Chr.2: 70281664-70281664 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538908688 Chr.2: 70281135-70281135 on Build GRCh38
rs969509238 Chr.2: 70281343-70281343 on Build GRCh38
rs752921568 Chr.2: 70281592-70281593 on Build GRCh38
rs575430841 Chr.2: 70281139-70281139 on Build GRCh38
rs1024313239 Chr.2: 70281467-70281467 on Build GRCh38
rs958021417 Chr.2: 70281608-70281608 on Build GRCh38
rs374618408 Chr.2: 70281624-70281624 on Build GRCh38
rs889595775 Chr.2: 70281322-70281323 on Build GRCh38
rs929866211 Chr.2: 70281413-70281413 on Build GRCh38
rs989544225 Chr.2: 70281308-70281308 on Build GRCh38
rs1026642222 Chr.2: 70281420-70281420 on Build GRCh38
rs555033273 Chr.2: 70281182-70281182 on Build GRCh38
rs952994066 Chr.2: 70281239-70281239 on Build GRCh38
rs369021360 Chr.2: 70281598-70281598 on Build GRCh38
rs558829267 Chr.2: 70281137-70281137 on Build GRCh38
rs999795279 Chr.2: 70281288-70281288 on Build GRCh38
rs1030976792 Chr.2: 70281302-70281302 on Build GRCh38
rs1053808275 Chr.2: 70281309-70281310 on Build GRCh38
rs548116682 Chr.2: 70281327-70281327 on Build GRCh38
rs772584486 Chr.2: 70281389-70281389 on Build GRCh38
rs574865664 Chr.2: 70281233-70281233 on Build GRCh38
rs747508089 Chr.2: 70281433-70281433 on Build GRCh38
rs768846130 Chr.2: 70281591-70281591 on Build GRCh38
rs899546376 Chr.2: 70281512-70281512 on Build GRCh38
rs999763437 Chr.2: 70281585-70281585 on Build GRCh38
rs376520008 Chr.2: 70281148-70281148 on Build GRCh38
rs1007940815 Chr.2: 70281101-70281101 on Build GRCh38
rs1052340231 Chr.2: 70281571-70281571 on Build GRCh38
rs1039447167 Chr.2: 70281487-70281487 on Build GRCh38
rs916687260 Chr.2: 70281318-70281318 on Build GRCh38
rs900668202 Chr.2: 70281253-70281253 on Build GRCh38
rs111510626 Chr.2: 70281621-70281621 on Build GRCh38
rs1050242 Chr.2: 70281497-70281497 on Build GRCh38
rs961000795 Chr.2: 70281214-70281214 on Build GRCh38
rs767160133 Chr.2: 70281637-70281637 on Build GRCh38
rs377653110 Chr.2: 70281593-70281593 on Build GRCh38
rs915207550 Chr.2: 70281588-70281588 on Build GRCh38
rs14631 Chr.2: 70281457-70281457 on Build GRCh38
rs924033177 Chr.2: 70281285-70281285 on Build GRCh38
rs771487967 Chr.2: 70281476-70281476 on Build GRCh38
rs927839339 Chr.2: 70281147-70281147 on Build GRCh38
rs185195240 Chr.2: 70281181-70281181 on Build GRCh38
rs547051744 Chr.2: 70281132-70281132 on Build GRCh38
rs924001449 Chr.2: 70281532-70281532 on Build GRCh38
rs2272530 Chr.2: 70281427-70281427 on Build GRCh38
rs927840214 Chr.2: 70281387-70281387 on Build GRCh38
rs763564874 Chr.2: 70281603-70281603 on Build GRCh38
rs952961415 Chr.2: 70281428-70281428 on Build GRCh38
rs759307677 Chr.2: 70281599-70281599 on Build GRCh38
rs1046304649 Chr.2: 70281143-70281143 on Build GRCh38
rs753227560 Chr.2: 70281175-70281175 on Build GRCh38
rs748355544 Chr.2: 70281594-70281594 on Build GRCh38
rs4853438 Chr.2: 70281247-70281247 on Build GRCh38
rs762516246 Chr.2: 70281602-70281602 on Build GRCh38
rs534440407 Chr.2: 70281575-70281575 on Build GRCh38
rs997947487 Chr.2: 70281556-70281556 on Build GRCh38
rs756127320 Chr.2: 70281596-70281597 on Build GRCh38
rs890639099 Chr.2: 70281577-70281577 on Build GRCh38
rs765099160 Chr.2: 70281600-70281600 on Build GRCh38
rs764219909 Chr.2: 70281616-70281618 on Build GRCh38
rs989480809 Chr.2: 70281587-70281587 on Build GRCh38
rs561798025 Chr.2: 70281348-70281348 on Build GRCh38
rs527385265 Chr.2: 70281273-70281273 on Build GRCh38
rs545227654 Chr.2: 70281199-70281199 on Build GRCh38
rs546072481 Chr.2: 70281264-70281264 on Build GRCh38
rs533764536 Chr.2: 70281130-70281130 on Build GRCh38
rs993812434 Chr.2: 70281215-70281215 on Build GRCh38
rs982316409 Chr.2: 70281386-70281386 on Build GRCh38
rs560481541 Chr.2: 70281248-70281248 on Build GRCh38
rs982703853 Chr.2: 70281518-70281518 on Build GRCh38
rs772399256 Chr.2: 70281370-70281370 on Build GRCh38
rs941071006 Chr.2: 70281483-70281483 on Build GRCh38
rs993781160 Chr.2: 70281404-70281404 on Build GRCh38
rs947328923 Chr.2: 70281320-70281320 on Build GRCh38
rs1131031 Chr.2: 70281451-70281451 on Build GRCh38
rs527364226 Chr.2: 70281354-70281354 on Build GRCh38
rs914486894 Chr.2: 70281315-70281315 on Build GRCh38
rs1027176044 Chr.2: 70281231-70281231 on Build GRCh38
rs778147781 Chr.2: 70281384-70281384 on Build GRCh38
rs1050113078 Chr.2: 70281321-70281321 on Build GRCh38
rs189411195 Chr.2: 70281255-70281255 on Build GRCh38
rs570129940 Chr.2: 70281134-70281134 on Build GRCh38
rs1046692818 Chr.2: 70281229-70281229 on Build GRCh38
rs1031280041 Chr.2: 70281590-70281590 on Build GRCh38
rs751262864 Chr.2: 70281618-70281618 on Build GRCh38
rs896933760 Chr.2: 70281391-70281391 on Build GRCh38
rs749396936 Chr.2: 70281586-70281586 on Build GRCh38
rs956811081 Chr.2: 70281563-70281563 on Build GRCh38
rs147744340 Chr.2: 70281306-70281306 on Build GRCh38

View Full Product Details