Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 205742758-205743357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRP2
Entrez Gene ID
8828
Gene Name
neuropilin 2
Gene Aliases
NP2, NPN2, PRO2714, VEGF165R2
UniGene
Hs.471200
Gene Chromosome Location
Chr.2: 205682450-205798133 on Build GRCh38
Transcripts
NM_003872.2 NM_018534.3 NM_201264.1 NM_201266.1 NM_201267.1 NM_201279.1 XM_005246933.3 XM_005246934.3 XM_017005185.1 XM_017005186.1 XM_017005187.1 XM_017005188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6729888 c.1396C>T p.R466C Chr.2: 205743307-205743307 on Build GRCh38
1217853 c.1396C>T p.R466C Chr.2: 205743307-205743307 on Build GRCh38
5442502 c.1421G>A p.W474* Chr.2: 205743332-205743332 on Build GRCh38
7177339 c.1321C>T p.L441F Chr.2: 205743232-205743232 on Build GRCh38
5301989 c.1352C>T p.S451F Chr.2: 205743263-205743263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000704513 Chr.2: 205742798-205742798 on Build GRCh38
rs150214373 Chr.2: 205742875-205742875 on Build GRCh38
rs758275583 Chr.2: 205743283-205743283 on Build GRCh38
rs1030549824 Chr.2: 205743143-205743143 on Build GRCh38
rs990373295 Chr.2: 205743251-205743251 on Build GRCh38
rs1034506408 Chr.2: 205743024-205743024 on Build GRCh38
rs1009662363 Chr.2: 205742935-205742935 on Build GRCh38
rs530582033 Chr.2: 205743293-205743293 on Build GRCh38
rs749728296 Chr.2: 205743317-205743317 on Build GRCh38
rs201900948 Chr.2: 205743244-205743244 on Build GRCh38
rs958866840 Chr.2: 205743057-205743057 on Build GRCh38
rs765588675 Chr.2: 205743276-205743276 on Build GRCh38
rs757531378 Chr.2: 205743296-205743296 on Build GRCh38
rs751890151 Chr.2: 205743294-205743294 on Build GRCh38
rs551278469 Chr.2: 205743232-205743232 on Build GRCh38
rs1018027647 Chr.2: 205742805-205742805 on Build GRCh38
rs770776073 Chr.2: 205742852-205742852 on Build GRCh38
rs921057351 Chr.2: 205743280-205743280 on Build GRCh38
rs772852411 Chr.2: 205743243-205743243 on Build GRCh38
rs117695821 Chr.2: 205742832-205742832 on Build GRCh38
rs1021437408 Chr.2: 205743264-205743264 on Build GRCh38
rs763246719 Chr.2: 205743240-205743240 on Build GRCh38
rs746710925 Chr.2: 205743322-205743322 on Build GRCh38
rs894592388 Chr.2: 205743055-205743055 on Build GRCh38
rs769071559 Chr.2: 205743319-205743319 on Build GRCh38
rs189762105 Chr.2: 205743068-205743068 on Build GRCh38
rs781119570 Chr.2: 205743212-205743212 on Build GRCh38
rs559110601 Chr.2: 205743050-205743050 on Build GRCh38
rs942179443 Chr.2: 205743067-205743067 on Build GRCh38
rs849568 Chr.2: 205743315-205743315 on Build GRCh38
rs914464135 Chr.2: 205742919-205742919 on Build GRCh38
rs546952277 Chr.2: 205743337-205743337 on Build GRCh38
rs928584613 Chr.2: 205742944-205742944 on Build GRCh38
rs1014580065 Chr.2: 205742860-205742860 on Build GRCh38
rs752470835 Chr.2: 205743158-205743158 on Build GRCh38
rs776410675 Chr.2: 205742920-205742920 on Build GRCh38
rs554922921 Chr.2: 205743097-205743097 on Build GRCh38
rs993683342 Chr.2: 205743135-205743135 on Build GRCh38
rs200291757 Chr.2: 205743170-205743170 on Build GRCh38
rs756167318 Chr.2: 205743307-205743307 on Build GRCh38
rs1024672786 Chr.2: 205742965-205742965 on Build GRCh38
rs747163585 Chr.2: 205743180-205743180 on Build GRCh38
rs114761500 Chr.2: 205743175-205743175 on Build GRCh38
rs539121118 Chr.2: 205743013-205743013 on Build GRCh38
rs374695783 Chr.2: 205743161-205743161 on Build GRCh38
rs764031538 Chr.2: 205743338-205743338 on Build GRCh38
rs200906927 Chr.2: 205743326-205743326 on Build GRCh38
rs575067877 Chr.2: 205743138-205743138 on Build GRCh38
rs575721297 Chr.2: 205743064-205743064 on Build GRCh38
rs529980618 Chr.2: 205743327-205743327 on Build GRCh38
rs770168217 Chr.2: 205743225-205743225 on Build GRCh38
rs57269940 Chr.2: 205742976-205742976 on Build GRCh38
rs745964819 Chr.2: 205743301-205743301 on Build GRCh38
rs775358153 Chr.2: 205743336-205743336 on Build GRCh38
rs774513723 Chr.2: 205743286-205743286 on Build GRCh38
rs768950652 Chr.2: 205743241-205743241 on Build GRCh38
rs547016113 Chr.2: 205742930-205742930 on Build GRCh38
rs149849497 Chr.2: 205743323-205743323 on Build GRCh38
rs951384209 Chr.2: 205743145-205743145 on Build GRCh38
rs765113700 Chr.2: 205743268-205743268 on Build GRCh38
rs986923117 Chr.2: 205742780-205742780 on Build GRCh38
rs906503267 Chr.2: 205743284-205743284 on Build GRCh38
rs753261998 Chr.2: 205743265-205743265 on Build GRCh38
rs771203400 Chr.2: 205743198-205743198 on Build GRCh38
rs752624992 Chr.2: 205743282-205743282 on Build GRCh38
rs781373400 Chr.2: 205743299-205743299 on Build GRCh38
rs774680234 Chr.2: 205743321-205743321 on Build GRCh38
rs765927642 Chr.2: 205743260-205743260 on Build GRCh38
rs1037897221 Chr.2: 205743081-205743081 on Build GRCh38

View Full Product Details