Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 23644534-23645094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEL2
Entrez Gene ID
54551
Gene Name
MAGE family member L2
Gene Aliases
NDNL1, PWLS, SHFYNG, nM15
UniGene
Hs.141496
Gene Chromosome Location
Chr.15: 23643549-23647846 on Build GRCh38
Transcripts
NM_019066.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383064 c.1474A>G p.N492D Chr.15: 23644555-23644555 on Build GRCh38
5419768 c.1061A>G p.Q354R Chr.15: 23644968-23644968 on Build GRCh38
7376056 c.1347G>C p.L449F Chr.15: 23644682-23644682 on Build GRCh38
959908 c.1052G>T p.W351L Chr.15: 23644977-23644977 on Build GRCh38
433640 c.1347G>A p.L449L Chr.15: 23644682-23644682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769569057 Chr.15: 23645001-23645001 on Build GRCh38
rs978861820 Chr.15: 23644783-23644783 on Build GRCh38
rs138628273 Chr.15: 23644726-23644726 on Build GRCh38
rs376791514 Chr.15: 23644926-23644926 on Build GRCh38
rs371333192 Chr.15: 23644731-23644731 on Build GRCh38
rs772078014 Chr.15: 23644822-23644822 on Build GRCh38
rs754097303 Chr.15: 23644945-23644945 on Build GRCh38
rs778826873 Chr.15: 23645005-23645005 on Build GRCh38
rs753429920 Chr.15: 23644767-23644767 on Build GRCh38
rs747500040 Chr.15: 23644675-23644675 on Build GRCh38
rs750090677 Chr.15: 23644605-23644605 on Build GRCh38
rs779589753 Chr.15: 23644611-23644611 on Build GRCh38
rs924698907 Chr.15: 23644792-23644792 on Build GRCh38
rs771085386 Chr.15: 23644565-23644565 on Build GRCh38
rs761049409 Chr.15: 23644933-23644933 on Build GRCh38
rs774536525 Chr.15: 23644816-23644816 on Build GRCh38
rs371498748 Chr.15: 23644665-23644665 on Build GRCh38
rs555378546 Chr.15: 23644779-23644779 on Build GRCh38
rs770798048 Chr.15: 23644921-23644921 on Build GRCh38
rs757665824 Chr.15: 23644677-23644677 on Build GRCh38
rs934663418 Chr.15: 23644586-23644586 on Build GRCh38
rs373597907 Chr.15: 23644917-23644917 on Build GRCh38
rs768824507 Chr.15: 23644812-23644812 on Build GRCh38
rs567298347 Chr.15: 23644946-23644946 on Build GRCh38
rs545274091 Chr.15: 23644867-23644867 on Build GRCh38
rs780388413 Chr.15: 23644796-23644796 on Build GRCh38
rs763578669 Chr.15: 23644925-23644925 on Build GRCh38
rs778013247 Chr.15: 23644617-23644617 on Build GRCh38
rs754124963 Chr.15: 23644885-23644885 on Build GRCh38
rs777681443 Chr.15: 23644995-23644995 on Build GRCh38
rs991731216 Chr.15: 23644732-23644732 on Build GRCh38
rs772582222 Chr.15: 23644616-23644616 on Build GRCh38
rs753010151 Chr.15: 23645047-23645047 on Build GRCh38
rs534141639 Chr.15: 23644940-23644940 on Build GRCh38
rs751401500 Chr.15: 23644659-23644659 on Build GRCh38
rs527396971 Chr.15: 23644906-23644906 on Build GRCh38
rs538176500 Chr.15: 23644657-23644657 on Build GRCh38
rs781054286 Chr.15: 23645068-23645068 on Build GRCh38
rs745919740 Chr.15: 23645037-23645037 on Build GRCh38
rs189752384 Chr.15: 23644959-23644959 on Build GRCh38
rs930436568 Chr.15: 23644577-23644577 on Build GRCh38
rs775596633 Chr.15: 23644653-23644653 on Build GRCh38
rs368338798 Chr.15: 23644662-23644662 on Build GRCh38
rs771281637 Chr.15: 23644626-23644626 on Build GRCh38
rs746183066 Chr.15: 23644780-23644780 on Build GRCh38
rs796606490 Chr.15: 23644637-23644637 on Build GRCh38
rs755737731 Chr.15: 23644608-23644608 on Build GRCh38
rs769378200 Chr.15: 23644924-23644924 on Build GRCh38
rs767942371 Chr.15: 23644875-23644875 on Build GRCh38
rs398122417 Chr.15: 23644561-23644562 on Build GRCh38
rs560259716 Chr.15: 23644873-23644873 on Build GRCh38
rs370956711 Chr.15: 23645036-23645036 on Build GRCh38
rs762915060 Chr.15: 23644589-23644589 on Build GRCh38
rs558186319 Chr.15: 23644754-23644754 on Build GRCh38
rs756981993 Chr.15: 23644598-23644598 on Build GRCh38
rs776697724 Chr.15: 23644576-23644576 on Build GRCh38
rs544938560 Chr.15: 23644824-23644824 on Build GRCh38
rs781747956 Chr.15: 23644735-23644735 on Build GRCh38
rs376987796 Chr.15: 23644773-23644773 on Build GRCh38
rs2233069 Chr.15: 23644697-23644697 on Build GRCh38
rs765345034 Chr.15: 23644585-23644585 on Build GRCh38
rs748122624 Chr.15: 23644821-23644821 on Build GRCh38
rs769805022 Chr.15: 23644652-23644652 on Build GRCh38
rs757223336 Chr.15: 23645066-23645066 on Build GRCh38
rs150094328 Chr.15: 23644579-23644579 on Build GRCh38
rs374261657 Chr.15: 23644625-23644625 on Build GRCh38
rs368008966 Chr.15: 23644859-23644859 on Build GRCh38
rs370022000 Chr.15: 23644666-23644666 on Build GRCh38
rs779151079 Chr.15: 23644900-23644900 on Build GRCh38
rs956898064 Chr.15: 23644685-23644685 on Build GRCh38
rs2233070 Chr.15: 23644592-23644592 on Build GRCh38
rs377086939 Chr.15: 23645004-23645004 on Build GRCh38
rs573975752 Chr.15: 23644791-23644791 on Build GRCh38
rs1035333736 Chr.15: 23644725-23644725 on Build GRCh38
rs766076021 Chr.15: 23644855-23644855 on Build GRCh38
rs548982518 Chr.15: 23644922-23644922 on Build GRCh38
rs770005224 Chr.15: 23644716-23644716 on Build GRCh38
rs369568784 Chr.15: 23644950-23644950 on Build GRCh38
rs767400841 Chr.15: 23644663-23644663 on Build GRCh38
rs576711188 Chr.15: 23644603-23644603 on Build GRCh38
rs398122418 Chr.15: 23644619-23644619 on Build GRCh38
rs756557403 Chr.15: 23644882-23644882 on Build GRCh38
rs371613799 Chr.15: 23644615-23644615 on Build GRCh38
rs781054761 Chr.15: 23644998-23644998 on Build GRCh38
rs773146235 Chr.15: 23644825-23644825 on Build GRCh38
rs746118539 Chr.15: 23644710-23644710 on Build GRCh38
rs185434800 Chr.15: 23644778-23644778 on Build GRCh38
rs773587128 Chr.15: 23644554-23644554 on Build GRCh38
rs749321139 Chr.15: 23644728-23644728 on Build GRCh38
rs762817125 Chr.15: 23644655-23644655 on Build GRCh38
rs776894525 Chr.15: 23644633-23644633 on Build GRCh38
rs562929572 Chr.15: 23644842-23644842 on Build GRCh38
rs760258934 Chr.15: 23644755-23644755 on Build GRCh38
rs746667147 Chr.15: 23644873-23644908 on Build GRCh38
rs932677720 Chr.15: 23645058-23645058 on Build GRCh38
rs531503349 Chr.15: 23644951-23644951 on Build GRCh38
rs760909346 Chr.15: 23644563-23644563 on Build GRCh38
rs1050365820 Chr.15: 23644884-23644884 on Build GRCh38
rs573023653 Chr.15: 23644766-23644766 on Build GRCh38
rs375960990 Chr.15: 23644740-23644740 on Build GRCh38
rs777917199 Chr.15: 23644919-23644919 on Build GRCh38
rs770724034 Chr.15: 23644996-23644996 on Build GRCh38
rs776392903 Chr.15: 23644856-23644856 on Build GRCh38
rs1048925339 Chr.15: 23644800-23644800 on Build GRCh38
rs941843764 Chr.15: 23644902-23644902 on Build GRCh38
rs180690391 Chr.15: 23645051-23645051 on Build GRCh38
rs764254872 Chr.15: 23645052-23645052 on Build GRCh38
rs755206356 Chr.15: 23644896-23644896 on Build GRCh38
rs778412481 Chr.15: 23644672-23644672 on Build GRCh38
rs918999260 Chr.15: 23644785-23644785 on Build GRCh38
rs1004039085 Chr.15: 23644629-23644629 on Build GRCh38
rs370813839 Chr.15: 23645061-23645061 on Build GRCh38
rs758835928 Chr.15: 23644668-23644668 on Build GRCh38
rs749451613 Chr.15: 23644805-23644805 on Build GRCh38
rs745591482 Chr.15: 23645000-23645000 on Build GRCh38
rs567531448 Chr.15: 23644645-23644645 on Build GRCh38
rs2233068 Chr.15: 23644857-23644857 on Build GRCh38
rs773688828 Chr.15: 23645006-23645006 on Build GRCh38
rs750887387 Chr.15: 23644878-23644878 on Build GRCh38
rs373945272 Chr.15: 23645038-23645038 on Build GRCh38
rs752626295 Chr.15: 23644588-23644588 on Build GRCh38
rs780253061 Chr.15: 23644883-23644883 on Build GRCh38
rs368189645 Chr.15: 23645075-23645075 on Build GRCh38
rs752153357 Chr.15: 23644862-23644862 on Build GRCh38
rs753331962 Chr.15: 23644667-23644667 on Build GRCh38
rs369891333 Chr.15: 23644764-23644764 on Build GRCh38

View Full Product Details