Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69415816-69416364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF23
Entrez Gene ID
9493
Gene Name
kinesin family member 23
Gene Aliases
CHO1, KNSL5, MKLP-1, MKLP1
UniGene
Hs.270845
Gene Chromosome Location
Chr.15: 69414246-69448427 on Build GRCh38
Transcripts
NM_001281301.1 NM_004856.6 NM_138555.3 XM_005254796.2 XM_005254797.2 XM_005254798.2 XM_005254799.2 XM_011522238.1 XM_011522239.2 XM_017022733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370239 c.28C>T p.R10W Chr.15: 69416010-69416010 on Build GRCh38
1129133 c.49G>A p.G17R Chr.15: 69416031-69416031 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369931528 Chr.15: 69415946-69415946 on Build GRCh38
rs368198580 Chr.15: 69416198-69416198 on Build GRCh38
rs915776207 Chr.15: 69416217-69416217 on Build GRCh38
rs922722206 Chr.15: 69415867-69415867 on Build GRCh38
rs373867203 Chr.15: 69416039-69416039 on Build GRCh38
rs748122634 Chr.15: 69416060-69416060 on Build GRCh38
rs760453944 Chr.15: 69416258-69416258 on Build GRCh38
rs146134944 Chr.15: 69416011-69416011 on Build GRCh38
rs546256393 Chr.15: 69416027-69416027 on Build GRCh38
rs774923273 Chr.15: 69416109-69416109 on Build GRCh38
rs960245455 Chr.15: 69416175-69416175 on Build GRCh38
rs755081616 Chr.15: 69416046-69416046 on Build GRCh38
rs560244349 Chr.15: 69415868-69415868 on Build GRCh38
rs143818035 Chr.15: 69416177-69416179 on Build GRCh38
rs955693690 Chr.15: 69416166-69416166 on Build GRCh38
rs765647999 Chr.15: 69416035-69416035 on Build GRCh38
rs1050263693 Chr.15: 69415913-69415913 on Build GRCh38
rs567391736 Chr.15: 69415914-69415914 on Build GRCh38
rs1023117449 Chr.15: 69416123-69416123 on Build GRCh38
rs773971888 Chr.15: 69416089-69416089 on Build GRCh38
rs749033383 Chr.15: 69416070-69416070 on Build GRCh38
rs999266892 Chr.15: 69416146-69416146 on Build GRCh38
rs558435692 Chr.15: 69416161-69416161 on Build GRCh38
rs766017103 Chr.15: 69415956-69415957 on Build GRCh38
rs764623875 Chr.15: 69416019-69416019 on Build GRCh38
rs751218869 Chr.15: 69415966-69415968 on Build GRCh38
rs752490219 Chr.15: 69416106-69416107 on Build GRCh38
rs969107298 Chr.15: 69415922-69415922 on Build GRCh38
rs759147281 Chr.15: 69416106-69416106 on Build GRCh38
rs767365881 Chr.15: 69416091-69416091 on Build GRCh38
rs762683504 Chr.15: 69415984-69415984 on Build GRCh38
rs1047870541 Chr.15: 69416320-69416320 on Build GRCh38
rs372224314 Chr.15: 69416108-69416108 on Build GRCh38
rs1037441915 Chr.15: 69415969-69415969 on Build GRCh38
rs61751118 Chr.15: 69416015-69416015 on Build GRCh38
rs762394079 Chr.15: 69415957-69415957 on Build GRCh38
rs183790083 Chr.15: 69416241-69416241 on Build GRCh38
rs754752859 Chr.15: 69416024-69416025 on Build GRCh38
rs999957248 Chr.15: 69416131-69416131 on Build GRCh38
rs188578503 Chr.15: 69416299-69416299 on Build GRCh38
rs770677372 Chr.15: 69416087-69416087 on Build GRCh38
rs969860546 Chr.15: 69416232-69416232 on Build GRCh38
rs575046333 Chr.15: 69416235-69416235 on Build GRCh38
rs948464053 Chr.15: 69415967-69415967 on Build GRCh38
rs181341703 Chr.15: 69416300-69416300 on Build GRCh38
rs112860684 Chr.15: 69416069-69416069 on Build GRCh38
rs536210797 Chr.15: 69416022-69416022 on Build GRCh38
rs770423663 Chr.15: 69416004-69416004 on Build GRCh38
rs758710646 Chr.15: 69416041-69416041 on Build GRCh38
rs905637488 Chr.15: 69416127-69416127 on Build GRCh38
rs915722379 Chr.15: 69415966-69415966 on Build GRCh38
rs955724983 Chr.15: 69416309-69416309 on Build GRCh38
rs773977730 Chr.15: 69416010-69416010 on Build GRCh38
rs772838359 Chr.15: 69415965-69415965 on Build GRCh38
rs566508814 Chr.15: 69416044-69416044 on Build GRCh38
rs139012996 Chr.15: 69416058-69416058 on Build GRCh38
rs929032202 Chr.15: 69416304-69416304 on Build GRCh38
rs911347921 Chr.15: 69415962-69415962 on Build GRCh38
rs754580439 Chr.15: 69415844-69415844 on Build GRCh38

View Full Product Details