Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124405212-124405658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB7
Entrez Gene ID
5695
Gene Name
proteasome subunit beta 7
Gene Aliases
Z
UniGene
Hs.213470
Gene Chromosome Location
Chr.9: 124353465-124415473 on Build GRCh38
Transcripts
NM_002799.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3324578 c.427T>G p.L143V Chr.9: 124405401-124405401 on Build GRCh38
6182315 c.432G>T p.G144G Chr.9: 124405396-124405396 on Build GRCh38
4370353 c.427T>G p.L143V Chr.9: 124405401-124405401 on Build GRCh38
4387294 c.440A>T p.D147V Chr.9: 124405388-124405388 on Build GRCh38
7155589 c.400C>G p.Q134E Chr.9: 124405428-124405428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960883654 Chr.9: 124405577-124405577 on Build GRCh38
rs144936453 Chr.9: 124405267-124405267 on Build GRCh38
rs189585240 Chr.9: 124405286-124405286 on Build GRCh38
rs367807454 Chr.9: 124405467-124405467 on Build GRCh38
rs182278812 Chr.9: 124405445-124405445 on Build GRCh38
rs377374304 Chr.9: 124405448-124405448 on Build GRCh38
rs764488096 Chr.9: 124405464-124405464 on Build GRCh38
rs779131826 Chr.9: 124405310-124405310 on Build GRCh38
rs773029620 Chr.9: 124405452-124405452 on Build GRCh38
rs1011674000 Chr.9: 124405596-124405596 on Build GRCh38
rs1041906773 Chr.9: 124405610-124405610 on Build GRCh38
rs746054413 Chr.9: 124405319-124405319 on Build GRCh38
rs747341041 Chr.9: 124405455-124405456 on Build GRCh38
rs765971898 Chr.9: 124405454-124405454 on Build GRCh38
rs10986331 Chr.9: 124405281-124405281 on Build GRCh38
rs141948150 Chr.9: 124405372-124405372 on Build GRCh38
rs142174520 Chr.9: 124405507-124405507 on Build GRCh38
rs111424805 Chr.9: 124405624-124405624 on Build GRCh38
rs1045958574 Chr.9: 124405444-124405444 on Build GRCh38
rs377142672 Chr.9: 124405270-124405270 on Build GRCh38
rs1033891281 Chr.9: 124405555-124405555 on Build GRCh38
rs548942727 Chr.9: 124405569-124405569 on Build GRCh38
rs1002464865 Chr.9: 124405509-124405509 on Build GRCh38
rs903020173 Chr.9: 124405628-124405628 on Build GRCh38
rs187697241 Chr.9: 124405619-124405619 on Build GRCh38
rs766801412 Chr.9: 124405561-124405561 on Build GRCh38
rs62581780 Chr.9: 124405523-124405523 on Build GRCh38
rs185514981 Chr.9: 124405269-124405269 on Build GRCh38
rs74699968 Chr.9: 124405463-124405463 on Build GRCh38
rs776526802 Chr.9: 124405399-124405399 on Build GRCh38
rs551509849 Chr.9: 124405328-124405328 on Build GRCh38
rs77255967 Chr.9: 124405629-124405629 on Build GRCh38
rs997441695 Chr.9: 124405259-124405259 on Build GRCh38
rs763407032 Chr.9: 124405593-124405593 on Build GRCh38
rs199533198 Chr.9: 124405361-124405361 on Build GRCh38
rs761677209 Chr.9: 124405437-124405437 on Build GRCh38
rs768471397 Chr.9: 124405396-124405396 on Build GRCh38
rs775370564 Chr.9: 124405336-124405336 on Build GRCh38
rs749586418 Chr.9: 124405277-124405277 on Build GRCh38
rs761285972 Chr.9: 124405457-124405457 on Build GRCh38
rs893186198 Chr.9: 124405572-124405572 on Build GRCh38
rs3209221 Chr.9: 124405394-124405394 on Build GRCh38
rs1022716739 Chr.9: 124405456-124405456 on Build GRCh38
rs908938545 Chr.9: 124405283-124405283 on Build GRCh38
rs149094712 Chr.9: 124405306-124405306 on Build GRCh38
rs1030241276 Chr.9: 124405633-124405633 on Build GRCh38
rs751027561 Chr.9: 124405455-124405455 on Build GRCh38
rs772147967 Chr.9: 124405330-124405330 on Build GRCh38

View Full Product Details