Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18242178-18242724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCML2
Entrez Gene ID
10389
Gene Name
sex comb on midleg-like 2 (Drosophila)
Gene Aliases
-
UniGene
Hs.495774
Gene Chromosome Location
Chr.X: 18239313-18355089 on Build GRCh38
Transcripts
NM_006089.2 NR_033717.1 XM_006724458.1 XM_017029219.1 XM_017029220.1 XM_017029221.1 XM_017029222.1

SNP Detail

SNP ID Chromosome Location
rs768888311 Chr.X: 18242560-18242560 on Build GRCh38
rs751981144 Chr.X: 18242324-18242324 on Build GRCh38
rs754710317 Chr.X: 18242414-18242414 on Build GRCh38
rs945658084 Chr.X: 18242580-18242580 on Build GRCh38
rs781559244 Chr.X: 18242382-18242382 on Build GRCh38
rs749773517 Chr.X: 18242598-18242598 on Build GRCh38
rs777118606 Chr.X: 18242565-18242565 on Build GRCh38
rs10521679 Chr.X: 18242206-18242206 on Build GRCh38
rs772670145 Chr.X: 18242599-18242599 on Build GRCh38
rs758948793 Chr.X: 18242279-18242279 on Build GRCh38
rs910060617 Chr.X: 18242268-18242268 on Build GRCh38
rs895181919 Chr.X: 18242422-18242422 on Build GRCh38
rs999336713 Chr.X: 18242659-18242659 on Build GRCh38
rs1043920853 Chr.X: 18242525-18242525 on Build GRCh38
rs1055166105 Chr.X: 18242401-18242401 on Build GRCh38
rs745963601 Chr.X: 18242475-18242475 on Build GRCh38
rs779075189 Chr.X: 18242474-18242474 on Build GRCh38
rs376375521 Chr.X: 18242456-18242456 on Build GRCh38
rs769311159 Chr.X: 18242532-18242533 on Build GRCh38
rs946829355 Chr.X: 18242433-18242433 on Build GRCh38
rs769459791 Chr.X: 18242596-18242596 on Build GRCh38
rs747599703 Chr.X: 18242529-18242529 on Build GRCh38
rs753640908 Chr.X: 18242308-18242308 on Build GRCh38
rs761315081 Chr.X: 18242568-18242568 on Build GRCh38
rs930732449 Chr.X: 18242298-18242298 on Build GRCh38
rs928226382 Chr.X: 18242348-18242348 on Build GRCh38
rs972999847 Chr.X: 18242323-18242323 on Build GRCh38
rs776108971 Chr.X: 18242490-18242490 on Build GRCh38
rs184300113 Chr.X: 18242376-18242376 on Build GRCh38
rs780842337 Chr.X: 18242525-18242526 on Build GRCh38
rs756868143 Chr.X: 18242490-18242490 on Build GRCh38
rs745396352 Chr.X: 18242530-18242531 on Build GRCh38
rs190465403 Chr.X: 18242420-18242420 on Build GRCh38
rs758283304 Chr.X: 18242394-18242394 on Build GRCh38
rs772663129 Chr.X: 18242485-18242485 on Build GRCh38
rs766335036 Chr.X: 18242408-18242408 on Build GRCh38
rs757432051 Chr.X: 18242338-18242338 on Build GRCh38
rs951870720 Chr.X: 18242219-18242219 on Build GRCh38
rs112628761 Chr.X: 18242290-18242290 on Build GRCh38
rs779013143 Chr.X: 18242363-18242363 on Build GRCh38
rs762422520 Chr.X: 18242602-18242602 on Build GRCh38
rs61751427 Chr.X: 18242528-18242528 on Build GRCh38
rs1018803406 Chr.X: 18242701-18242701 on Build GRCh38
rs760742728 Chr.X: 18242208-18242208 on Build GRCh38
rs751488055 Chr.X: 18242636-18242636 on Build GRCh38
rs769069232 Chr.X: 18242605-18242605 on Build GRCh38
rs781101887 Chr.X: 18242418-18242418 on Build GRCh38
rs748336734 Chr.X: 18242600-18242600 on Build GRCh38
rs765925877 Chr.X: 18242635-18242635 on Build GRCh38
rs182197283 Chr.X: 18242700-18242700 on Build GRCh38
rs974461117 Chr.X: 18242205-18242205 on Build GRCh38
rs984658978 Chr.X: 18242224-18242224 on Build GRCh38
rs775902143 Chr.X: 18242597-18242598 on Build GRCh38
rs944692413 Chr.X: 18242407-18242407 on Build GRCh38
rs745720826 Chr.X: 18242590-18242590 on Build GRCh38
rs751542848 Chr.X: 18242412-18242412 on Build GRCh38
rs758500403 Chr.X: 18242390-18242390 on Build GRCh38
rs749469844 Chr.X: 18242419-18242419 on Build GRCh38

View Full Product Details