Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47406544-47407004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB1
Entrez Gene ID
2560
Gene Name
gamma-aminobutyric acid type A receptor beta1 subunit
Gene Aliases
-
UniGene
Hs.254117
Gene Chromosome Location
Chr.4: 47031278-47438409 on Build GRCh38
Transcripts
NM_000812.3 XM_017007985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1055701 c.954C>A p.F318L Chr.4: 47406800-47406800 on Build GRCh38
4400805 c.900C>T p.I300I Chr.4: 47406746-47406746 on Build GRCh38
5995332 c.860C>A p.T287K Chr.4: 47406706-47406706 on Build GRCh38
5444305 c.910A>C p.K304Q Chr.4: 47406756-47406756 on Build GRCh38
4399881 c.1029A>G p.K343K Chr.4: 47406875-47406875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769038641 Chr.4: 47406910-47406910 on Build GRCh38
rs1050513607 Chr.4: 47406846-47406846 on Build GRCh38
rs6290 Chr.4: 47406821-47406821 on Build GRCh38
rs768805804 Chr.4: 47406752-47406752 on Build GRCh38
rs756987530 Chr.4: 47406867-47406867 on Build GRCh38
rs780557867 Chr.4: 47406875-47406875 on Build GRCh38
rs539442260 Chr.4: 47406947-47406947 on Build GRCh38
rs780797468 Chr.4: 47406712-47406712 on Build GRCh38
rs548869600 Chr.4: 47406761-47406761 on Build GRCh38
rs866922910 Chr.4: 47406710-47406710 on Build GRCh38
rs1003137152 Chr.4: 47406972-47406972 on Build GRCh38
rs184713186 Chr.4: 47406577-47406577 on Build GRCh38
rs775709875 Chr.4: 47406728-47406728 on Build GRCh38
rs775949181 Chr.4: 47406949-47406949 on Build GRCh38
rs749841128 Chr.4: 47406960-47406960 on Build GRCh38
rs80178222 Chr.4: 47406725-47406725 on Build GRCh38
rs951653630 Chr.4: 47406939-47406939 on Build GRCh38
rs530767211 Chr.4: 47406743-47406743 on Build GRCh38
rs767463256 Chr.4: 47406776-47406776 on Build GRCh38
rs765037938 Chr.4: 47406945-47406945 on Build GRCh38
rs763027668 Chr.4: 47406946-47406946 on Build GRCh38
rs377260679 Chr.4: 47406918-47406918 on Build GRCh38
rs763802053 Chr.4: 47406858-47406858 on Build GRCh38
rs780497165 Chr.4: 47406971-47406971 on Build GRCh38
rs752605683 Chr.4: 47406665-47406665 on Build GRCh38
rs939181875 Chr.4: 47406647-47406647 on Build GRCh38
rs1011717950 Chr.4: 47406612-47406612 on Build GRCh38
rs1020859953 Chr.4: 47406613-47406613 on Build GRCh38
rs6289 Chr.4: 47406692-47406692 on Build GRCh38
rs541045568 Chr.4: 47406618-47406618 on Build GRCh38
rs761839362 Chr.4: 47406774-47406774 on Build GRCh38
rs559346676 Chr.4: 47406638-47406638 on Build GRCh38
rs886039818 Chr.4: 47406706-47406706 on Build GRCh38
rs904539553 Chr.4: 47406758-47406758 on Build GRCh38
rs919651362 Chr.4: 47406715-47406715 on Build GRCh38
rs755669497 Chr.4: 47406961-47406961 on Build GRCh38
rs778550734 Chr.4: 47406663-47406663 on Build GRCh38
rs6287 Chr.4: 47406648-47406648 on Build GRCh38
rs573622433 Chr.4: 47406606-47406606 on Build GRCh38
rs758058750 Chr.4: 47406675-47406675 on Build GRCh38
rs759198088 Chr.4: 47406689-47406689 on Build GRCh38
rs760604569 Chr.4: 47406835-47406835 on Build GRCh38
rs115114447 Chr.4: 47406833-47406833 on Build GRCh38
rs565774468 Chr.4: 47406886-47406886 on Build GRCh38
rs151232894 Chr.4: 47406951-47406951 on Build GRCh38
rs766966325 Chr.4: 47406950-47406950 on Build GRCh38
rs982663843 Chr.4: 47406726-47406726 on Build GRCh38
rs749603787 Chr.4: 47406894-47406894 on Build GRCh38
rs962697887 Chr.4: 47406956-47406956 on Build GRCh38
rs372746217 Chr.4: 47406666-47406666 on Build GRCh38
rs780621488 Chr.4: 47406887-47406887 on Build GRCh38
rs748346475 Chr.4: 47406926-47406926 on Build GRCh38
rs577240874 Chr.4: 47406935-47406935 on Build GRCh38
rs751229590 Chr.4: 47406865-47406865 on Build GRCh38
rs142758601 Chr.4: 47406847-47406847 on Build GRCh38
rs967680534 Chr.4: 47406643-47406643 on Build GRCh38
rs79183288 Chr.4: 47406586-47406586 on Build GRCh38
rs545362380 Chr.4: 47406662-47406662 on Build GRCh38
rs773389083 Chr.4: 47406779-47406779 on Build GRCh38
rs746343478 Chr.4: 47406719-47406719 on Build GRCh38
rs1029047798 Chr.4: 47406598-47406598 on Build GRCh38
rs35402897 Chr.4: 47406727-47406727 on Build GRCh38
rs140339384 Chr.4: 47406980-47406980 on Build GRCh38
rs772396298 Chr.4: 47406931-47406931 on Build GRCh38
rs150598937 Chr.4: 47406879-47406879 on Build GRCh38
rs369153302 Chr.4: 47406649-47406649 on Build GRCh38
rs747582639 Chr.4: 47406977-47406978 on Build GRCh38
rs763656247 Chr.4: 47406836-47406836 on Build GRCh38
rs980422200 Chr.4: 47406965-47406965 on Build GRCh38

View Full Product Details