Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170892355-170892743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP17
Entrez Gene ID
64901
Gene Name
RAN binding protein 17
Gene Aliases
-
UniGene
Hs.410810
Gene Chromosome Location
Chr.5: 170861882-171300015 on Build GRCh38
Transcripts
NM_022897.4 XM_011534627.1 XM_011534631.2 XM_011534636.2 XM_011534637.2 XM_017009736.1 XM_017009737.1 XM_017009738.1 XM_017009739.1 XM_017009740.1 XM_017009741.1 XM_017009742.1 XM_017009743.1 XM_017009744.1 XM_017009746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012144 c.264C>T p.Y88Y Chr.5: 170892394-170892394 on Build GRCh38
1065783 c.277G>A p.V93M Chr.5: 170892407-170892407 on Build GRCh38
1310959 c.420C>G p.L140L Chr.5: 170892550-170892550 on Build GRCh38
1486596 c.353G>T p.G118V Chr.5: 170892483-170892483 on Build GRCh38
3339321 c.386T>C p.F129S Chr.5: 170892516-170892516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023748492 Chr.5: 170892656-170892656 on Build GRCh38
rs149823158 Chr.5: 170892685-170892685 on Build GRCh38
rs558055947 Chr.5: 170892628-170892628 on Build GRCh38
rs760415267 Chr.5: 170892524-170892524 on Build GRCh38
rs761599887 Chr.5: 170892531-170892531 on Build GRCh38
rs745372789 Chr.5: 170892508-170892508 on Build GRCh38
rs1010886647 Chr.5: 170892688-170892688 on Build GRCh38
rs755062162 Chr.5: 170892459-170892459 on Build GRCh38
rs561106815 Chr.5: 170892662-170892662 on Build GRCh38
rs372451784 Chr.5: 170892406-170892406 on Build GRCh38
rs763591202 Chr.5: 170892397-170892397 on Build GRCh38
rs749458069 Chr.5: 170892583-170892583 on Build GRCh38
rs778141018 Chr.5: 170892579-170892579 on Build GRCh38
rs141535602 Chr.5: 170892402-170892402 on Build GRCh38
rs867165048 Chr.5: 170892431-170892431 on Build GRCh38
rs781080945 Chr.5: 170892464-170892464 on Build GRCh38
rs1016512600 Chr.5: 170892680-170892680 on Build GRCh38
rs966550255 Chr.5: 170892462-170892462 on Build GRCh38
rs940207058 Chr.5: 170892617-170892619 on Build GRCh38
rs772428006 Chr.5: 170892600-170892600 on Build GRCh38
rs199744676 Chr.5: 170892535-170892535 on Build GRCh38
rs141503316 Chr.5: 170892509-170892509 on Build GRCh38
rs775006467 Chr.5: 170892503-170892503 on Build GRCh38
rs200625625 Chr.5: 170892572-170892572 on Build GRCh38
rs548752197 Chr.5: 170892398-170892398 on Build GRCh38
rs896541024 Chr.5: 170892640-170892640 on Build GRCh38
rs760064184 Chr.5: 170892506-170892506 on Build GRCh38
rs753698563 Chr.5: 170892528-170892528 on Build GRCh38
rs764952588 Chr.5: 170892550-170892550 on Build GRCh38
rs773412829 Chr.5: 170892390-170892390 on Build GRCh38
rs570828025 Chr.5: 170892453-170892453 on Build GRCh38
rs769872460 Chr.5: 170892485-170892485 on Build GRCh38
rs1006427607 Chr.5: 170892673-170892673 on Build GRCh38
rs763640787 Chr.5: 170892526-170892526 on Build GRCh38
rs79372097 Chr.5: 170892571-170892571 on Build GRCh38
rs903138079 Chr.5: 170892624-170892624 on Build GRCh38
rs534903364 Chr.5: 170892589-170892589 on Build GRCh38
rs372086128 Chr.5: 170892493-170892493 on Build GRCh38
rs139889860 Chr.5: 170892671-170892671 on Build GRCh38
rs779373634 Chr.5: 170892593-170892593 on Build GRCh38
rs763981721 Chr.5: 170892420-170892420 on Build GRCh38
rs367760791 Chr.5: 170892574-170892574 on Build GRCh38
rs34726683 Chr.5: 170892652-170892653 on Build GRCh38
rs79439344 Chr.5: 170892513-170892513 on Build GRCh38
rs901319479 Chr.5: 170892652-170892652 on Build GRCh38
rs201660848 Chr.5: 170892565-170892565 on Build GRCh38
rs77741839 Chr.5: 170892615-170892615 on Build GRCh38
rs377186217 Chr.5: 170892491-170892491 on Build GRCh38
rs758225041 Chr.5: 170892570-170892570 on Build GRCh38
rs752410005 Chr.5: 170892410-170892410 on Build GRCh38
rs748429696 Chr.5: 170892476-170892476 on Build GRCh38
rs778253214 Chr.5: 170892486-170892486 on Build GRCh38
rs760197441 Chr.5: 170892415-170892415 on Build GRCh38
rs531086103 Chr.5: 170892407-170892407 on Build GRCh38
rs1052872891 Chr.5: 170892630-170892630 on Build GRCh38
rs1013694625 Chr.5: 170892655-170892655 on Build GRCh38
rs753607706 Chr.5: 170892422-170892422 on Build GRCh38
rs1050397474 Chr.5: 170892681-170892684 on Build GRCh38
rs200291830 Chr.5: 170892449-170892449 on Build GRCh38
rs752943557 Chr.5: 170892594-170892595 on Build GRCh38
rs144659787 Chr.5: 170892447-170892447 on Build GRCh38
rs759685136 Chr.5: 170892404-170892405 on Build GRCh38
rs765419064 Chr.5: 170892595-170892595 on Build GRCh38
rs746409638 Chr.5: 170892597-170892597 on Build GRCh38

View Full Product Details