Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212365141-212365650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM206
Entrez Gene ID
55248
Gene Name
transmembrane protein 206
Gene Aliases
C1orf75
UniGene
Hs.445386
Gene Chromosome Location
Chr.1: 212364474-212415508 on Build GRCh38
Transcripts
NM_001198862.1 NM_018252.2 XM_005273177.3 XM_011509717.2 XM_011509718.2 XM_017001634.1 XM_017001635.1 XM_017001636.1 XM_017001637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3483137 c.1037C>A p.T346K Chr.1: 212365231-212365231 on Build GRCh38
463901 c.925C>A p.L309I Chr.1: 212365343-212365343 on Build GRCh38
5845389 c.1078G>A p.V360I Chr.1: 212365373-212365373 on Build GRCh38
7109812 c.1174A>G p.M392V Chr.1: 212365277-212365277 on Build GRCh38
7104240 c.930C>T p.L310L Chr.1: 212365338-212365338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371692119 Chr.1: 212365491-212365491 on Build GRCh38
rs756072956 Chr.1: 212365224-212365224 on Build GRCh38
rs879161931 Chr.1: 212365430-212365431 on Build GRCh38
rs925223548 Chr.1: 212365515-212365515 on Build GRCh38
rs904129768 Chr.1: 212365535-212365535 on Build GRCh38
rs751282505 Chr.1: 212365205-212365205 on Build GRCh38
rs754502878 Chr.1: 212365395-212365395 on Build GRCh38
rs761457991 Chr.1: 212365336-212365336 on Build GRCh38
rs146634843 Chr.1: 212365574-212365574 on Build GRCh38
rs374434268 Chr.1: 212365195-212365195 on Build GRCh38
rs192890813 Chr.1: 212365303-212365303 on Build GRCh38
rs778369163 Chr.1: 212365405-212365405 on Build GRCh38
rs1055366503 Chr.1: 212365532-212365532 on Build GRCh38
rs537692214 Chr.1: 212365462-212365462 on Build GRCh38
rs370633643 Chr.1: 212365204-212365204 on Build GRCh38
rs1034429230 Chr.1: 212365260-212365260 on Build GRCh38
rs780942180 Chr.1: 212365209-212365209 on Build GRCh38
rs75113230 Chr.1: 212365369-212365369 on Build GRCh38
rs933670407 Chr.1: 212365165-212365165 on Build GRCh38
rs373903663 Chr.1: 212365429-212365429 on Build GRCh38
rs778564779 Chr.1: 212365241-212365241 on Build GRCh38
rs1019667034 Chr.1: 212365584-212365584 on Build GRCh38
rs549694696 Chr.1: 212365465-212365465 on Build GRCh38
rs750389531 Chr.1: 212365220-212365220 on Build GRCh38
rs12087727 Chr.1: 212365230-212365230 on Build GRCh38
rs930541806 Chr.1: 212365415-212365415 on Build GRCh38
rs765414938 Chr.1: 212365197-212365197 on Build GRCh38
rs921879945 Chr.1: 212365404-212365404 on Build GRCh38
rs747285739 Chr.1: 212365261-212365261 on Build GRCh38
rs747838186 Chr.1: 212365406-212365406 on Build GRCh38
rs570379933 Chr.1: 212365430-212365430 on Build GRCh38
rs764118974 Chr.1: 212365426-212365427 on Build GRCh38
rs763168765 Chr.1: 212365326-212365326 on Build GRCh38
rs148556253 Chr.1: 212365178-212365178 on Build GRCh38
rs753303951 Chr.1: 212365385-212365385 on Build GRCh38
rs907691084 Chr.1: 212365439-212365439 on Build GRCh38
rs1043652726 Chr.1: 212365500-212365500 on Build GRCh38
rs376667767 Chr.1: 212365555-212365555 on Build GRCh38
rs761267565 Chr.1: 212365399-212365400 on Build GRCh38
rs12127560 Chr.1: 212365557-212365557 on Build GRCh38
rs527819365 Chr.1: 212365487-212365487 on Build GRCh38
rs748844764 Chr.1: 212365231-212365231 on Build GRCh38
rs774109526 Chr.1: 212365257-212365262 on Build GRCh38
rs955308646 Chr.1: 212365480-212365480 on Build GRCh38
rs1014305257 Chr.1: 212365556-212365556 on Build GRCh38
rs201074648 Chr.1: 212365429-212365429 on Build GRCh38
rs985067664 Chr.1: 212365430-212365438 on Build GRCh38
rs768870660 Chr.1: 212365428-212365428 on Build GRCh38
rs986438846 Chr.1: 212365486-212365486 on Build GRCh38
rs767350301 Chr.1: 212365347-212365347 on Build GRCh38
rs746270019 Chr.1: 212365419-212365419 on Build GRCh38
rs903667702 Chr.1: 212365227-212365227 on Build GRCh38
rs964082401 Chr.1: 212365623-212365623 on Build GRCh38
rs746212824 Chr.1: 212365259-212365262 on Build GRCh38
rs777212888 Chr.1: 212365192-212365192 on Build GRCh38
rs945049782 Chr.1: 212365495-212365495 on Build GRCh38
rs768265470 Chr.1: 212365236-212365236 on Build GRCh38
rs772524769 Chr.1: 212365298-212365298 on Build GRCh38
rs761969875 Chr.1: 212365428-212365429 on Build GRCh38
rs775840505 Chr.1: 212365384-212365385 on Build GRCh38
rs947715250 Chr.1: 212365497-212365497 on Build GRCh38
rs776931830 Chr.1: 212365276-212365276 on Build GRCh38
rs771335748 Chr.1: 212365272-212365272 on Build GRCh38
rs201833200 Chr.1: 212365387-212365387 on Build GRCh38
rs986897990 Chr.1: 212365285-212365285 on Build GRCh38
rs775691932 Chr.1: 212365316-212365316 on Build GRCh38
rs945290705 Chr.1: 212365202-212365202 on Build GRCh38
rs894589240 Chr.1: 212365291-212365291 on Build GRCh38
rs761140523 Chr.1: 212365175-212365175 on Build GRCh38
rs371580121 Chr.1: 212365417-212365417 on Build GRCh38
rs796456839 Chr.1: 212365428-212365429 on Build GRCh38
rs12138533 Chr.1: 212365581-212365581 on Build GRCh38
rs750242868 Chr.1: 212365353-212365353 on Build GRCh38
rs374301527 Chr.1: 212365381-212365381 on Build GRCh38
rs896213974 Chr.1: 212365493-212365493 on Build GRCh38
rs937854833 Chr.1: 212365516-212365516 on Build GRCh38
rs756023068 Chr.1: 212365375-212365375 on Build GRCh38
rs112629390 Chr.1: 212365613-212365613 on Build GRCh38
rs376261747 Chr.1: 212365203-212365203 on Build GRCh38
rs771692716 Chr.1: 212365414-212365414 on Build GRCh38
rs962903436 Chr.1: 212365461-212365461 on Build GRCh38
rs774787040 Chr.1: 212365573-212365573 on Build GRCh38
rs200795875 Chr.1: 212365331-212365331 on Build GRCh38
rs776174126 Chr.1: 212365425-212365425 on Build GRCh38
rs371543331 Chr.1: 212365332-212365332 on Build GRCh38
rs770396135 Chr.1: 212365422-212365422 on Build GRCh38
rs896161480 Chr.1: 212365186-212365186 on Build GRCh38
rs1050752074 Chr.1: 212365191-212365191 on Build GRCh38
rs760087895 Chr.1: 212365286-212365286 on Build GRCh38
rs374085267 Chr.1: 212365206-212365206 on Build GRCh38

View Full Product Details