Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 96341232-96341806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAML2
Entrez Gene ID
84441
Gene Name
mastermind like transcriptional coactivator 2
Gene Aliases
MAM-3, MAM2, MAM3, MLL-MAML2
UniGene
Hs.428214
Gene Chromosome Location
Chr.11: 95976593-96343245 on Build GRCh38
Transcripts
NM_032427.3 XM_011543024.2 XM_011543025.1 NR_036125.1

Gene Symbol
MIR1260B
Entrez Gene ID
100422991
Gene Name
microRNA 1260b
Gene Aliases
mir-1260b
UniGene
Gene Chromosome Location
Chr.11: 96341438-96341526 on Build GRCh38
Transcripts
NM_032427.3 XM_011543024.2 XM_011543025.1 NR_036125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467611 c.122G>A p.R41Q Chr.11: 96341774-96341774 on Build GRCh38
4497623 c.500C>T p.S167L Chr.11: 96341396-96341396 on Build GRCh38
7265902 c.321G>A p.P107P Chr.11: 96341575-96341575 on Build GRCh38
5763005 c.320C>T p.P107L Chr.11: 96341576-96341576 on Build GRCh38
5506379 c.465G>A p.Q155Q Chr.11: 96341431-96341431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531769026 Chr.11: 96341287-96341287 on Build GRCh38
rs751345099 Chr.11: 96341617-96341618 on Build GRCh38
rs984534521 Chr.11: 96341365-96341365 on Build GRCh38
rs576626011 Chr.11: 96341722-96341722 on Build GRCh38
rs964420056 Chr.11: 96341582-96341582 on Build GRCh38
rs984648906 Chr.11: 96341541-96341541 on Build GRCh38
rs750224740 Chr.11: 96341549-96341549 on Build GRCh38
rs1039885238 Chr.11: 96341449-96341449 on Build GRCh38
rs565979295 Chr.11: 96341577-96341577 on Build GRCh38
rs1003195565 Chr.11: 96341519-96341519 on Build GRCh38
rs950510736 Chr.11: 96341528-96341528 on Build GRCh38
rs1009976167 Chr.11: 96341520-96341520 on Build GRCh38
rs372484046 Chr.11: 96341726-96341726 on Build GRCh38
rs768173637 Chr.11: 96341709-96341709 on Build GRCh38
rs539799372 Chr.11: 96341707-96341707 on Build GRCh38
rs950611458 Chr.11: 96341340-96341340 on Build GRCh38
rs1030079534 Chr.11: 96341507-96341507 on Build GRCh38
rs764279104 Chr.11: 96341479-96341479 on Build GRCh38
rs971696804 Chr.11: 96341283-96341283 on Build GRCh38
rs776275951 Chr.11: 96341527-96341527 on Build GRCh38
rs142270768 Chr.11: 96341607-96341608 on Build GRCh38
rs374801746 Chr.11: 96341746-96341746 on Build GRCh38
rs988707190 Chr.11: 96341308-96341308 on Build GRCh38
rs781515806 Chr.11: 96341656-96341656 on Build GRCh38
rs371771349 Chr.11: 96341443-96341443 on Build GRCh38
rs752999878 Chr.11: 96341677-96341677 on Build GRCh38
rs372626208 Chr.11: 96341646-96341646 on Build GRCh38
rs757572949 Chr.11: 96341773-96341773 on Build GRCh38
rs764292190 Chr.11: 96341761-96341761 on Build GRCh38
rs367937959 Chr.11: 96341439-96341439 on Build GRCh38
rs1047342515 Chr.11: 96341425-96341425 on Build GRCh38
rs978546058 Chr.11: 96341344-96341344 on Build GRCh38
rs747767344 Chr.11: 96341714-96341714 on Build GRCh38
rs978578739 Chr.11: 96341611-96341611 on Build GRCh38
rs930226525 Chr.11: 96341392-96341392 on Build GRCh38
rs117349218 Chr.11: 96341369-96341369 on Build GRCh38
rs933040683 Chr.11: 96341668-96341668 on Build GRCh38
rs369363538 Chr.11: 96341753-96341753 on Build GRCh38
rs754161824 Chr.11: 96341767-96341767 on Build GRCh38
rs868783900 Chr.11: 96341735-96341735 on Build GRCh38
rs902678649 Chr.11: 96341442-96341442 on Build GRCh38
rs768228590 Chr.11: 96341515-96341515 on Build GRCh38
rs749676362 Chr.11: 96341687-96341687 on Build GRCh38
rs778107186 Chr.11: 96341437-96341437 on Build GRCh38
rs762012524 Chr.11: 96341407-96341407 on Build GRCh38
rs761543021 Chr.11: 96341538-96341538 on Build GRCh38
rs756348917 Chr.11: 96341754-96341754 on Build GRCh38
rs913065852 Chr.11: 96341311-96341311 on Build GRCh38
rs1057217606 Chr.11: 96341363-96341363 on Build GRCh38
rs766283585 Chr.11: 96341728-96341728 on Build GRCh38
rs778179629 Chr.11: 96341686-96341686 on Build GRCh38
rs779395127 Chr.11: 96341698-96341698 on Build GRCh38
rs781570943 Chr.11: 96341384-96341384 on Build GRCh38
rs547474756 Chr.11: 96341474-96341474 on Build GRCh38
rs922966717 Chr.11: 96341398-96341398 on Build GRCh38
rs751735419 Chr.11: 96341358-96341358 on Build GRCh38
rs771462313 Chr.11: 96341693-96341693 on Build GRCh38
rs11021541 Chr.11: 96341713-96341713 on Build GRCh38
rs766378089 Chr.11: 96341353-96341353 on Build GRCh38
rs568215293 Chr.11: 96341429-96341429 on Build GRCh38
rs748466064 Chr.11: 96341424-96341424 on Build GRCh38
rs375418213 Chr.11: 96341706-96341706 on Build GRCh38
rs34913856 Chr.11: 96341485-96341485 on Build GRCh38
rs992555033 Chr.11: 96341712-96341712 on Build GRCh38
rs774452766 Chr.11: 96341740-96341740 on Build GRCh38
rs773045051 Chr.11: 96341725-96341725 on Build GRCh38
rs1026268005 Chr.11: 96341523-96341523 on Build GRCh38
rs539622314 Chr.11: 96341587-96341587 on Build GRCh38
rs1043014041 Chr.11: 96341676-96341676 on Build GRCh38
rs981384325 Chr.11: 96341322-96341322 on Build GRCh38
rs779339782 Chr.11: 96341780-96341780 on Build GRCh38
rs755124554 Chr.11: 96341654-96341654 on Build GRCh38
rs970760102 Chr.11: 96341513-96341513 on Build GRCh38
rs199840764 Chr.11: 96341555-96341555 on Build GRCh38
rs751624667 Chr.11: 96341645-96341645 on Build GRCh38
rs756335282 Chr.11: 96341679-96341679 on Build GRCh38
rs376355064 Chr.11: 96341466-96341466 on Build GRCh38
rs1038698345 Chr.11: 96341393-96341393 on Build GRCh38
rs998258087 Chr.11: 96341494-96341494 on Build GRCh38
rs968560499 Chr.11: 96341320-96341320 on Build GRCh38
rs892433964 Chr.11: 96341511-96341511 on Build GRCh38
rs537072178 Chr.11: 96341751-96341751 on Build GRCh38
rs919050540 Chr.11: 96341325-96341325 on Build GRCh38
rs982839152 Chr.11: 96341749-96341749 on Build GRCh38

View Full Product Details