Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82993734-82994296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB30
Entrez Gene ID
27314
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.40758
Gene Chromosome Location
Chr.11: 82973133-83071923 on Build GRCh38
Transcripts
NM_001286059.1 NM_001286060.1 NM_001286061.1 NM_014488.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
246925 c.101T>C p.F34S Chr.11: 82994115-82994115 on Build GRCh38
6134872 c.107C>T p.P36L Chr.11: 82994109-82994109 on Build GRCh38
6134873 c.120C>T p.A40A Chr.11: 82994096-82994096 on Build GRCh38
4457311 c.103C>T p.P35S Chr.11: 82994113-82994113 on Build GRCh38
7426895 c.119C>A p.A40D Chr.11: 82994097-82994097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763492156 Chr.11: 82994192-82994192 on Build GRCh38
rs377704020 Chr.11: 82994033-82994033 on Build GRCh38
rs753144915 Chr.11: 82994139-82994139 on Build GRCh38
rs200470465 Chr.11: 82993925-82993925 on Build GRCh38
rs979910825 Chr.11: 82993909-82993909 on Build GRCh38
rs142067995 Chr.11: 82993831-82993831 on Build GRCh38
rs960650066 Chr.11: 82994210-82994210 on Build GRCh38
rs755846589 Chr.11: 82994125-82994125 on Build GRCh38
rs533899747 Chr.11: 82993893-82993893 on Build GRCh38
rs936905386 Chr.11: 82994155-82994155 on Build GRCh38
rs1001058260 Chr.11: 82993764-82993764 on Build GRCh38
rs954725896 Chr.11: 82993807-82993807 on Build GRCh38
rs73508492 Chr.11: 82994212-82994212 on Build GRCh38
rs764271950 Chr.11: 82993960-82993960 on Build GRCh38
rs746082999 Chr.11: 82994000-82994000 on Build GRCh38
rs912828513 Chr.11: 82993950-82993952 on Build GRCh38
rs751860799 Chr.11: 82993988-82993988 on Build GRCh38
rs116840426 Chr.11: 82993955-82993955 on Build GRCh38
rs966842762 Chr.11: 82993857-82993857 on Build GRCh38
rs1033465367 Chr.11: 82994209-82994209 on Build GRCh38
rs114610273 Chr.11: 82993803-82993803 on Build GRCh38
rs761290275 Chr.11: 82994077-82994077 on Build GRCh38
rs202153794 Chr.11: 82994140-82994140 on Build GRCh38
rs944281939 Chr.11: 82993954-82993954 on Build GRCh38
rs900677197 Chr.11: 82994174-82994174 on Build GRCh38
rs767052105 Chr.11: 82994103-82994103 on Build GRCh38
rs1002000780 Chr.11: 82994204-82994204 on Build GRCh38
rs980544481 Chr.11: 82993774-82993774 on Build GRCh38
rs762325396 Chr.11: 82993920-82993920 on Build GRCh38
rs75529202 Chr.11: 82993829-82993829 on Build GRCh38
rs766081317 Chr.11: 82994128-82994128 on Build GRCh38
rs777148634 Chr.11: 82993918-82993918 on Build GRCh38
rs890498292 Chr.11: 82994262-82994262 on Build GRCh38
rs3793986 Chr.11: 82993925-82993925 on Build GRCh38
rs961568815 Chr.11: 82993880-82993880 on Build GRCh38
rs372247286 Chr.11: 82994020-82994020 on Build GRCh38
rs774327481 Chr.11: 82994066-82994066 on Build GRCh38
rs758804331 Chr.11: 82994165-82994165 on Build GRCh38
rs778193070 Chr.11: 82994168-82994168 on Build GRCh38
rs3793987 Chr.11: 82993926-82993926 on Build GRCh38
rs900624992 Chr.11: 82993782-82993782 on Build GRCh38
rs1049299523 Chr.11: 82994028-82994028 on Build GRCh38
rs763982612 Chr.11: 82994065-82994065 on Build GRCh38
rs749838638 Chr.11: 82994120-82994120 on Build GRCh38
rs539237929 Chr.11: 82994259-82994259 on Build GRCh38
rs1032443889 Chr.11: 82993769-82993769 on Build GRCh38
rs564731397 Chr.11: 82993795-82993795 on Build GRCh38
rs757445685 Chr.11: 82994163-82994163 on Build GRCh38
rs71465967 Chr.11: 82993925-82993926 on Build GRCh38
rs776775510 Chr.11: 82993989-82993989 on Build GRCh38
rs996251324 Chr.11: 82994199-82994199 on Build GRCh38

View Full Product Details