Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55467617-55468195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT5A
Entrez Gene ID
7474
Gene Name
Wnt family member 5A
Gene Aliases
hWNT5A
UniGene
Hs.643085
Gene Chromosome Location
Chr.3: 55465715-55505261 on Build GRCh38
Transcripts
NM_001256105.1 NM_003392.4 XM_006713324.1 XM_011534085.2 XM_011534086.2 XM_011534087.2 XM_011534088.2 XM_011534089.1 XM_017007127.1 XM_017007128.1

SNP Detail

SNP ID Chromosome Location
rs931353777 Chr.3: 55468001-55468001 on Build GRCh38
rs1055405273 Chr.3: 55467876-55467876 on Build GRCh38
rs139339191 Chr.3: 55468082-55468082 on Build GRCh38
rs1045433709 Chr.3: 55467897-55467897 on Build GRCh38
rs537179179 Chr.3: 55467672-55467672 on Build GRCh38
rs555168456 Chr.3: 55467683-55467683 on Build GRCh38
rs796958500 Chr.3: 55468139-55468140 on Build GRCh38
rs942263944 Chr.3: 55467785-55467785 on Build GRCh38
rs1056594 Chr.3: 55467917-55467917 on Build GRCh38
rs909397312 Chr.3: 55467773-55467773 on Build GRCh38
rs779212131 Chr.3: 55467905-55467905 on Build GRCh38
rs998654671 Chr.3: 55467654-55467654 on Build GRCh38
rs568443423 Chr.3: 55468040-55468040 on Build GRCh38
rs1020540032 Chr.3: 55467882-55467882 on Build GRCh38
rs78756487 Chr.3: 55468153-55468156 on Build GRCh38
rs973620519 Chr.3: 55467969-55467969 on Build GRCh38
rs900105854 Chr.3: 55468082-55468085 on Build GRCh38
rs1030309580 Chr.3: 55468139-55468139 on Build GRCh38
rs892997125 Chr.3: 55467992-55467992 on Build GRCh38
rs3178762 Chr.3: 55467889-55467889 on Build GRCh38
rs923326217 Chr.3: 55467726-55467726 on Build GRCh38
rs970354232 Chr.3: 55468152-55468153 on Build GRCh38
rs115634773 Chr.3: 55468081-55468081 on Build GRCh38
rs893978730 Chr.3: 55468083-55468083 on Build GRCh38
rs566637224 Chr.3: 55467931-55467931 on Build GRCh38
rs778251605 Chr.3: 55467693-55467693 on Build GRCh38
rs397795589 Chr.3: 55467816-55467817 on Build GRCh38
rs878998603 Chr.3: 55468139-55468139 on Build GRCh38
rs758540786 Chr.3: 55467699-55467699 on Build GRCh38
rs878952703 Chr.3: 55468140-55468140 on Build GRCh38
rs569536036 Chr.3: 55467638-55467638 on Build GRCh38
rs920453279 Chr.3: 55467976-55467976 on Build GRCh38
rs374485466 Chr.3: 55467892-55467892 on Build GRCh38
rs748180463 Chr.3: 55467676-55467676 on Build GRCh38
rs375205756 Chr.3: 55467936-55467936 on Build GRCh38
rs757717433 Chr.3: 55467879-55467879 on Build GRCh38
rs368581342 Chr.3: 55468138-55468139 on Build GRCh38
rs546844099 Chr.3: 55467913-55467913 on Build GRCh38
rs990095606 Chr.3: 55467675-55467675 on Build GRCh38
rs929398150 Chr.3: 55467732-55467732 on Build GRCh38
rs997449780 Chr.3: 55467769-55467769 on Build GRCh38
rs1056613 Chr.3: 55467659-55467659 on Build GRCh38
rs1031504779 Chr.3: 55467666-55467666 on Build GRCh38
rs537104308 Chr.3: 55468057-55468057 on Build GRCh38
rs58034285 Chr.3: 55467736-55467736 on Build GRCh38
rs1020695614 Chr.3: 55468150-55468151 on Build GRCh38
rs11544761 Chr.3: 55467966-55467966 on Build GRCh38
rs976018024 Chr.3: 55468105-55468105 on Build GRCh38
rs957492241 Chr.3: 55467673-55467673 on Build GRCh38
rs772032186 Chr.3: 55468039-55468039 on Build GRCh38
rs771112286 Chr.3: 55468082-55468083 on Build GRCh38
rs764660661 Chr.3: 55467802-55467803 on Build GRCh38
rs114045392 Chr.3: 55467741-55467741 on Build GRCh38

View Full Product Details