Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26791763-26792350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPA
Entrez Gene ID
1058
Gene Name
centromere protein A
Gene Aliases
CENP-A, CenH3
UniGene
Hs.1594
Gene Chromosome Location
Chr.2: 26786014-26794589 on Build GRCh38
Transcripts
NM_001042426.1 NM_001809.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6626476 c.205C>T p.R69C Chr.2: 26792235-26792235 on Build GRCh38
6980352 c.128G>T p.R43L Chr.2: 26792158-26792158 on Build GRCh38
6932421 c.154C>T p.R52* Chr.2: 26792184-26792184 on Build GRCh38
6626478 c.172A>G p.T58A Chr.2: 26792202-26792202 on Build GRCh38
720690 c.150G>C p.E50D Chr.2: 26792180-26792180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901645370 Chr.2: 26791981-26791982 on Build GRCh38
rs559699683 Chr.2: 26792088-26792088 on Build GRCh38
rs987552696 Chr.2: 26791927-26791927 on Build GRCh38
rs1031103638 Chr.2: 26792228-26792228 on Build GRCh38
rs766046974 Chr.2: 26792271-26792271 on Build GRCh38
rs1044124753 Chr.2: 26791991-26791991 on Build GRCh38
rs113442357 Chr.2: 26792210-26792210 on Build GRCh38
rs914149139 Chr.2: 26791914-26791916 on Build GRCh38
rs553922223 Chr.2: 26792289-26792289 on Build GRCh38
rs780899209 Chr.2: 26792162-26792162 on Build GRCh38
rs141275453 Chr.2: 26792133-26792133 on Build GRCh38
rs373697581 Chr.2: 26792176-26792176 on Build GRCh38
rs905510068 Chr.2: 26791992-26791992 on Build GRCh38
rs780148367 Chr.2: 26792138-26792138 on Build GRCh38
rs761409036 Chr.2: 26792111-26792111 on Build GRCh38
rs980922465 Chr.2: 26791800-26791800 on Build GRCh38
rs988650637 Chr.2: 26792238-26792238 on Build GRCh38
rs549906943 Chr.2: 26792188-26792188 on Build GRCh38
rs35344721 Chr.2: 26792273-26792274 on Build GRCh38
rs751059407 Chr.2: 26792273-26792273 on Build GRCh38
rs912411461 Chr.2: 26791945-26791945 on Build GRCh38
rs749972073 Chr.2: 26792122-26792122 on Build GRCh38
rs151098324 Chr.2: 26792185-26792185 on Build GRCh38
rs771164020 Chr.2: 26792086-26792086 on Build GRCh38
rs202063570 Chr.2: 26792155-26792155 on Build GRCh38
rs112122856 Chr.2: 26791917-26791917 on Build GRCh38
rs761238808 Chr.2: 26792094-26792094 on Build GRCh38
rs754912844 Chr.2: 26792274-26792274 on Build GRCh38
rs761484514 Chr.2: 26792320-26792320 on Build GRCh38
rs914494853 Chr.2: 26791967-26791967 on Build GRCh38
rs879559709 Chr.2: 26792172-26792172 on Build GRCh38
rs368168633 Chr.2: 26792222-26792222 on Build GRCh38
rs1052649901 Chr.2: 26791838-26791838 on Build GRCh38
rs12991888 Chr.2: 26792290-26792290 on Build GRCh38
rs772490526 Chr.2: 26791954-26791954 on Build GRCh38
rs762692797 Chr.2: 26792266-26792266 on Build GRCh38
rs889190640 Chr.2: 26791964-26791964 on Build GRCh38
rs886166901 Chr.2: 26792034-26792034 on Build GRCh38
rs749533227 Chr.2: 26792085-26792085 on Build GRCh38
rs993297228 Chr.2: 26791984-26791984 on Build GRCh38
rs371483224 Chr.2: 26792249-26792249 on Build GRCh38
rs954996658 Chr.2: 26791852-26791852 on Build GRCh38
rs764422803 Chr.2: 26792098-26792098 on Build GRCh38
rs150238806 Chr.2: 26792241-26792241 on Build GRCh38
rs887731908 Chr.2: 26792062-26792062 on Build GRCh38
rs974724723 Chr.2: 26791805-26791805 on Build GRCh38
rs12992046 Chr.2: 26792112-26792112 on Build GRCh38
rs77738261 Chr.2: 26791957-26791957 on Build GRCh38
rs145089579 Chr.2: 26791823-26791823 on Build GRCh38
rs369992100 Chr.2: 26792227-26792227 on Build GRCh38
rs13014800 Chr.2: 26792250-26792250 on Build GRCh38
rs564257882 Chr.2: 26792189-26792189 on Build GRCh38
rs778053298 Chr.2: 26792272-26792273 on Build GRCh38
rs754206711 Chr.2: 26792103-26792103 on Build GRCh38
rs762695876 Chr.2: 26791928-26791931 on Build GRCh38
rs189677211 Chr.2: 26791960-26791960 on Build GRCh38
rs529917343 Chr.2: 26792089-26792089 on Build GRCh38
rs201807511 Chr.2: 26792284-26792284 on Build GRCh38
rs111376909 Chr.2: 26792307-26792307 on Build GRCh38
rs781144905 Chr.2: 26792277-26792277 on Build GRCh38
rs980003296 Chr.2: 26792281-26792281 on Build GRCh38
rs771173342 Chr.2: 26792183-26792183 on Build GRCh38
rs542164441 Chr.2: 26791836-26791836 on Build GRCh38
rs776697460 Chr.2: 26792184-26792184 on Build GRCh38
rs138973443 Chr.2: 26791931-26791931 on Build GRCh38
rs1040402166 Chr.2: 26792268-26792268 on Build GRCh38
rs754932895 Chr.2: 26792157-26792157 on Build GRCh38
rs1056896413 Chr.2: 26792020-26792020 on Build GRCh38
rs921927726 Chr.2: 26791811-26791811 on Build GRCh38
rs963559947 Chr.2: 26791799-26791799 on Build GRCh38
rs751521612 Chr.2: 26792154-26792154 on Build GRCh38
rs143726653 Chr.2: 26792017-26792017 on Build GRCh38
rs397782204 Chr.2: 26792275-26792276 on Build GRCh38
rs915661186 Chr.2: 26791789-26791789 on Build GRCh38
rs769172129 Chr.2: 26792232-26792232 on Build GRCh38
rs549362021 Chr.2: 26792090-26792090 on Build GRCh38
rs887689782 Chr.2: 26792317-26792317 on Build GRCh38
rs368264561 Chr.2: 26792257-26792257 on Build GRCh38
rs956965320 Chr.2: 26792235-26792235 on Build GRCh38
rs761132908 Chr.2: 26792205-26792205 on Build GRCh38
rs140589208 Chr.2: 26791971-26791971 on Build GRCh38

View Full Product Details