Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48787356-48787935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL6
Entrez Gene ID
284076
Gene Name
tubulin tyrosine ligase like 6
Gene Aliases
TTL.6
UniGene
Hs.91930
Gene Chromosome Location
Chr.17: 48762231-48817253 on Build GRCh38
Transcripts
NM_001130918.1 NM_173623.3 XM_017024491.1 XM_017024492.1 XM_017024493.1 XM_017024494.1 XM_017024495.1 XM_017024496.1 XM_017024497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518787 c.1429C>T p.R477W Chr.17: 48787827-48787827 on Build GRCh38
5386604 c.1377G>A p.K459K Chr.17: 48787879-48787879 on Build GRCh38
3819878 c.561C>G p.F187L Chr.17: 48787873-48787873 on Build GRCh38
3795741 c.650C>A p.A217D Chr.17: 48787829-48787829 on Build GRCh38
1493940 c.1445G>A p.R482Q Chr.17: 48787811-48787811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200150437 Chr.17: 48787855-48787855 on Build GRCh38
rs149748911 Chr.17: 48787894-48787894 on Build GRCh38
rs778570529 Chr.17: 48787898-48787898 on Build GRCh38
rs930175877 Chr.17: 48787869-48787869 on Build GRCh38
rs777175795 Chr.17: 48787903-48787903 on Build GRCh38
rs867632237 Chr.17: 48787849-48787849 on Build GRCh38
rs374777398 Chr.17: 48787811-48787811 on Build GRCh38
rs117729065 Chr.17: 48787797-48787797 on Build GRCh38
rs944509530 Chr.17: 48787481-48787481 on Build GRCh38
rs1005746024 Chr.17: 48787625-48787625 on Build GRCh38
rs866038514 Chr.17: 48787909-48787910 on Build GRCh38
rs752744799 Chr.17: 48787785-48787785 on Build GRCh38
rs746261714 Chr.17: 48787914-48787914 on Build GRCh38
rs916381006 Chr.17: 48787753-48787753 on Build GRCh38
rs956583940 Chr.17: 48787779-48787779 on Build GRCh38
rs146000914 Chr.17: 48787825-48787825 on Build GRCh38
rs1051042146 Chr.17: 48787872-48787872 on Build GRCh38
rs953933769 Chr.17: 48787459-48787459 on Build GRCh38
rs576033608 Chr.17: 48787429-48787429 on Build GRCh38
rs961049327 Chr.17: 48787675-48787675 on Build GRCh38
rs371709239 Chr.17: 48787453-48787453 on Build GRCh38
rs994706114 Chr.17: 48787547-48787547 on Build GRCh38
rs949512120 Chr.17: 48787758-48787759 on Build GRCh38
rs779122684 Chr.17: 48787839-48787839 on Build GRCh38
rs1054099855 Chr.17: 48787725-48787725 on Build GRCh38
rs764214963 Chr.17: 48787764-48787765 on Build GRCh38
rs920931663 Chr.17: 48787716-48787716 on Build GRCh38
rs1040517975 Chr.17: 48787500-48787500 on Build GRCh38
rs775166066 Chr.17: 48787399-48787399 on Build GRCh38
rs200455261 Chr.17: 48787808-48787808 on Build GRCh38
rs541096545 Chr.17: 48787440-48787440 on Build GRCh38
rs952600208 Chr.17: 48787709-48787709 on Build GRCh38
rs544138652 Chr.17: 48787731-48787731 on Build GRCh38
rs888764204 Chr.17: 48787479-48787479 on Build GRCh38
rs139949113 Chr.17: 48787826-48787826 on Build GRCh38
rs747621806 Chr.17: 48787902-48787902 on Build GRCh38
rs762948444 Chr.17: 48787425-48787425 on Build GRCh38
rs765461200 Chr.17: 48787782-48787782 on Build GRCh38
rs1007945673 Chr.17: 48787504-48787504 on Build GRCh38
rs558547845 Chr.17: 48787436-48787436 on Build GRCh38
rs186877051 Chr.17: 48787721-48787721 on Build GRCh38
rs990654254 Chr.17: 48787733-48787733 on Build GRCh38
rs544680520 Chr.17: 48787435-48787435 on Build GRCh38
rs748786433 Chr.17: 48787812-48787812 on Build GRCh38
rs763013800 Chr.17: 48787844-48787844 on Build GRCh38
rs140617905 Chr.17: 48787809-48787809 on Build GRCh38
rs901348365 Chr.17: 48787511-48787511 on Build GRCh38
rs528586299 Chr.17: 48787888-48787888 on Build GRCh38
rs748446224 Chr.17: 48787887-48787887 on Build GRCh38
rs777169014 Chr.17: 48787827-48787827 on Build GRCh38
rs561491865 Chr.17: 48787450-48787450 on Build GRCh38
rs751568509 Chr.17: 48787864-48787864 on Build GRCh38
rs754618995 Chr.17: 48787897-48787897 on Build GRCh38
rs919870802 Chr.17: 48787687-48787687 on Build GRCh38
rs777731701 Chr.17: 48787786-48787786 on Build GRCh38
rs764133792 Chr.17: 48787617-48787617 on Build GRCh38
rs751344443 Chr.17: 48787645-48787645 on Build GRCh38
rs781055455 Chr.17: 48787881-48787881 on Build GRCh38
rs759897970 Chr.17: 48787833-48787833 on Build GRCh38
rs776544413 Chr.17: 48787776-48787776 on Build GRCh38
rs1035474419 Chr.17: 48787722-48787722 on Build GRCh38
rs376317719 Chr.17: 48787895-48787895 on Build GRCh38
rs1003666672 Chr.17: 48787685-48787685 on Build GRCh38
rs894102722 Chr.17: 48787810-48787810 on Build GRCh38
rs773913129 Chr.17: 48787822-48787822 on Build GRCh38
rs147242441 Chr.17: 48787796-48787796 on Build GRCh38
rs935361247 Chr.17: 48787800-48787800 on Build GRCh38
rs1046836975 Chr.17: 48787387-48787387 on Build GRCh38
rs756098038 Chr.17: 48787688-48787688 on Build GRCh38
rs754197678 Chr.17: 48787772-48787772 on Build GRCh38
rs757060108 Chr.17: 48787758-48787758 on Build GRCh38
rs1019384028 Chr.17: 48787904-48787904 on Build GRCh38
rs888671742 Chr.17: 48787593-48787593 on Build GRCh38
rs761743188 Chr.17: 48787878-48787886 on Build GRCh38
rs768276232 Chr.17: 48787816-48787816 on Build GRCh38
rs969792887 Chr.17: 48787671-48787671 on Build GRCh38
rs201516203 Chr.17: 48787819-48787819 on Build GRCh38
rs759682490 Chr.17: 48787774-48787774 on Build GRCh38
rs572186208 Chr.17: 48787438-48787438 on Build GRCh38
rs765429403 Chr.17: 48787835-48787835 on Build GRCh38
rs530215412 Chr.17: 48787684-48787684 on Build GRCh38
rs752935645 Chr.17: 48787837-48787837 on Build GRCh38
rs766836734 Chr.17: 48787769-48787769 on Build GRCh38
rs757213987 Chr.17: 48787868-48787868 on Build GRCh38
rs1025873367 Chr.17: 48787553-48787553 on Build GRCh38
rs1023616153 Chr.17: 48787659-48787659 on Build GRCh38
rs780965587 Chr.17: 48787802-48787802 on Build GRCh38

View Full Product Details