Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 68838695-68839263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC102B
Entrez Gene ID
79839
Gene Name
coiled-coil domain containing 102B
Gene Aliases
ACY1L, AN, C18orf14, HsT1731
UniGene
Hs.280781
Gene Chromosome Location
Chr.18: 68715221-69214350 on Build GRCh38
Transcripts
NM_001093729.1 NM_024781.2 XM_011526174.2 XM_011526175.2 XM_017025973.1 XM_017025974.1 XM_017025975.1 XM_017025976.1 XM_017025977.1 XM_017025978.1 XM_017025979.1 XM_017025980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148815 c.625G>C p.D209H Chr.18: 68838724-68838724 on Build GRCh38
4276477 c.664G>T p.D222Y Chr.18: 68838763-68838763 on Build GRCh38
7400456 c.773C>G p.S258* Chr.18: 68838872-68838872 on Build GRCh38
1523492 c.664G>T p.D222Y Chr.18: 68838763-68838763 on Build GRCh38
5458687 c.725G>T p.R242I Chr.18: 68838824-68838824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774811363 Chr.18: 68838934-68838934 on Build GRCh38
rs766698906 Chr.18: 68838957-68838957 on Build GRCh38
rs760908062 Chr.18: 68838954-68838954 on Build GRCh38
rs369280734 Chr.18: 68838893-68838893 on Build GRCh38
rs560614390 Chr.18: 68838803-68838803 on Build GRCh38
rs773917132 Chr.18: 68839050-68839050 on Build GRCh38
rs754784468 Chr.18: 68838970-68838970 on Build GRCh38
rs751590960 Chr.18: 68838749-68838749 on Build GRCh38
rs967002875 Chr.18: 68839097-68839097 on Build GRCh38
rs746533397 Chr.18: 68838874-68838874 on Build GRCh38
rs1005382858 Chr.18: 68839068-68839077 on Build GRCh38
rs72951343 Chr.18: 68839190-68839190 on Build GRCh38
rs967704326 Chr.18: 68839205-68839205 on Build GRCh38
rs746516719 Chr.18: 68839005-68839005 on Build GRCh38
rs749475061 Chr.18: 68839047-68839048 on Build GRCh38
rs752555590 Chr.18: 68838990-68838990 on Build GRCh38
rs764595291 Chr.18: 68838739-68838739 on Build GRCh38
rs147553785 Chr.18: 68838810-68838810 on Build GRCh38
rs906351770 Chr.18: 68838944-68838944 on Build GRCh38
rs756817557 Chr.18: 68839010-68839010 on Build GRCh38
rs978922237 Chr.18: 68838983-68838983 on Build GRCh38
rs921014935 Chr.18: 68839158-68839158 on Build GRCh38
rs569241787 Chr.18: 68839068-68839068 on Build GRCh38
rs771527399 Chr.18: 68838793-68838793 on Build GRCh38
rs777096973 Chr.18: 68838872-68838872 on Build GRCh38
rs61910729 Chr.18: 68838802-68838802 on Build GRCh38
rs1035883057 Chr.18: 68839124-68839124 on Build GRCh38
rs947575629 Chr.18: 68838974-68838974 on Build GRCh38
rs12954054 Chr.18: 68838801-68838801 on Build GRCh38
rs747684869 Chr.18: 68838788-68838788 on Build GRCh38
rs772456539 Chr.18: 68839041-68839041 on Build GRCh38
rs745482917 Chr.18: 68839019-68839019 on Build GRCh38
rs912088099 Chr.18: 68839134-68839134 on Build GRCh38
rs577627837 Chr.18: 68839081-68839081 on Build GRCh38
rs748072909 Chr.18: 68838853-68838853 on Build GRCh38
rs778884474 Chr.18: 68838848-68838848 on Build GRCh38
rs375280010 Chr.18: 68838942-68838942 on Build GRCh38
rs112988548 Chr.18: 68838882-68838882 on Build GRCh38
rs144468255 Chr.18: 68838875-68838875 on Build GRCh38
rs762179508 Chr.18: 68838938-68838938 on Build GRCh38
rs749112534 Chr.18: 68838767-68838767 on Build GRCh38
rs562903183 Chr.18: 68838754-68838754 on Build GRCh38
rs754166882 Chr.18: 68838958-68838958 on Build GRCh38
rs766715174 Chr.18: 68838836-68838836 on Build GRCh38
rs911353230 Chr.18: 68838997-68838997 on Build GRCh38
rs956462855 Chr.18: 68839146-68839146 on Build GRCh38
rs768941828 Chr.18: 68838901-68838901 on Build GRCh38
rs753089174 Chr.18: 68838728-68838728 on Build GRCh38
rs376558161 Chr.18: 68838861-68838861 on Build GRCh38
rs912036329 Chr.18: 68838795-68838795 on Build GRCh38
rs750539989 Chr.18: 68838821-68838821 on Build GRCh38
rs1003663302 Chr.18: 68838992-68838992 on Build GRCh38
rs764896728 Chr.18: 68838982-68838982 on Build GRCh38
rs944866756 Chr.18: 68839138-68839138 on Build GRCh38
rs755323638 Chr.18: 68838927-68838928 on Build GRCh38
rs151064634 Chr.18: 68838751-68838751 on Build GRCh38
rs901188558 Chr.18: 68839040-68839040 on Build GRCh38
rs772886327 Chr.18: 68838796-68838796 on Build GRCh38
rs775658277 Chr.18: 68839028-68839028 on Build GRCh38
rs201839445 Chr.18: 68838746-68838746 on Build GRCh38
rs753685014 Chr.18: 68838840-68838840 on Build GRCh38
rs769509455 Chr.18: 68839021-68839021 on Build GRCh38
rs1004507767 Chr.18: 68838887-68838887 on Build GRCh38
rs756348556 Chr.18: 68838829-68838829 on Build GRCh38
rs758389400 Chr.18: 68838863-68838863 on Build GRCh38
rs778787878 Chr.18: 68838785-68838785 on Build GRCh38
rs201428626 Chr.18: 68838718-68838718 on Build GRCh38
rs989087012 Chr.18: 68839150-68839150 on Build GRCh38
rs989505645 Chr.18: 68839074-68839074 on Build GRCh38
rs535318337 Chr.18: 68839060-68839060 on Build GRCh38
rs1002981290 Chr.18: 68839112-68839112 on Build GRCh38
rs978729778 Chr.18: 68839128-68839128 on Build GRCh38
rs978299459 Chr.18: 68838760-68838760 on Build GRCh38
rs776331384 Chr.18: 68838878-68838878 on Build GRCh38
rs372234152 Chr.18: 68838939-68838939 on Build GRCh38
rs568035178 Chr.18: 68838993-68838993 on Build GRCh38
rs976510014 Chr.18: 68839155-68839155 on Build GRCh38
rs777769419 Chr.18: 68838994-68838994 on Build GRCh38
rs780799978 Chr.18: 68839017-68839017 on Build GRCh38
rs754882534 Chr.18: 68838844-68838844 on Build GRCh38
rs756438339 Chr.18: 68838758-68838758 on Build GRCh38
rs989489588 Chr.18: 68838903-68838903 on Build GRCh38
rs753388927 Chr.18: 68838750-68838750 on Build GRCh38
rs189696700 Chr.18: 68839189-68839189 on Build GRCh38
rs779090600 Chr.18: 68839024-68839024 on Build GRCh38
rs749638792 Chr.18: 68838891-68838891 on Build GRCh38
rs888363613 Chr.18: 68839046-68839046 on Build GRCh38
rs748552209 Chr.18: 68839025-68839025 on Build GRCh38
rs763399922 Chr.18: 68838730-68838730 on Build GRCh38
rs969109287 Chr.18: 68838843-68838843 on Build GRCh38
rs746621352 Chr.18: 68838797-68838797 on Build GRCh38
rs75713049 Chr.18: 68838809-68838809 on Build GRCh38
rs779947462 Chr.18: 68838766-68838766 on Build GRCh38

View Full Product Details