Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81428314-81428869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP2
Entrez Gene ID
23635
Gene Name
single stranded DNA binding protein 2
Gene Aliases
HSPC116, SOSS-B2
UniGene
Hs.102735
Gene Chromosome Location
Chr.5: 81417360-81751253 on Build GRCh38
Transcripts
NM_001256732.1 NM_001256733.1 NM_001256734.1 NM_001256735.1 NM_001256736.1 NM_012446.3 XM_011543297.2 XM_017009304.1 XM_017009305.1 XM_017009306.1 XM_017009307.1 XM_017009308.1 XM_017009309.1 XM_017009310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518594 c.1050T>C p.S350S Chr.5: 81428591-81428591 on Build GRCh38
7392059 c.994G>T p.G332C Chr.5: 81428647-81428647 on Build GRCh38
1438922 c.960T>G p.N320K Chr.5: 81428681-81428681 on Build GRCh38
3856653 c.1029T>A p.N343K Chr.5: 81428612-81428612 on Build GRCh38
3139414 c.973A>G p.M325V Chr.5: 81428668-81428668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767225673 Chr.5: 81428568-81428568 on Build GRCh38
rs372569515 Chr.5: 81428728-81428728 on Build GRCh38
rs180869620 Chr.5: 81428688-81428688 on Build GRCh38
rs750022630 Chr.5: 81428588-81428588 on Build GRCh38
rs771356145 Chr.5: 81428615-81428616 on Build GRCh38
rs190396546 Chr.5: 81428668-81428668 on Build GRCh38
rs533170020 Chr.5: 81428414-81428414 on Build GRCh38
rs1055802365 Chr.5: 81428404-81428404 on Build GRCh38
rs889352884 Chr.5: 81428419-81428419 on Build GRCh38
rs571562379 Chr.5: 81428494-81428494 on Build GRCh38
rs749707183 Chr.5: 81428560-81428560 on Build GRCh38
rs758786150 Chr.5: 81428648-81428648 on Build GRCh38
rs767105152 Chr.5: 81428631-81428631 on Build GRCh38
rs772987770 Chr.5: 81428696-81428696 on Build GRCh38
rs77097724 Chr.5: 81428437-81428437 on Build GRCh38
rs898031607 Chr.5: 81428551-81428551 on Build GRCh38
rs553405001 Chr.5: 81428752-81428752 on Build GRCh38
rs766002143 Chr.5: 81428705-81428705 on Build GRCh38
rs770889602 Chr.5: 81428544-81428544 on Build GRCh38
rs774139146 Chr.5: 81428561-81428561 on Build GRCh38
rs758710073 Chr.5: 81428479-81428479 on Build GRCh38
rs768613434 Chr.5: 81428669-81428669 on Build GRCh38
rs1005359678 Chr.5: 81428639-81428639 on Build GRCh38
rs778137542 Chr.5: 81428779-81428779 on Build GRCh38
rs1001010638 Chr.5: 81428366-81428366 on Build GRCh38
rs3764985 Chr.5: 81428753-81428753 on Build GRCh38
rs945844272 Chr.5: 81428794-81428794 on Build GRCh38
rs963557001 Chr.5: 81428662-81428662 on Build GRCh38
rs923322223 Chr.5: 81428814-81428814 on Build GRCh38
rs1013599893 Chr.5: 81428464-81428464 on Build GRCh38
rs139624975 Chr.5: 81428635-81428635 on Build GRCh38
rs753228946 Chr.5: 81428735-81428735 on Build GRCh38
rs766245221 Chr.5: 81428762-81428762 on Build GRCh38
rs200365873 Chr.5: 81428566-81428566 on Build GRCh38
rs147887538 Chr.5: 81428353-81428353 on Build GRCh38
rs976600772 Chr.5: 81428783-81428783 on Build GRCh38
rs781115896 Chr.5: 81428667-81428667 on Build GRCh38
rs538986179 Chr.5: 81428795-81428795 on Build GRCh38
rs977470008 Chr.5: 81428342-81428342 on Build GRCh38
rs746139216 Chr.5: 81428643-81428648 on Build GRCh38
rs765804037 Chr.5: 81428613-81428613 on Build GRCh38
rs753400180 Chr.5: 81428719-81428719 on Build GRCh38
rs966495607 Chr.5: 81428758-81428758 on Build GRCh38
rs763932368 Chr.5: 81428558-81428558 on Build GRCh38
rs904004567 Chr.5: 81428362-81428362 on Build GRCh38
rs760234170 Chr.5: 81428699-81428699 on Build GRCh38
rs754427857 Chr.5: 81428621-81428621 on Build GRCh38
rs759073314 Chr.5: 81428727-81428727 on Build GRCh38
rs774822614 Chr.5: 81428616-81428618 on Build GRCh38
rs774225155 Chr.5: 81428675-81428675 on Build GRCh38
rs757037545 Chr.5: 81428836-81428836 on Build GRCh38
rs149755720 Chr.5: 81428645-81428645 on Build GRCh38
rs761645168 Chr.5: 81428682-81428682 on Build GRCh38
rs771106595 Chr.5: 81428651-81428651 on Build GRCh38
rs760273824 Chr.5: 81428592-81428592 on Build GRCh38
rs762871231 Chr.5: 81428553-81428553 on Build GRCh38
rs923382752 Chr.5: 81428387-81428387 on Build GRCh38
rs747165170 Chr.5: 81428649-81428649 on Build GRCh38
rs199659509 Chr.5: 81428627-81428627 on Build GRCh38
rs774214935 Chr.5: 81428371-81428371 on Build GRCh38

View Full Product Details