Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224387633-224388217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
UniGene
Hs.497873
Gene Chromosome Location
Chr.1: 224385143-224434299 on Build GRCh38
Transcripts
NM_001115113.2 NM_025160.6 XM_005273273.4 XM_006711817.3 XM_011544281.2 XM_011544282.2 XM_017002396.1

SNP Detail

SNP ID Chromosome Location
rs955318766 Chr.1: 224388111-224388111 on Build GRCh38
rs765225378 Chr.1: 224387715-224387715 on Build GRCh38
rs531445123 Chr.1: 224387845-224387845 on Build GRCh38
rs965934565 Chr.1: 224387772-224387772 on Build GRCh38
rs1038287608 Chr.1: 224388030-224388030 on Build GRCh38
rs564971134 Chr.1: 224388031-224388031 on Build GRCh38
rs75277931 Chr.1: 224388132-224388132 on Build GRCh38
rs913229178 Chr.1: 224387860-224387861 on Build GRCh38
rs4653586 Chr.1: 224387671-224387671 on Build GRCh38
rs754261873 Chr.1: 224388037-224388037 on Build GRCh38
rs901325415 Chr.1: 224388057-224388057 on Build GRCh38
rs76591934 Chr.1: 224388038-224388038 on Build GRCh38
rs200936577 Chr.1: 224388169-224388169 on Build GRCh38
rs1018844783 Chr.1: 224388162-224388162 on Build GRCh38
rs1006663973 Chr.1: 224388140-224388140 on Build GRCh38
rs11809710 Chr.1: 224388172-224388172 on Build GRCh38
rs889456395 Chr.1: 224388023-224388023 on Build GRCh38
rs752544864 Chr.1: 224387760-224387760 on Build GRCh38
rs949260546 Chr.1: 224387974-224387974 on Build GRCh38
rs952732729 Chr.1: 224387826-224387826 on Build GRCh38
rs547406494 Chr.1: 224388080-224388080 on Build GRCh38
rs939172399 Chr.1: 224387913-224387913 on Build GRCh38
rs1034280170 Chr.1: 224388187-224388187 on Build GRCh38
rs1025889051 Chr.1: 224388074-224388074 on Build GRCh38
rs566421799 Chr.1: 224388087-224388087 on Build GRCh38
rs957561344 Chr.1: 224388166-224388166 on Build GRCh38
rs1053486784 Chr.1: 224388022-224388022 on Build GRCh38
rs1005413918 Chr.1: 224387983-224387983 on Build GRCh38
rs896785270 Chr.1: 224388036-224388036 on Build GRCh38
rs5781364 Chr.1: 224388129-224388130 on Build GRCh38
rs902153378 Chr.1: 224387707-224387707 on Build GRCh38
rs946070609 Chr.1: 224387885-224387885 on Build GRCh38
rs536897904 Chr.1: 224388165-224388165 on Build GRCh38
rs902456464 Chr.1: 224388007-224388007 on Build GRCh38
rs758552799 Chr.1: 224387800-224387800 on Build GRCh38
rs1032080551 Chr.1: 224387731-224387731 on Build GRCh38
rs1007060508 Chr.1: 224387759-224387759 on Build GRCh38
rs889573260 Chr.1: 224387741-224387741 on Build GRCh38
rs993874544 Chr.1: 224387692-224387692 on Build GRCh38
rs936313808 Chr.1: 224388019-224388019 on Build GRCh38
rs560872686 Chr.1: 224387832-224387832 on Build GRCh38
rs777664684 Chr.1: 224387821-224387821 on Build GRCh38
rs779577242 Chr.1: 224388076-224388076 on Build GRCh38
rs1042288376 Chr.1: 224387989-224387989 on Build GRCh38
rs994775432 Chr.1: 224388065-224388065 on Build GRCh38
rs990428427 Chr.1: 224387777-224387777 on Build GRCh38
rs548673738 Chr.1: 224387721-224387721 on Build GRCh38
rs917730097 Chr.1: 224387936-224387936 on Build GRCh38
rs1045777692 Chr.1: 224387892-224387892 on Build GRCh38
rs397831363 Chr.1: 224388131-224388132 on Build GRCh38
rs927379780 Chr.1: 224387879-224387879 on Build GRCh38
rs549582181 Chr.1: 224387899-224387899 on Build GRCh38
rs148513645 Chr.1: 224387921-224387922 on Build GRCh38
rs961060629 Chr.1: 224387684-224387684 on Build GRCh38
rs79556179 Chr.1: 224388129-224388129 on Build GRCh38
rs895004498 Chr.1: 224387942-224387942 on Build GRCh38
rs953117850 Chr.1: 224388163-224388163 on Build GRCh38
rs984121595 Chr.1: 224387830-224387830 on Build GRCh38
rs992880759 Chr.1: 224387930-224387930 on Build GRCh38

View Full Product Details