Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10045716-10046256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF11
Entrez Gene ID
8462
Gene Name
Kruppel like factor 11
Gene Aliases
FKLF, FKLF1, MODY7, TIEG2, Tieg3
UniGene
Hs.12229
Gene Chromosome Location
Chr.2: 10043555-10054836 on Build GRCh38
Transcripts
NM_001177716.1 NM_001177718.1 NM_003597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508538 c.97G>T p.E33* Chr.2: 10046204-10046204 on Build GRCh38
1738776 c.86G>C p.R29P Chr.2: 10046193-10046193 on Build GRCh38
2980163 c.90T>C p.H30H Chr.2: 10046197-10046197 on Build GRCh38
6696214 c.144C>T p.V48V Chr.2: 10046251-10046251 on Build GRCh38
3425044 c.97G>A p.E33K Chr.2: 10046204-10046204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927544195 Chr.2: 10045791-10045791 on Build GRCh38
rs1009834370 Chr.2: 10046019-10046019 on Build GRCh38
rs148388504 Chr.2: 10045795-10045795 on Build GRCh38
rs905725819 Chr.2: 10045936-10045936 on Build GRCh38
rs988863410 Chr.2: 10045813-10045813 on Build GRCh38
rs746269825 Chr.2: 10045792-10045792 on Build GRCh38
rs958944278 Chr.2: 10045961-10045962 on Build GRCh38
rs759576007 Chr.2: 10046168-10046168 on Build GRCh38
rs775967362 Chr.2: 10046144-10046144 on Build GRCh38
rs867320051 Chr.2: 10046030-10046030 on Build GRCh38
rs146577005 Chr.2: 10046197-10046197 on Build GRCh38
rs1048502745 Chr.2: 10045868-10045868 on Build GRCh38
rs1024336951 Chr.2: 10046114-10046114 on Build GRCh38
rs921435747 Chr.2: 10045846-10045846 on Build GRCh38
rs767511496 Chr.2: 10046053-10046053 on Build GRCh38
rs571706616 Chr.2: 10045811-10045811 on Build GRCh38
rs976936753 Chr.2: 10045841-10045841 on Build GRCh38
rs150096859 Chr.2: 10046193-10046193 on Build GRCh38
rs780154479 Chr.2: 10046205-10046205 on Build GRCh38
rs955759413 Chr.2: 10045810-10045810 on Build GRCh38
rs932814197 Chr.2: 10045860-10045860 on Build GRCh38
rs1048810983 Chr.2: 10045852-10045852 on Build GRCh38
rs775910736 Chr.2: 10045825-10045825 on Build GRCh38
rs370032206 Chr.2: 10045789-10045789 on Build GRCh38
rs549385160 Chr.2: 10046075-10046075 on Build GRCh38
rs1009959086 Chr.2: 10046006-10046006 on Build GRCh38
rs1000772563 Chr.2: 10046062-10046062 on Build GRCh38
rs763069090 Chr.2: 10045858-10045858 on Build GRCh38
rs200783123 Chr.2: 10046044-10046044 on Build GRCh38
rs549299869 Chr.2: 10045749-10045749 on Build GRCh38
rs763280352 Chr.2: 10045873-10045873 on Build GRCh38
rs532841444 Chr.2: 10046140-10046140 on Build GRCh38
rs55734254 Chr.2: 10046072-10046072 on Build GRCh38
rs373784080 Chr.2: 10045828-10045828 on Build GRCh38
rs993275131 Chr.2: 10045970-10045970 on Build GRCh38
rs780770561 Chr.2: 10046192-10046192 on Build GRCh38
rs937350995 Chr.2: 10045794-10045794 on Build GRCh38
rs373697732 Chr.2: 10046159-10046159 on Build GRCh38
rs761932489 Chr.2: 10045895-10045895 on Build GRCh38
rs984714588 Chr.2: 10046093-10046093 on Build GRCh38
rs956000258 Chr.2: 10045995-10045995 on Build GRCh38
rs750272368 Chr.2: 10046181-10046181 on Build GRCh38
rs367633966 Chr.2: 10045923-10045923 on Build GRCh38
rs965643704 Chr.2: 10046009-10046009 on Build GRCh38
rs375783438 Chr.2: 10045766-10045766 on Build GRCh38
rs561466910 Chr.2: 10046177-10046177 on Build GRCh38
rs367958764 Chr.2: 10046098-10046098 on Build GRCh38
rs940097308 Chr.2: 10045892-10045892 on Build GRCh38
rs572782762 Chr.2: 10046028-10046028 on Build GRCh38
rs556075033 Chr.2: 10045874-10045874 on Build GRCh38
rs543062014 Chr.2: 10045887-10045887 on Build GRCh38
rs576110995 Chr.2: 10045798-10045798 on Build GRCh38
rs757091205 Chr.2: 10046103-10046103 on Build GRCh38
rs758083789 Chr.2: 10046155-10046157 on Build GRCh38
rs778693434 Chr.2: 10046129-10046129 on Build GRCh38
rs757223238 Chr.2: 10046179-10046179 on Build GRCh38
rs940786182 Chr.2: 10045981-10045981 on Build GRCh38
rs371327904 Chr.2: 10046200-10046200 on Build GRCh38
rs774599917 Chr.2: 10045893-10045893 on Build GRCh38
rs1040721766 Chr.2: 10046012-10046012 on Build GRCh38
rs763407739 Chr.2: 10046158-10046158 on Build GRCh38
rs774529711 Chr.2: 10046213-10046213 on Build GRCh38
rs759816118 Chr.2: 10046226-10046226 on Build GRCh38
rs569501166 Chr.2: 10046090-10046090 on Build GRCh38
rs777149055 Chr.2: 10045755-10045755 on Build GRCh38
rs79237909 Chr.2: 10045797-10045797 on Build GRCh38
rs1032784338 Chr.2: 10045988-10045988 on Build GRCh38
rs965065587 Chr.2: 10045752-10045752 on Build GRCh38
rs200094318 Chr.2: 10046156-10046156 on Build GRCh38
rs190130541 Chr.2: 10045894-10045894 on Build GRCh38
rs574119447 Chr.2: 10045882-10045882 on Build GRCh38
rs904235807 Chr.2: 10046055-10046055 on Build GRCh38
rs367593984 Chr.2: 10045932-10045932 on Build GRCh38
rs56101580 Chr.2: 10046142-10046142 on Build GRCh38
rs551775297 Chr.2: 10046165-10046165 on Build GRCh38
rs754540806 Chr.2: 10046191-10046191 on Build GRCh38
rs979404172 Chr.2: 10045790-10045790 on Build GRCh38
rs1036430316 Chr.2: 10045982-10045982 on Build GRCh38
rs984401644 Chr.2: 10046210-10046210 on Build GRCh38
rs900827888 Chr.2: 10046017-10046017 on Build GRCh38
rs995463027 Chr.2: 10045849-10045849 on Build GRCh38
rs988941389 Chr.2: 10045762-10045762 on Build GRCh38
rs761869566 Chr.2: 10045980-10045980 on Build GRCh38
rs962059701 Chr.2: 10046119-10046119 on Build GRCh38
rs1048193300 Chr.2: 10045998-10045998 on Build GRCh38
rs773983272 Chr.2: 10045900-10045900 on Build GRCh38
rs779719904 Chr.2: 10046185-10046185 on Build GRCh38
rs1044335894 Chr.2: 10045933-10045933 on Build GRCh38
rs751757470 Chr.2: 10046099-10046099 on Build GRCh38
rs367566983 Chr.2: 10046175-10046175 on Build GRCh38
rs771775881 Chr.2: 10046133-10046133 on Build GRCh38
rs778816307 Chr.2: 10046180-10046180 on Build GRCh38
rs751475306 Chr.2: 10046216-10046217 on Build GRCh38
rs182632864 Chr.2: 10046024-10046024 on Build GRCh38
rs187797997 Chr.2: 10046032-10046032 on Build GRCh38
rs140643007 Chr.2: 10046188-10046188 on Build GRCh38
rs895740488 Chr.2: 10045735-10045735 on Build GRCh38
rs1034519774 Chr.2: 10046063-10046063 on Build GRCh38
rs750267236 Chr.2: 10046105-10046106 on Build GRCh38
rs767656816 Chr.2: 10046173-10046173 on Build GRCh38
rs767567762 Chr.2: 10046230-10046230 on Build GRCh38
rs771393366 Chr.2: 10046203-10046203 on Build GRCh38
rs777622001 Chr.2: 10046100-10046100 on Build GRCh38
rs371633895 Chr.2: 10046143-10046143 on Build GRCh38
rs1000015589 Chr.2: 10045983-10045983 on Build GRCh38
rs770371375 Chr.2: 10046155-10046155 on Build GRCh38
rs753844037 Chr.2: 10046101-10046101 on Build GRCh38
rs544934038 Chr.2: 10045985-10045985 on Build GRCh38
rs1054708267 Chr.2: 10046229-10046229 on Build GRCh38
rs565127878 Chr.2: 10046007-10046007 on Build GRCh38
rs1009927748 Chr.2: 10045880-10045880 on Build GRCh38
rs534339373 Chr.2: 10045827-10045827 on Build GRCh38

View Full Product Details