Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45110460-45110968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF39
Entrez Gene ID
55015
Gene Name
pre-mRNA processing factor 39
Gene Aliases
-
UniGene
Hs.274337
Gene Chromosome Location
Chr.14: 45084099-45115601 on Build GRCh38
Transcripts
NM_017922.3 NR_003691.1

Gene Symbol
SNORD127
Entrez Gene ID
100113389
Gene Name
small nucleolar RNA, C/D box 127
Gene Aliases
-
UniGene
Hs.712501
Gene Chromosome Location
Chr.14: 45110875-45110975 on Build GRCh38
Transcripts
NM_017922.3 NR_003691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6228983 c.1497C>T p.A499A Chr.14: 45110742-45110742 on Build GRCh38
1728008 c.1349G>A p.C450Y Chr.14: 45110594-45110594 on Build GRCh38
6222608 c.1378C>T p.R460* Chr.14: 45110623-45110623 on Build GRCh38
7001103 c.1164_1172delTCCAAAATC p.P389_S391delPKS Chr.14: 45110772-45110780 on Build GRCh38
1477571 c.1346A>C p.E449A Chr.14: 45110591-45110591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199930230 Chr.14: 45110623-45110623 on Build GRCh38
rs950981673 Chr.14: 45110828-45110828 on Build GRCh38
rs767948473 Chr.14: 45110682-45110682 on Build GRCh38
rs922050691 Chr.14: 45110900-45110900 on Build GRCh38
rs916934711 Chr.14: 45110891-45110891 on Build GRCh38
rs902435466 Chr.14: 45110498-45110498 on Build GRCh38
rs763674943 Chr.14: 45110715-45110715 on Build GRCh38
rs780104678 Chr.14: 45110870-45110870 on Build GRCh38
rs774672911 Chr.14: 45110657-45110657 on Build GRCh38
rs377044579 Chr.14: 45110702-45110702 on Build GRCh38
rs1053682867 Chr.14: 45110935-45110935 on Build GRCh38
rs1035779629 Chr.14: 45110766-45110766 on Build GRCh38
rs909229342 Chr.14: 45110860-45110860 on Build GRCh38
rs751756532 Chr.14: 45110918-45110918 on Build GRCh38
rs745845794 Chr.14: 45110523-45110523 on Build GRCh38
rs766400733 Chr.14: 45110645-45110645 on Build GRCh38
rs758077499 Chr.14: 45110573-45110573 on Build GRCh38
rs923739599 Chr.14: 45110930-45110930 on Build GRCh38
rs753490586 Chr.14: 45110727-45110727 on Build GRCh38
rs970664591 Chr.14: 45110679-45110679 on Build GRCh38
rs756711891 Chr.14: 45110806-45110806 on Build GRCh38
rs190135276 Chr.14: 45110611-45110611 on Build GRCh38
rs958278186 Chr.14: 45110497-45110497 on Build GRCh38
rs201073023 Chr.14: 45110672-45110672 on Build GRCh38
rs999418303 Chr.14: 45110505-45110505 on Build GRCh38
rs758356183 Chr.14: 45110852-45110852 on Build GRCh38
rs756380947 Chr.14: 45110805-45110805 on Build GRCh38
rs775917565 Chr.14: 45110683-45110683 on Build GRCh38
rs771168541 Chr.14: 45110655-45110655 on Build GRCh38
rs1018300468 Chr.14: 45110528-45110528 on Build GRCh38
rs751346648 Chr.14: 45110781-45110781 on Build GRCh38
rs547387087 Chr.14: 45110914-45110914 on Build GRCh38
rs763946616 Chr.14: 45110717-45110717 on Build GRCh38
rs949729330 Chr.14: 45110901-45110901 on Build GRCh38
rs935262427 Chr.14: 45110932-45110932 on Build GRCh38
rs185454100 Chr.14: 45110929-45110929 on Build GRCh38
rs937269616 Chr.14: 45110864-45110864 on Build GRCh38
rs75940101 Chr.14: 45110926-45110926 on Build GRCh38
rs975268513 Chr.14: 45110878-45110878 on Build GRCh38
rs1053461729 Chr.14: 45110583-45110583 on Build GRCh38
rs868229919 Chr.14: 45110571-45110571 on Build GRCh38
rs764745759 Chr.14: 45110842-45110842 on Build GRCh38
rs1007113026 Chr.14: 45110506-45110506 on Build GRCh38
rs147657088 Chr.14: 45110869-45110869 on Build GRCh38
rs201831950 Chr.14: 45110652-45110652 on Build GRCh38
rs951323282 Chr.14: 45110743-45110743 on Build GRCh38
rs749945098 Chr.14: 45110849-45110849 on Build GRCh38
rs200143415 Chr.14: 45110593-45110593 on Build GRCh38
rs182348145 Chr.14: 45110903-45110903 on Build GRCh38
rs200537096 Chr.14: 45110574-45110574 on Build GRCh38
rs566286272 Chr.14: 45110921-45110921 on Build GRCh38
rs758197072 Chr.14: 45110647-45110647 on Build GRCh38

View Full Product Details