Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88010537-88011062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR65
Entrez Gene ID
8477
Gene Name
G protein-coupled receptor 65
Gene Aliases
TDAG8, hTDAG8
UniGene
Hs.443243
Gene Chromosome Location
Chr.14: 88005124-88014811 on Build GRCh38
Transcripts
NM_003608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5382771 c.149G>A p.G50E Chr.14: 88010996-88010996 on Build GRCh38
311587 c.138A>T p.E46D Chr.14: 88010985-88010985 on Build GRCh38
5478776 c.59T>C p.V20A Chr.14: 88010906-88010906 on Build GRCh38
958398 c.61T>C p.Y21H Chr.14: 88010908-88010908 on Build GRCh38
3886459 c.85A>C p.I29L Chr.14: 88010932-88010932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983718463 Chr.14: 88010962-88010962 on Build GRCh38
rs765745388 Chr.14: 88010854-88010854 on Build GRCh38
rs142533891 Chr.14: 88011022-88011022 on Build GRCh38
rs936359205 Chr.14: 88010787-88010787 on Build GRCh38
rs975934822 Chr.14: 88010747-88010747 on Build GRCh38
rs371143409 Chr.14: 88010841-88010841 on Build GRCh38
rs771711565 Chr.14: 88010814-88010814 on Build GRCh38
rs751705970 Chr.14: 88010884-88010884 on Build GRCh38
rs757554885 Chr.14: 88010886-88010886 on Build GRCh38
rs999420180 Chr.14: 88010781-88010781 on Build GRCh38
rs567358815 Chr.14: 88010756-88010756 on Build GRCh38
rs373256942 Chr.14: 88010816-88010816 on Build GRCh38
rs767908692 Chr.14: 88010618-88010618 on Build GRCh38
rs764064545 Chr.14: 88011004-88011004 on Build GRCh38
rs1027687236 Chr.14: 88010959-88010959 on Build GRCh38
rs776921083 Chr.14: 88010957-88010957 on Build GRCh38
rs1006686906 Chr.14: 88010832-88010832 on Build GRCh38
rs923063762 Chr.14: 88010760-88010760 on Build GRCh38
rs753641390 Chr.14: 88011034-88011034 on Build GRCh38
rs952855541 Chr.14: 88010630-88010630 on Build GRCh38
rs969630643 Chr.14: 88010573-88010573 on Build GRCh38
rs1026992229 Chr.14: 88010627-88010627 on Build GRCh38
rs139430517 Chr.14: 88010963-88010963 on Build GRCh38
rs944077055 Chr.14: 88010697-88010697 on Build GRCh38
rs1055300318 Chr.14: 88010720-88010720 on Build GRCh38
rs903767390 Chr.14: 88010779-88010779 on Build GRCh38
rs775939991 Chr.14: 88010994-88010994 on Build GRCh38
rs755211561 Chr.14: 88010954-88010955 on Build GRCh38
rs939507849 Chr.14: 88010719-88010719 on Build GRCh38
rs753693308 Chr.14: 88010903-88010903 on Build GRCh38
rs144531468 Chr.14: 88010796-88010797 on Build GRCh38
rs770245308 Chr.14: 88010986-88010986 on Build GRCh38
rs189583840 Chr.14: 88010763-88010763 on Build GRCh38
rs762974956 Chr.14: 88010860-88010860 on Build GRCh38
rs529969289 Chr.14: 88010680-88010680 on Build GRCh38
rs1043407576 Chr.14: 88010771-88010771 on Build GRCh38
rs757816094 Chr.14: 88010924-88010924 on Build GRCh38
rs1047990351 Chr.14: 88010695-88010695 on Build GRCh38
rs768484621 Chr.14: 88010826-88010826 on Build GRCh38
rs556374123 Chr.14: 88010773-88010779 on Build GRCh38
rs563532635 Chr.14: 88010765-88010765 on Build GRCh38
rs773964361 Chr.14: 88010973-88010973 on Build GRCh38
rs746253097 Chr.14: 88010970-88010970 on Build GRCh38
rs868319581 Chr.14: 88010996-88010996 on Build GRCh38
rs761455094 Chr.14: 88011003-88011003 on Build GRCh38
rs1002366983 Chr.14: 88010937-88010937 on Build GRCh38
rs754908890 Chr.14: 88010906-88010906 on Build GRCh38
rs950683606 Chr.14: 88010945-88010945 on Build GRCh38
rs535833827 Chr.14: 88010762-88010762 on Build GRCh38
rs764270910 Chr.14: 88010873-88010873 on Build GRCh38
rs1013265871 Chr.14: 88010827-88010827 on Build GRCh38
rs747191304 Chr.14: 88010842-88010843 on Build GRCh38
rs777392229 Chr.14: 88010932-88010932 on Build GRCh38
rs778502960 Chr.14: 88010805-88010805 on Build GRCh38
rs920617864 Chr.14: 88010916-88010916 on Build GRCh38
rs779020059 Chr.14: 88010910-88010910 on Build GRCh38
rs747625763 Chr.14: 88010919-88010919 on Build GRCh38
rs761876928 Chr.14: 88011009-88011009 on Build GRCh38
rs766270553 Chr.14: 88010897-88010897 on Build GRCh38
rs982799149 Chr.14: 88010642-88010642 on Build GRCh38
rs532186378 Chr.14: 88010782-88010782 on Build GRCh38
rs777039575 Chr.14: 88010837-88010837 on Build GRCh38
rs1025099201 Chr.14: 88010864-88010864 on Build GRCh38
rs920588328 Chr.14: 88010665-88010665 on Build GRCh38
rs762611711 Chr.14: 88010755-88010755 on Build GRCh38
rs1052036967 Chr.14: 88010795-88010795 on Build GRCh38
rs756618557 Chr.14: 88010921-88010921 on Build GRCh38
rs771290650 Chr.14: 88010576-88010576 on Build GRCh38
rs1013734754 Chr.14: 88010556-88010556 on Build GRCh38
rs746629867 Chr.14: 88010934-88010934 on Build GRCh38
rs561177179 Chr.14: 88010624-88010624 on Build GRCh38
rs373108252 Chr.14: 88010859-88010859 on Build GRCh38
rs776868279 Chr.14: 88010742-88010742 on Build GRCh38
rs1015581588 Chr.14: 88011031-88011031 on Build GRCh38
rs1055264317 Chr.14: 88011036-88011036 on Build GRCh38
rs771179586 Chr.14: 88010836-88010836 on Build GRCh38
rs747494202 Chr.14: 88010564-88010564 on Build GRCh38
rs747678359 Chr.14: 88010808-88010808 on Build GRCh38

View Full Product Details