Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50965463-50966037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAG9
Entrez Gene ID
9043
Gene Name
sperm associated antigen 9
Gene Aliases
CT89, HLC-6, HLC4, HLC6, JIP-4, JIP4, JLP, PHET, PIG6
UniGene
Hs.463439
Gene Chromosome Location
Chr.17: 50962174-51120865 on Build GRCh38
Transcripts
NM_001130527.2 NM_001130528.2 NM_001251971.1 NM_003971.5 XM_005257768.2 XM_005257771.2 XM_005257774.3 XM_017025283.1 XM_017025284.1 XM_017025285.1

SNP Detail

SNP ID Chromosome Location
rs545041879 Chr.17: 50965756-50965760 on Build GRCh38
rs560033355 Chr.17: 50965754-50965754 on Build GRCh38
rs773806317 Chr.17: 50965804-50965804 on Build GRCh38
rs147399641 Chr.17: 50965850-50965850 on Build GRCh38
rs762656549 Chr.17: 50965636-50965636 on Build GRCh38
rs576777761 Chr.17: 50965522-50965522 on Build GRCh38
rs991618527 Chr.17: 50965879-50965879 on Build GRCh38
rs940801298 Chr.17: 50965498-50965498 on Build GRCh38
rs148040600 Chr.17: 50965968-50965968 on Build GRCh38
rs777088557 Chr.17: 50965888-50965888 on Build GRCh38
rs937552724 Chr.17: 50965505-50965505 on Build GRCh38
rs8125 Chr.17: 50965987-50965987 on Build GRCh38
rs1055303 Chr.17: 50965646-50965646 on Build GRCh38
rs987850343 Chr.17: 50965759-50965759 on Build GRCh38
rs1014914802 Chr.17: 50965772-50965772 on Build GRCh38
rs917474545 Chr.17: 50965880-50965880 on Build GRCh38
rs768314607 Chr.17: 50965650-50965650 on Build GRCh38
rs948934017 Chr.17: 50965882-50965882 on Build GRCh38
rs962378876 Chr.17: 50965789-50965789 on Build GRCh38
rs919699483 Chr.17: 50965788-50965788 on Build GRCh38
rs542435346 Chr.17: 50965910-50965910 on Build GRCh38
rs1029546899 Chr.17: 50965724-50965724 on Build GRCh38
rs896077741 Chr.17: 50965573-50965573 on Build GRCh38
rs973926677 Chr.17: 50965501-50965501 on Build GRCh38
rs899660492 Chr.17: 50965710-50965710 on Build GRCh38
rs1020798046 Chr.17: 50965959-50965959 on Build GRCh38
rs1055170374 Chr.17: 50965831-50965831 on Build GRCh38
rs920757899 Chr.17: 50965504-50965504 on Build GRCh38
rs920711265 Chr.17: 50965828-50965828 on Build GRCh38
rs1033199071 Chr.17: 50965705-50965705 on Build GRCh38
rs953916212 Chr.17: 50965642-50965642 on Build GRCh38
rs545786848 Chr.17: 50965731-50965731 on Build GRCh38
rs1028341655 Chr.17: 50965612-50965612 on Build GRCh38
rs902902549 Chr.17: 50965658-50965658 on Build GRCh38
rs779462205 Chr.17: 50965682-50965682 on Build GRCh38
rs986724841 Chr.17: 50965656-50965656 on Build GRCh38
rs866571369 Chr.17: 50965508-50965508 on Build GRCh38
rs1041400570 Chr.17: 50965655-50965655 on Build GRCh38
rs999005495 Chr.17: 50965948-50965948 on Build GRCh38
rs1055961928 Chr.17: 50965560-50965560 on Build GRCh38
rs990577725 Chr.17: 50965565-50965566 on Build GRCh38
rs773370581 Chr.17: 50965610-50965611 on Build GRCh38
rs996690926 Chr.17: 50965721-50965721 on Build GRCh38
rs907371421 Chr.17: 50965931-50965931 on Build GRCh38
rs924281672 Chr.17: 50965939-50965939 on Build GRCh38
rs1042390759 Chr.17: 50965914-50965914 on Build GRCh38

View Full Product Details