Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116252330-116252891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPYL4
Entrez Gene ID
23270
Gene Name
TSPY like 4
Gene Aliases
dJ486I3.2
UniGene
Hs.284141
Gene Chromosome Location
Chr.6: 116249964-116254098 on Build GRCh38
Transcripts
NM_021648.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3157070 c.1216G>A p.A406T Chr.6: 116252793-116252793 on Build GRCh38
5011663 c.1180G>A p.G394R Chr.6: 116252829-116252829 on Build GRCh38
277860 c.1186C>T p.R396* Chr.6: 116252823-116252823 on Build GRCh38
1072370 c.1134G>A p.W378* Chr.6: 116252875-116252875 on Build GRCh38
5749399 c.1145T>C p.L382P Chr.6: 116252864-116252864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015018666 Chr.6: 116252508-116252508 on Build GRCh38
rs1036825028 Chr.6: 116252619-116252619 on Build GRCh38
rs768466156 Chr.6: 116252736-116252737 on Build GRCh38
rs769169017 Chr.6: 116252752-116252752 on Build GRCh38
rs769125416 Chr.6: 116252782-116252782 on Build GRCh38
rs754002170 Chr.6: 116252831-116252831 on Build GRCh38
rs1046266386 Chr.6: 116252386-116252386 on Build GRCh38
rs55804829 Chr.6: 116252745-116252745 on Build GRCh38
rs183681428 Chr.6: 116252795-116252795 on Build GRCh38
rs772833530 Chr.6: 116252784-116252784 on Build GRCh38
rs962004294 Chr.6: 116252513-116252513 on Build GRCh38
rs1002423617 Chr.6: 116252439-116252439 on Build GRCh38
rs939843827 Chr.6: 116252573-116252573 on Build GRCh38
rs1045058789 Chr.6: 116252666-116252666 on Build GRCh38
rs776378293 Chr.6: 116252768-116252768 on Build GRCh38
rs747429780 Chr.6: 116252746-116252746 on Build GRCh38
rs757535016 Chr.6: 116252835-116252835 on Build GRCh38
rs905122442 Chr.6: 116252416-116252416 on Build GRCh38
rs538039455 Chr.6: 116252707-116252707 on Build GRCh38
rs368091812 Chr.6: 116252794-116252794 on Build GRCh38
rs747781152 Chr.6: 116252410-116252410 on Build GRCh38
rs779064603 Chr.6: 116252719-116252719 on Build GRCh38
rs373677929 Chr.6: 116252773-116252773 on Build GRCh38
rs555105693 Chr.6: 116252460-116252460 on Build GRCh38
rs767304423 Chr.6: 116252814-116252814 on Build GRCh38
rs975603445 Chr.6: 116252797-116252797 on Build GRCh38
rs758710823 Chr.6: 116252846-116252846 on Build GRCh38
rs976093650 Chr.6: 116252567-116252567 on Build GRCh38
rs558664499 Chr.6: 116252526-116252526 on Build GRCh38
rs995352459 Chr.6: 116252658-116252658 on Build GRCh38
rs1051048942 Chr.6: 116252523-116252523 on Build GRCh38
rs771611277 Chr.6: 116252461-116252461 on Build GRCh38
rs746349684 Chr.6: 116252721-116252721 on Build GRCh38
rs772704745 Chr.6: 116252736-116252736 on Build GRCh38
rs899264798 Chr.6: 116252652-116252652 on Build GRCh38
rs186899508 Chr.6: 116252796-116252796 on Build GRCh38
rs905233995 Chr.6: 116252692-116252692 on Build GRCh38
rs759278000 Chr.6: 116252812-116252812 on Build GRCh38
rs9387389 Chr.6: 116252504-116252504 on Build GRCh38
rs774125311 Chr.6: 116252759-116252760 on Build GRCh38
rs563441481 Chr.6: 116252861-116252861 on Build GRCh38
rs760515696 Chr.6: 116252503-116252503 on Build GRCh38
rs920928684 Chr.6: 116252829-116252829 on Build GRCh38
rs986815019 Chr.6: 116252716-116252716 on Build GRCh38
rs1009757958 Chr.6: 116252789-116252789 on Build GRCh38
rs879408721 Chr.6: 116252474-116252474 on Build GRCh38
rs1035244962 Chr.6: 116252709-116252709 on Build GRCh38
rs535606537 Chr.6: 116252454-116252454 on Build GRCh38
rs975088547 Chr.6: 116252483-116252483 on Build GRCh38
rs1030975982 Chr.6: 116252634-116252634 on Build GRCh38
rs759943043 Chr.6: 116252716-116252718 on Build GRCh38
rs915012280 Chr.6: 116252370-116252370 on Build GRCh38
rs570050445 Chr.6: 116252490-116252490 on Build GRCh38
rs2232473 Chr.6: 116252839-116252839 on Build GRCh38
rs773128551 Chr.6: 116252486-116252486 on Build GRCh38

View Full Product Details