Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37907537-37908070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFRP4
Entrez Gene ID
6424
Gene Name
secreted frizzled related protein 4
Gene Aliases
FRP-4, FRPHE, sFRP-4
UniGene
Hs.658169
Gene Chromosome Location
Chr.7: 37905932-37916923 on Build GRCh38
Transcripts
NM_003014.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736442 c.958C>A p.P320T Chr.7: 37907562-37907562 on Build GRCh38
4560913 c.861G>A p.W287* Chr.7: 37907659-37907659 on Build GRCh38
1569001 c.919C>T p.R307C Chr.7: 37907601-37907601 on Build GRCh38
6498040 c.928C>T p.R310C Chr.7: 37907592-37907592 on Build GRCh38
4561452 c.885G>A p.R295R Chr.7: 37907635-37907635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931444140 Chr.7: 37907946-37907946 on Build GRCh38
rs755118256 Chr.7: 37907560-37907560 on Build GRCh38
rs978199399 Chr.7: 37907584-37907584 on Build GRCh38
rs908771287 Chr.7: 37907974-37907974 on Build GRCh38
rs765815507 Chr.7: 37907637-37907637 on Build GRCh38
rs749643299 Chr.7: 37907965-37907966 on Build GRCh38
rs751344262 Chr.7: 37907690-37907691 on Build GRCh38
rs183271354 Chr.7: 37907879-37907879 on Build GRCh38
rs568754305 Chr.7: 37907636-37907636 on Build GRCh38
rs200892120 Chr.7: 37907666-37907666 on Build GRCh38
rs577145980 Chr.7: 37907909-37907909 on Build GRCh38
rs745902127 Chr.7: 37907705-37907705 on Build GRCh38
rs554983686 Chr.7: 37907832-37907832 on Build GRCh38
rs368083798 Chr.7: 37907676-37907676 on Build GRCh38
rs1057510838 Chr.7: 37908021-37908021 on Build GRCh38
rs1012984744 Chr.7: 37907756-37907756 on Build GRCh38
rs757108214 Chr.7: 37907699-37907700 on Build GRCh38
rs922906008 Chr.7: 37907995-37907995 on Build GRCh38
rs748472179 Chr.7: 37907575-37907575 on Build GRCh38
rs940331565 Chr.7: 37907714-37907714 on Build GRCh38
rs771031038 Chr.7: 37907591-37907591 on Build GRCh38
rs758881962 Chr.7: 37907576-37907576 on Build GRCh38
rs1010193424 Chr.7: 37907894-37907894 on Build GRCh38
rs766989220 Chr.7: 37907557-37907557 on Build GRCh38
rs763194830 Chr.7: 37908044-37908044 on Build GRCh38
rs1003570406 Chr.7: 37907923-37907923 on Build GRCh38
rs546932583 Chr.7: 37907605-37907605 on Build GRCh38
rs753298451 Chr.7: 37907658-37907658 on Build GRCh38
rs906514703 Chr.7: 37907908-37907908 on Build GRCh38
rs755468190 Chr.7: 37907563-37907563 on Build GRCh38
rs373677734 Chr.7: 37907817-37907817 on Build GRCh38
rs893175943 Chr.7: 37907893-37907893 on Build GRCh38
rs577058963 Chr.7: 37907872-37907872 on Build GRCh38
rs112569801 Chr.7: 37907822-37907822 on Build GRCh38
rs1057147074 Chr.7: 37907672-37907672 on Build GRCh38
rs769681617 Chr.7: 37907600-37907600 on Build GRCh38
rs1034156910 Chr.7: 37907927-37907927 on Build GRCh38
rs761310895 Chr.7: 37907608-37907608 on Build GRCh38
rs566893141 Chr.7: 37907718-37907718 on Build GRCh38
rs777719122 Chr.7: 37907644-37907645 on Build GRCh38
rs778026259 Chr.7: 37907579-37907579 on Build GRCh38
rs867572291 Chr.7: 37907983-37907983 on Build GRCh38
rs975184383 Chr.7: 37907899-37907899 on Build GRCh38
rs569095967 Chr.7: 37907811-37907811 on Build GRCh38
rs1023505204 Chr.7: 37907810-37907810 on Build GRCh38
rs73340883 Chr.7: 37907850-37907850 on Build GRCh38
rs1048564851 Chr.7: 37907956-37907956 on Build GRCh38
rs769914026 Chr.7: 37907713-37907713 on Build GRCh38
rs965454097 Chr.7: 37907880-37907880 on Build GRCh38
rs762703116 Chr.7: 37907604-37907604 on Build GRCh38
rs781512197 Chr.7: 37907704-37907704 on Build GRCh38
rs984255082 Chr.7: 37907947-37907947 on Build GRCh38
rs147145122 Chr.7: 37907573-37907573 on Build GRCh38
rs775432602 Chr.7: 37907601-37907601 on Build GRCh38
rs921021507 Chr.7: 37907933-37907933 on Build GRCh38
rs932960232 Chr.7: 37908022-37908022 on Build GRCh38
rs778056562 Chr.7: 37907664-37907664 on Build GRCh38
rs538747626 Chr.7: 37907759-37907759 on Build GRCh38
rs1020248701 Chr.7: 37907907-37907907 on Build GRCh38
rs934226986 Chr.7: 37907799-37907799 on Build GRCh38
rs758831650 Chr.7: 37907661-37907661 on Build GRCh38
rs775399898 Chr.7: 37907715-37907715 on Build GRCh38
rs767203141 Chr.7: 37907611-37907611 on Build GRCh38
rs910261975 Chr.7: 37908043-37908043 on Build GRCh38
rs1017115737 Chr.7: 37907962-37907962 on Build GRCh38
rs749861176 Chr.7: 37907618-37907618 on Build GRCh38
rs774036745 Chr.7: 37907606-37907606 on Build GRCh38
rs1004146678 Chr.7: 37907635-37907635 on Build GRCh38
rs140322341 Chr.7: 37907629-37907629 on Build GRCh38
rs918838533 Chr.7: 37907740-37907740 on Build GRCh38
rs1051716255 Chr.7: 37907818-37907818 on Build GRCh38
rs867024602 Chr.7: 37907939-37907939 on Build GRCh38
rs552384393 Chr.7: 37907769-37907769 on Build GRCh38
rs776519995 Chr.7: 37907592-37907592 on Build GRCh38
rs747414219 Chr.7: 37907668-37907668 on Build GRCh38
rs969177040 Chr.7: 37907816-37907816 on Build GRCh38
rs1802073 Chr.7: 37907562-37907562 on Build GRCh38
rs750688162 Chr.7: 37907583-37907583 on Build GRCh38
rs533559168 Chr.7: 37907852-37907852 on Build GRCh38
rs781346230 Chr.7: 37907703-37907704 on Build GRCh38
rs190754950 Chr.7: 37907860-37907860 on Build GRCh38
rs148000412 Chr.7: 37908029-37908029 on Build GRCh38

View Full Product Details