Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124976929-124977524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF572
Entrez Gene ID
137209
Gene Name
zinc finger protein 572
Gene Aliases
-
UniGene
Hs.175350
Gene Chromosome Location
Chr.8: 124973297-124979389 on Build GRCh38
Transcripts
NM_152412.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1271589 c.1215A>G p.T405T Chr.8: 124977483-124977483 on Build GRCh38
324515 c.1007A>C p.N336T Chr.8: 124977275-124977275 on Build GRCh38
3834028 c.825T>C p.P275P Chr.8: 124977093-124977093 on Build GRCh38
6500119 c.1197C>G p.L399L Chr.8: 124977465-124977465 on Build GRCh38
6567643 c.849G>A p.Q283Q Chr.8: 124977117-124977117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750476124 Chr.8: 124977160-124977160 on Build GRCh38
rs371828013 Chr.8: 124977116-124977116 on Build GRCh38
rs78004199 Chr.8: 124977190-124977190 on Build GRCh38
rs552116943 Chr.8: 124977046-124977046 on Build GRCh38
rs768612216 Chr.8: 124977310-124977310 on Build GRCh38
rs760896701 Chr.8: 124977250-124977250 on Build GRCh38
rs781233770 Chr.8: 124977084-124977084 on Build GRCh38
rs776669295 Chr.8: 124977473-124977473 on Build GRCh38
rs765333525 Chr.8: 124977039-124977039 on Build GRCh38
rs769832727 Chr.8: 124977143-124977143 on Build GRCh38
rs138227822 Chr.8: 124977119-124977119 on Build GRCh38
rs115342993 Chr.8: 124977236-124977236 on Build GRCh38
rs145772498 Chr.8: 124977191-124977191 on Build GRCh38
rs1000923137 Chr.8: 124977371-124977371 on Build GRCh38
rs776278835 Chr.8: 124977134-124977134 on Build GRCh38
rs751366386 Chr.8: 124977155-124977155 on Build GRCh38
rs138353949 Chr.8: 124977213-124977213 on Build GRCh38
rs765381772 Chr.8: 124977369-124977369 on Build GRCh38
rs867622058 Chr.8: 124977311-124977311 on Build GRCh38
rs866354717 Chr.8: 124977253-124977253 on Build GRCh38
rs745458721 Chr.8: 124977289-124977289 on Build GRCh38
rs748989657 Chr.8: 124977401-124977401 on Build GRCh38
rs746534455 Chr.8: 124977138-124977139 on Build GRCh38
rs771792404 Chr.8: 124977328-124977328 on Build GRCh38
rs746616715 Chr.8: 124977131-124977131 on Build GRCh38
rs751736331 Chr.8: 124977082-124977082 on Build GRCh38
rs867154011 Chr.8: 124976956-124976956 on Build GRCh38
rs554359247 Chr.8: 124977129-124977129 on Build GRCh38
rs768541083 Chr.8: 124977227-124977227 on Build GRCh38
rs759586102 Chr.8: 124977365-124977365 on Build GRCh38
rs750749812 Chr.8: 124977326-124977326 on Build GRCh38
rs541553493 Chr.8: 124977283-124977283 on Build GRCh38
rs367729734 Chr.8: 124976979-124976979 on Build GRCh38
rs755887509 Chr.8: 124977292-124977292 on Build GRCh38
rs772876316 Chr.8: 124977012-124977012 on Build GRCh38
rs539029785 Chr.8: 124977379-124977379 on Build GRCh38
rs572331484 Chr.8: 124977280-124977280 on Build GRCh38
rs901731538 Chr.8: 124976991-124976991 on Build GRCh38
rs765989836 Chr.8: 124977385-124977385 on Build GRCh38
rs748023513 Chr.8: 124977429-124977429 on Build GRCh38
rs765923254 Chr.8: 124977016-124977016 on Build GRCh38
rs1025232157 Chr.8: 124977089-124977089 on Build GRCh38
rs767318769 Chr.8: 124977241-124977241 on Build GRCh38
rs775825279 Chr.8: 124976987-124976987 on Build GRCh38
rs766428416 Chr.8: 124977274-124977274 on Build GRCh38
rs188482651 Chr.8: 124977015-124977015 on Build GRCh38
rs199853193 Chr.8: 124977163-124977163 on Build GRCh38
rs760876458 Chr.8: 124977336-124977336 on Build GRCh38
rs778286370 Chr.8: 124977285-124977285 on Build GRCh38
rs148366492 Chr.8: 124977159-124977159 on Build GRCh38
rs201968418 Chr.8: 124977284-124977284 on Build GRCh38
rs375384539 Chr.8: 124977467-124977467 on Build GRCh38
rs750600647 Chr.8: 124977247-124977247 on Build GRCh38
rs754588368 Chr.8: 124977424-124977424 on Build GRCh38
rs762933274 Chr.8: 124977224-124977224 on Build GRCh38
rs550539701 Chr.8: 124976997-124976997 on Build GRCh38
rs753926505 Chr.8: 124977279-124977279 on Build GRCh38
rs10104558 Chr.8: 124977218-124977218 on Build GRCh38
rs758262358 Chr.8: 124977052-124977052 on Build GRCh38
rs866468414 Chr.8: 124977331-124977331 on Build GRCh38
rs372094161 Chr.8: 124977166-124977166 on Build GRCh38
rs530320212 Chr.8: 124977306-124977306 on Build GRCh38
rs143395753 Chr.8: 124977154-124977154 on Build GRCh38
rs143593483 Chr.8: 124977038-124977038 on Build GRCh38
rs758987141 Chr.8: 124977377-124977377 on Build GRCh38
rs374083749 Chr.8: 124977316-124977316 on Build GRCh38
rs753216515 Chr.8: 124977374-124977374 on Build GRCh38
rs776742147 Chr.8: 124977350-124977350 on Build GRCh38
rs755795239 Chr.8: 124977383-124977383 on Build GRCh38
rs745538040 Chr.8: 124977196-124977196 on Build GRCh38
rs1027186656 Chr.8: 124977474-124977474 on Build GRCh38
rs146940643 Chr.8: 124977062-124977062 on Build GRCh38
rs200935963 Chr.8: 124977423-124977423 on Build GRCh38
rs16900167 Chr.8: 124977456-124977456 on Build GRCh38
rs766579424 Chr.8: 124977338-124977338 on Build GRCh38
rs1003469734 Chr.8: 124977488-124977488 on Build GRCh38
rs771164123 Chr.8: 124977469-124977469 on Build GRCh38
rs757433136 Chr.8: 124977083-124977083 on Build GRCh38
rs143408022 Chr.8: 124977133-124977133 on Build GRCh38
rs748530541 Chr.8: 124977185-124977185 on Build GRCh38
rs758000145 Chr.8: 124977351-124977351 on Build GRCh38
rs775457833 Chr.8: 124977145-124977145 on Build GRCh38
rs1000849748 Chr.8: 124976971-124976971 on Build GRCh38
rs774162507 Chr.8: 124977233-124977233 on Build GRCh38
rs556345811 Chr.8: 124977104-124977104 on Build GRCh38
rs1023005571 Chr.8: 124977056-124977056 on Build GRCh38
rs772060013 Chr.8: 124977451-124977451 on Build GRCh38
rs776297314 Chr.8: 124977024-124977024 on Build GRCh38
rs773008523 Chr.8: 124977335-124977335 on Build GRCh38
rs756133536 Chr.8: 124977162-124977162 on Build GRCh38
rs1025642294 Chr.8: 124977416-124977416 on Build GRCh38
rs759158841 Chr.8: 124977142-124977142 on Build GRCh38
rs10105106 Chr.8: 124977407-124977407 on Build GRCh38
rs749776801 Chr.8: 124977122-124977122 on Build GRCh38
rs762802697 Chr.8: 124977146-124977146 on Build GRCh38
rs147804097 Chr.8: 124977000-124977000 on Build GRCh38
rs779618916 Chr.8: 124977395-124977395 on Build GRCh38
rs543991604 Chr.8: 124977378-124977378 on Build GRCh38
rs868244619 Chr.8: 124977212-124977212 on Build GRCh38
rs748111151 Chr.8: 124977325-124977325 on Build GRCh38
rs374716833 Chr.8: 124976967-124976967 on Build GRCh38
rs565659958 Chr.8: 124977097-124977097 on Build GRCh38
rs779610093 Chr.8: 124977304-124977304 on Build GRCh38
rs762088383 Chr.8: 124977157-124977157 on Build GRCh38
rs779325249 Chr.8: 124977178-124977178 on Build GRCh38

View Full Product Details