Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124450597-124451165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT12
Entrez Gene ID
55039
Gene Name
tRNA methyltransferase 12 homolog
Gene Aliases
TRM12, TYW2
UniGene
Hs.9925
Gene Chromosome Location
Chr.8: 124450807-124453026 on Build GRCh38
Transcripts
NM_017956.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1095869 c.103G>T p.E35* Chr.8: 124451030-124451030 on Build GRCh38
5039140 c.49C>A p.P17T Chr.8: 124450976-124450976 on Build GRCh38
1095871 c.117G>T p.R39S Chr.8: 124451044-124451044 on Build GRCh38
5812769 c.92A>T p.Q31L Chr.8: 124451019-124451019 on Build GRCh38
5821322 c.72G>T p.V24V Chr.8: 124450999-124450999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376700689 Chr.8: 124451032-124451032 on Build GRCh38
rs562285047 Chr.8: 124450833-124450833 on Build GRCh38
rs371321546 Chr.8: 124451064-124451064 on Build GRCh38
rs79997881 Chr.8: 124450760-124450760 on Build GRCh38
rs28444161 Chr.8: 124450671-124450671 on Build GRCh38
rs775356802 Chr.8: 124450910-124450911 on Build GRCh38
rs115174386 Chr.8: 124450732-124450732 on Build GRCh38
rs771073931 Chr.8: 124451137-124451137 on Build GRCh38
rs139253258 Chr.8: 124450978-124450978 on Build GRCh38
rs761270478 Chr.8: 124450934-124450934 on Build GRCh38
rs1028670365 Chr.8: 124450827-124450827 on Build GRCh38
rs764045536 Chr.8: 124450826-124450826 on Build GRCh38
rs558035527 Chr.8: 124450896-124450896 on Build GRCh38
rs150835991 Chr.8: 124450970-124450970 on Build GRCh38
rs369210980 Chr.8: 124450726-124450726 on Build GRCh38
rs772214879 Chr.8: 124451022-124451022 on Build GRCh38
rs768597891 Chr.8: 124451115-124451116 on Build GRCh38
rs546219242 Chr.8: 124450761-124450761 on Build GRCh38
rs987556253 Chr.8: 124450748-124450748 on Build GRCh38
rs779819750 Chr.8: 124451094-124451094 on Build GRCh38
rs906448860 Chr.8: 124450813-124450813 on Build GRCh38
rs773767312 Chr.8: 124450816-124450816 on Build GRCh38
rs377389381 Chr.8: 124450897-124450897 on Build GRCh38
rs548427345 Chr.8: 124450817-124450817 on Build GRCh38
rs773402210 Chr.8: 124451135-124451135 on Build GRCh38
rs755450694 Chr.8: 124450650-124450664 on Build GRCh38
rs369340865 Chr.8: 124451027-124451027 on Build GRCh38
rs895175124 Chr.8: 124450649-124450649 on Build GRCh38
rs539001041 Chr.8: 124450841-124450841 on Build GRCh38
rs766847093 Chr.8: 124450935-124450935 on Build GRCh38
rs868102974 Chr.8: 124450870-124450870 on Build GRCh38
rs973422586 Chr.8: 124450853-124450853 on Build GRCh38
rs749274141 Chr.8: 124451101-124451101 on Build GRCh38
rs767728139 Chr.8: 124450928-124450928 on Build GRCh38
rs1049543327 Chr.8: 124450729-124450729 on Build GRCh38
rs759777990 Chr.8: 124451060-124451060 on Build GRCh38
rs768530433 Chr.8: 124451001-124451001 on Build GRCh38
rs1048604736 Chr.8: 124450864-124450864 on Build GRCh38
rs1001576778 Chr.8: 124450915-124450916 on Build GRCh38
rs1034478292 Chr.8: 124450803-124450803 on Build GRCh38
rs780579629 Chr.8: 124450878-124450878 on Build GRCh38
rs762669515 Chr.8: 124451027-124451028 on Build GRCh38
rs1054241659 Chr.8: 124450880-124450883 on Build GRCh38
rs1046051643 Chr.8: 124451133-124451133 on Build GRCh38
rs760098141 Chr.8: 124451140-124451140 on Build GRCh38
rs761762527 Chr.8: 124450927-124450927 on Build GRCh38
rs386729566 Chr.8: 124450849-124450857 on Build GRCh38
rs916786447 Chr.8: 124451138-124451138 on Build GRCh38
rs758800934 Chr.8: 124450762-124450762 on Build GRCh38
rs3812475 Chr.8: 124451009-124451009 on Build GRCh38
rs775149324 Chr.8: 124450911-124450911 on Build GRCh38
rs201957009 Chr.8: 124450992-124450992 on Build GRCh38
rs763225799 Chr.8: 124450919-124450919 on Build GRCh38
rs139807028 Chr.8: 124450836-124450836 on Build GRCh38
rs999885296 Chr.8: 124450814-124450814 on Build GRCh38
rs1037127090 Chr.8: 124450684-124450684 on Build GRCh38
rs952944835 Chr.8: 124451093-124451093 on Build GRCh38
rs1026978104 Chr.8: 124450644-124450644 on Build GRCh38
rs373565254 Chr.8: 124451031-124451031 on Build GRCh38
rs376989869 Chr.8: 124450876-124450876 on Build GRCh38
rs999432480 Chr.8: 124450918-124450918 on Build GRCh38
rs751801443 Chr.8: 124450845-124450845 on Build GRCh38
rs751253208 Chr.8: 124450965-124450965 on Build GRCh38
rs772153773 Chr.8: 124450801-124450801 on Build GRCh38
rs989219321 Chr.8: 124450734-124450734 on Build GRCh38
rs775614170 Chr.8: 124450805-124450805 on Build GRCh38
rs756905425 Chr.8: 124451068-124451068 on Build GRCh38
rs77055381 Chr.8: 124450849-124450849 on Build GRCh38
rs779878756 Chr.8: 124450990-124450990 on Build GRCh38
rs562056044 Chr.8: 124451026-124451026 on Build GRCh38
rs1046835311 Chr.8: 124450806-124450806 on Build GRCh38
rs28607530 Chr.8: 124450764-124450764 on Build GRCh38
rs770823452 Chr.8: 124450890-124450890 on Build GRCh38
rs555531898 Chr.8: 124450715-124450715 on Build GRCh38
rs377493496 Chr.8: 124451002-124451002 on Build GRCh38
rs1052280319 Chr.8: 124450960-124450960 on Build GRCh38
rs756016879 Chr.8: 124451084-124451084 on Build GRCh38
rs931444178 Chr.8: 124450858-124450858 on Build GRCh38
rs953026703 Chr.8: 124450746-124450746 on Build GRCh38
rs912779960 Chr.8: 124450758-124450758 on Build GRCh38
rs933746548 Chr.8: 124450690-124450690 on Build GRCh38
rs752328483 Chr.8: 124450879-124450879 on Build GRCh38
rs778774155 Chr.8: 124451111-124451111 on Build GRCh38
rs754211628 Chr.8: 124450940-124450940 on Build GRCh38
rs895248191 Chr.8: 124450783-124450783 on Build GRCh38
rs774462489 Chr.8: 124450924-124450924 on Build GRCh38
rs528586166 Chr.8: 124450812-124450812 on Build GRCh38
rs777323223 Chr.8: 124450882-124450882 on Build GRCh38
rs780691540 Chr.8: 124450972-124450972 on Build GRCh38
rs149678009 Chr.8: 124450979-124450979 on Build GRCh38
rs78376625 Chr.8: 124450986-124450986 on Build GRCh38
rs957889537 Chr.8: 124450825-124450825 on Build GRCh38
rs570821503 Chr.8: 124450840-124450840 on Build GRCh38
rs1045699701 Chr.8: 124450900-124450900 on Build GRCh38
rs768812661 Chr.8: 124450921-124450921 on Build GRCh38
rs200524864 Chr.8: 124450889-124450889 on Build GRCh38
rs748378128 Chr.8: 124451131-124451131 on Build GRCh38
rs749026147 Chr.8: 124450736-124450736 on Build GRCh38
rs377302398 Chr.8: 124450908-124450908 on Build GRCh38
rs772481497 Chr.8: 124451132-124451132 on Build GRCh38
rs3812473 Chr.8: 124450842-124450842 on Build GRCh38
rs1048532289 Chr.8: 124450770-124450770 on Build GRCh38
rs6981043 Chr.8: 124450745-124450745 on Build GRCh38
rs531032767 Chr.8: 124450835-124450835 on Build GRCh38
rs754673222 Chr.8: 124451107-124451107 on Build GRCh38
rs749987580 Chr.8: 124451071-124451071 on Build GRCh38
rs201870760 Chr.8: 124450880-124450880 on Build GRCh38
rs777330173 Chr.8: 124450953-124450953 on Build GRCh38
rs952181675 Chr.8: 124450809-124450809 on Build GRCh38
rs1024866803 Chr.8: 124450654-124450654 on Build GRCh38
rs767006145 Chr.8: 124451070-124451070 on Build GRCh38
rs922351124 Chr.8: 124450689-124450689 on Build GRCh38
rs747435475 Chr.8: 124450919-124450919 on Build GRCh38
rs190418131 Chr.8: 124450843-124450843 on Build GRCh38
rs577994915 Chr.8: 124450941-124450941 on Build GRCh38
rs751110771 Chr.8: 124451067-124451067 on Build GRCh38
rs931370809 Chr.8: 124450766-124450766 on Build GRCh38
rs750902365 Chr.8: 124450932-124450932 on Build GRCh38
rs3812474 Chr.8: 124450857-124450857 on Build GRCh38
rs777399795 Chr.8: 124450653-124450653 on Build GRCh38

View Full Product Details