Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27325131-27325730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB3B
Entrez Gene ID
79817
Gene Name
MOB kinase activator 3B
Gene Aliases
C9orf35, MOB1D, MOBKL2B
UniGene
Hs.369022
Gene Chromosome Location
Chr.9: 27325209-27529852 on Build GRCh38
Transcripts
NM_024761.4

SNP Detail

SNP ID Chromosome Location
rs989496004 Chr.9: 27325477-27325477 on Build GRCh38
rs899041587 Chr.9: 27325611-27325611 on Build GRCh38
rs1008255987 Chr.9: 27325445-27325445 on Build GRCh38
rs1037612890 Chr.9: 27325319-27325319 on Build GRCh38
rs559424812 Chr.9: 27325655-27325655 on Build GRCh38
rs1056886955 Chr.9: 27325558-27325558 on Build GRCh38
rs1004395928 Chr.9: 27325352-27325352 on Build GRCh38
rs937792768 Chr.9: 27325557-27325557 on Build GRCh38
rs1050253206 Chr.9: 27325628-27325631 on Build GRCh38
rs535424842 Chr.9: 27325208-27325208 on Build GRCh38
rs927438176 Chr.9: 27325540-27325540 on Build GRCh38
rs867179550 Chr.9: 27325630-27325630 on Build GRCh38
rs373210671 Chr.9: 27325236-27325236 on Build GRCh38
rs996941048 Chr.9: 27325412-27325412 on Build GRCh38
rs899008692 Chr.9: 27325332-27325332 on Build GRCh38
rs938336366 Chr.9: 27325291-27325291 on Build GRCh38
rs533211836 Chr.9: 27325688-27325688 on Build GRCh38
rs1038038932 Chr.9: 27325581-27325581 on Build GRCh38
rs958053004 Chr.9: 27325421-27325421 on Build GRCh38
rs980233479 Chr.9: 27325508-27325508 on Build GRCh38
rs553580365 Chr.9: 27325325-27325325 on Build GRCh38
rs867702734 Chr.9: 27325487-27325487 on Build GRCh38
rs529289408 Chr.9: 27325586-27325586 on Build GRCh38
rs1010199395 Chr.9: 27325699-27325699 on Build GRCh38
rs931584382 Chr.9: 27325260-27325260 on Build GRCh38
rs898822618 Chr.9: 27325248-27325248 on Build GRCh38
rs920277466 Chr.9: 27325568-27325568 on Build GRCh38
rs147782374 Chr.9: 27325471-27325471 on Build GRCh38
rs1047818447 Chr.9: 27325635-27325635 on Build GRCh38
rs113158798 Chr.9: 27325244-27325244 on Build GRCh38
rs1005233856 Chr.9: 27325313-27325313 on Build GRCh38
rs1048620448 Chr.9: 27325293-27325293 on Build GRCh38
rs1028777934 Chr.9: 27325419-27325419 on Build GRCh38
rs749516065 Chr.9: 27325402-27325402 on Build GRCh38
rs12348286 Chr.9: 27325643-27325643 on Build GRCh38
rs969295611 Chr.9: 27325507-27325507 on Build GRCh38
rs374531950 Chr.9: 27325633-27325636 on Build GRCh38
rs940214100 Chr.9: 27325659-27325659 on Build GRCh38
rs912821002 Chr.9: 27325213-27325213 on Build GRCh38
rs1028334644 Chr.9: 27325161-27325161 on Build GRCh38
rs954061730 Chr.9: 27325204-27325204 on Build GRCh38
rs568737634 Chr.9: 27325688-27325688 on Build GRCh38
rs966571238 Chr.9: 27325476-27325476 on Build GRCh38
rs775630906 Chr.9: 27325362-27325362 on Build GRCh38
rs7049080 Chr.9: 27325454-27325454 on Build GRCh38
rs1019642498 Chr.9: 27325663-27325663 on Build GRCh38
rs914800418 Chr.9: 27325479-27325479 on Build GRCh38
rs930342883 Chr.9: 27325618-27325618 on Build GRCh38
rs989074209 Chr.9: 27325231-27325231 on Build GRCh38
rs905573423 Chr.9: 27325278-27325278 on Build GRCh38
rs981919000 Chr.9: 27325480-27325480 on Build GRCh38
rs887331442 Chr.9: 27325296-27325296 on Build GRCh38
rs934941395 Chr.9: 27325214-27325214 on Build GRCh38
rs927798168 Chr.9: 27325529-27325529 on Build GRCh38
rs541024189 Chr.9: 27325612-27325612 on Build GRCh38
rs546431723 Chr.9: 27325351-27325351 on Build GRCh38
rs766264196 Chr.9: 27325607-27325607 on Build GRCh38
rs10967889 Chr.9: 27325560-27325560 on Build GRCh38
rs1028365854 Chr.9: 27325420-27325420 on Build GRCh38
rs887362330 Chr.9: 27325649-27325649 on Build GRCh38
rs558038324 Chr.9: 27325359-27325359 on Build GRCh38
rs553946570 Chr.9: 27325211-27325211 on Build GRCh38
rs561726593 Chr.9: 27325310-27325313 on Build GRCh38
rs986721327 Chr.9: 27325209-27325209 on Build GRCh38

View Full Product Details