Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38981813-38982008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOS1
Entrez Gene ID
6654
Gene Name
SOS Ras/Rac guanine nucleotide exchange factor 1
Gene Aliases
GF1, GGF1, GINGF, HGF, NS4
UniGene
Hs.709893
Gene Chromosome Location
Chr.2: 38981549-39124959 on Build GRCh38
Transcripts
NM_005633.3

SNP Detail

SNP ID Chromosome Location
rs977983815 Chr.2: 38981842-38981842 on Build GRCh38
rs923836534 Chr.2: 38981844-38981844 on Build GRCh38
rs955962734 Chr.2: 38981884-38981884 on Build GRCh38
rs536821906 Chr.2: 38981894-38981894 on Build GRCh38
rs757234736 Chr.2: 38981870-38981870 on Build GRCh38
rs898667811 Chr.2: 38981915-38981915 on Build GRCh38
rs1043793 Chr.2: 38981906-38981906 on Build GRCh38
rs1051131824 Chr.2: 38981869-38981869 on Build GRCh38
rs759088430 Chr.2: 38981896-38981896 on Build GRCh38
rs933998713 Chr.2: 38981863-38981866 on Build GRCh38
rs968042160 Chr.2: 38981962-38981962 on Build GRCh38
rs1020927823 Chr.2: 38981912-38981912 on Build GRCh38
rs979830760 Chr.2: 38981965-38981965 on Build GRCh38

View Full Product Details