Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1219364-1219531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK11
Entrez Gene ID
6794
Gene Name
serine/threonine kinase 11
Gene Aliases
LKB1, PJS, hLKB1
UniGene
Hs.515005
Gene Chromosome Location
Chr.19: 1205799-1228435 on Build GRCh38
Transcripts
NM_000455.4 XM_005259617.2 XM_005259618.3 XM_011528209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6984763 c.464+1G>T p.? Chr.19: 1219414-1219414 on Build GRCh38
6922289 c.461A>T p.H154L Chr.19: 1219410-1219410 on Build GRCh38
6983748 c.441delT p.F148fs*13 Chr.19: 1219390-1219390 on Build GRCh38
6961044 c.440G>A p.R147H Chr.19: 1219389-1219389 on Build GRCh38
27023 c.1_*del p.0 Chr.19: 1206914-1226647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869129729 Chr.19: 1219460-1219461 on Build GRCh38
rs766345051 Chr.19: 1219447-1219447 on Build GRCh38
rs748330307 Chr.19: 1219463-1219463 on Build GRCh38
rs113069959 Chr.19: 1219425-1219425 on Build GRCh38
rs951888087 Chr.19: 1219453-1219453 on Build GRCh38
rs538840976 Chr.19: 1219507-1219507 on Build GRCh38
rs994299997 Chr.19: 1219449-1219449 on Build GRCh38
rs587781802 Chr.19: 1219398-1219398 on Build GRCh38
rs369764220 Chr.19: 1219383-1219383 on Build GRCh38
rs12608721 Chr.19: 1219444-1219444 on Build GRCh38
rs781353508 Chr.19: 1219460-1219460 on Build GRCh38
rs780264271 Chr.19: 1219463-1219464 on Build GRCh38
rs909289240 Chr.19: 1219501-1219501 on Build GRCh38
rs968991091 Chr.19: 1219439-1219439 on Build GRCh38
rs786201418 Chr.19: 1219411-1219411 on Build GRCh38
rs376788924 Chr.19: 1219381-1219381 on Build GRCh38
rs587782369 Chr.19: 1219433-1219433 on Build GRCh38
rs587782452 Chr.19: 1219426-1219426 on Build GRCh38
rs544282452 Chr.19: 1219452-1219453 on Build GRCh38
rs587780718 Chr.19: 1219396-1219396 on Build GRCh38
rs587781579 Chr.19: 1219420-1219420 on Build GRCh38
rs587782453 Chr.19: 1219431-1219431 on Build GRCh38
rs587781681 Chr.19: 1219418-1219418 on Build GRCh38
rs869153329 Chr.19: 1219476-1219476 on Build GRCh38
rs878853989 Chr.19: 1219421-1219421 on Build GRCh38
rs751617266 Chr.19: 1219448-1219448 on Build GRCh38
rs587780717 Chr.19: 1219389-1219389 on Build GRCh38
rs750293847 Chr.19: 1219438-1219438 on Build GRCh38
rs757335801 Chr.19: 1219445-1219451 on Build GRCh38
rs863224669 Chr.19: 1219421-1219421 on Build GRCh38
rs587782429 Chr.19: 1219430-1219430 on Build GRCh38
rs376313955 Chr.19: 1219422-1219422 on Build GRCh38
rs948063169 Chr.19: 1219481-1219481 on Build GRCh38
rs730881960 Chr.19: 1219433-1219433 on Build GRCh38
rs587782421 Chr.19: 1219427-1219427 on Build GRCh38
rs748464757 Chr.19: 1219388-1219388 on Build GRCh38
rs373167735 Chr.19: 1219417-1219417 on Build GRCh38
rs758268029 Chr.19: 1219446-1219446 on Build GRCh38
rs981295184 Chr.19: 1219492-1219492 on Build GRCh38
rs730881971 Chr.19: 1219414-1219414 on Build GRCh38
rs763353991 Chr.19: 1219412-1219412 on Build GRCh38
rs917889736 Chr.19: 1219480-1219480 on Build GRCh38
rs878853988 Chr.19: 1219409-1219409 on Build GRCh38
rs571895916 Chr.19: 1219424-1219424 on Build GRCh38
rs587782430 Chr.19: 1219432-1219432 on Build GRCh38
rs992134759 Chr.19: 1219471-1219471 on Build GRCh38
rs58579265 Chr.19: 1219459-1219460 on Build GRCh38
rs587782445 Chr.19: 1219423-1219423 on Build GRCh38
rs786201510 Chr.19: 1219402-1219402 on Build GRCh38
rs369515604 Chr.19: 1219452-1219452 on Build GRCh38
rs786201456 Chr.19: 1219390-1219390 on Build GRCh38
rs869127541 Chr.19: 1219445-1219446 on Build GRCh38
rs769837743 Chr.19: 1219460-1219460 on Build GRCh38
rs777847857 Chr.19: 1219440-1219441 on Build GRCh38

View Full Product Details