Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38754882-38755385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR5
Entrez Gene ID
79913
Gene Name
ARP5 actin-related protein 5 homolog
Gene Aliases
Arp5, INO80M
UniGene
Hs.371585
Gene Chromosome Location
Chr.20: 38748454-38772446 on Build GRCh38
Transcripts
NM_024855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389742 c.817G>C p.V273L Chr.20: 38754998-38754998 on Build GRCh38
6593118 c.955C>T p.L319L Chr.20: 38755136-38755136 on Build GRCh38
3423640 c.804T>G p.Y268* Chr.20: 38754985-38754985 on Build GRCh38
6593121 c.790C>T p.R264W Chr.20: 38754971-38754971 on Build GRCh38
3405072 c.934C>T p.R312W Chr.20: 38755115-38755115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778068411 Chr.20: 38755084-38755084 on Build GRCh38
rs371726552 Chr.20: 38755184-38755184 on Build GRCh38
rs754891634 Chr.20: 38755126-38755126 on Build GRCh38
rs766320537 Chr.20: 38755050-38755050 on Build GRCh38
rs763823068 Chr.20: 38754922-38754922 on Build GRCh38
rs773605328 Chr.20: 38754983-38754983 on Build GRCh38
rs775002324 Chr.20: 38755089-38755089 on Build GRCh38
rs538066451 Chr.20: 38755294-38755294 on Build GRCh38
rs2757521 Chr.20: 38755198-38755198 on Build GRCh38
rs894135672 Chr.20: 38755193-38755193 on Build GRCh38
rs2748663 Chr.20: 38754997-38754997 on Build GRCh38
rs752818849 Chr.20: 38755138-38755138 on Build GRCh38
rs771466412 Chr.20: 38754985-38754985 on Build GRCh38
rs368018075 Chr.20: 38755107-38755107 on Build GRCh38
rs761408786 Chr.20: 38755111-38755111 on Build GRCh38
rs768302928 Chr.20: 38755106-38755106 on Build GRCh38
rs779456397 Chr.20: 38755218-38755218 on Build GRCh38
rs751434584 Chr.20: 38755063-38755063 on Build GRCh38
rs781203112 Chr.20: 38755071-38755071 on Build GRCh38
rs1028739010 Chr.20: 38755250-38755250 on Build GRCh38
rs766125785 Chr.20: 38755118-38755118 on Build GRCh38
rs944295614 Chr.20: 38755120-38755120 on Build GRCh38
rs999867150 Chr.20: 38755205-38755205 on Build GRCh38
rs769121543 Chr.20: 38755165-38755165 on Build GRCh38
rs533306645 Chr.20: 38755215-38755215 on Build GRCh38
rs771362765 Chr.20: 38755088-38755088 on Build GRCh38
rs191589290 Chr.20: 38755292-38755292 on Build GRCh38
rs1024043828 Chr.20: 38755195-38755195 on Build GRCh38
rs770080319 Chr.20: 38754975-38754975 on Build GRCh38
rs779369830 Chr.20: 38755150-38755150 on Build GRCh38
rs923368645 Chr.20: 38755338-38755338 on Build GRCh38
rs758329717 Chr.20: 38754952-38754952 on Build GRCh38
rs757007106 Chr.20: 38754930-38754930 on Build GRCh38
rs529387368 Chr.20: 38755272-38755272 on Build GRCh38
rs748333992 Chr.20: 38754972-38754972 on Build GRCh38
rs754111067 Chr.20: 38755208-38755208 on Build GRCh38
rs747084550 Chr.20: 38754962-38754962 on Build GRCh38
rs756351499 Chr.20: 38755139-38755139 on Build GRCh38
rs571836749 Chr.20: 38755349-38755349 on Build GRCh38
rs989171800 Chr.20: 38755232-38755232 on Build GRCh38
rs760272731 Chr.20: 38755000-38755000 on Build GRCh38
rs770766360 Chr.20: 38755189-38755189 on Build GRCh38
rs368582667 Chr.20: 38754944-38754944 on Build GRCh38
rs552006034 Chr.20: 38755332-38755332 on Build GRCh38
rs374715117 Chr.20: 38755066-38755066 on Build GRCh38
rs773613569 Chr.20: 38754943-38754943 on Build GRCh38
rs760472700 Chr.20: 38754917-38754917 on Build GRCh38
rs772671458 Chr.20: 38755157-38755157 on Build GRCh38
rs375244248 Chr.20: 38755147-38755147 on Build GRCh38
rs772952815 Chr.20: 38755179-38755179 on Build GRCh38
rs749402313 Chr.20: 38755148-38755148 on Build GRCh38
rs753481694 Chr.20: 38754925-38754925 on Build GRCh38
rs990074574 Chr.20: 38755258-38755258 on Build GRCh38
rs1030636466 Chr.20: 38755205-38755205 on Build GRCh38
rs778729065 Chr.20: 38754934-38754934 on Build GRCh38
rs780144893 Chr.20: 38754956-38754956 on Build GRCh38
rs367988045 Chr.20: 38755158-38755158 on Build GRCh38
rs758331207 Chr.20: 38755044-38755044 on Build GRCh38
rs371854747 Chr.20: 38755113-38755113 on Build GRCh38
rs566908191 Chr.20: 38755266-38755266 on Build GRCh38
rs368659646 Chr.20: 38755149-38755149 on Build GRCh38
rs748369066 Chr.20: 38755073-38755073 on Build GRCh38
rs746421999 Chr.20: 38755153-38755153 on Build GRCh38
rs138432468 Chr.20: 38755018-38755018 on Build GRCh38
rs957970505 Chr.20: 38755213-38755213 on Build GRCh38
rs113029636 Chr.20: 38755277-38755277 on Build GRCh38
rs776014468 Chr.20: 38754978-38754978 on Build GRCh38
rs762814710 Chr.20: 38755116-38755116 on Build GRCh38
rs944281263 Chr.20: 38755311-38755311 on Build GRCh38
rs957556835 Chr.20: 38755239-38755239 on Build GRCh38
rs761475461 Chr.20: 38755007-38755007 on Build GRCh38
rs112273594 Chr.20: 38755263-38755263 on Build GRCh38
rs746336224 Chr.20: 38755092-38755092 on Build GRCh38
rs976029649 Chr.20: 38755286-38755286 on Build GRCh38
rs747679194 Chr.20: 38755161-38755161 on Build GRCh38
rs761153442 Chr.20: 38755161-38755162 on Build GRCh38
rs912726737 Chr.20: 38755309-38755309 on Build GRCh38
rs763630364 Chr.20: 38755001-38755001 on Build GRCh38
rs184783832 Chr.20: 38755199-38755199 on Build GRCh38
rs370363764 Chr.20: 38754946-38754946 on Build GRCh38
rs753575773 Chr.20: 38755006-38755006 on Build GRCh38
rs188129219 Chr.20: 38755207-38755207 on Build GRCh38
rs766963412 Chr.20: 38755169-38755169 on Build GRCh38
rs755084972 Chr.20: 38755068-38755068 on Build GRCh38
rs767625308 Chr.20: 38755128-38755128 on Build GRCh38
rs759537662 Chr.20: 38755197-38755197 on Build GRCh38
rs150584876 Chr.20: 38755194-38755194 on Build GRCh38
rs765115786 Chr.20: 38755064-38755064 on Build GRCh38
rs768668556 Chr.20: 38754967-38754967 on Build GRCh38
rs772935923 Chr.20: 38755115-38755115 on Build GRCh38
rs201062646 Chr.20: 38755119-38755119 on Build GRCh38
rs17853829 Chr.20: 38755074-38755074 on Build GRCh38
rs756347740 Chr.20: 38755262-38755262 on Build GRCh38
rs781400177 Chr.20: 38754970-38754970 on Build GRCh38
rs750199504 Chr.20: 38755042-38755042 on Build GRCh38

View Full Product Details