Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116832659-116833161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOC3
Entrez Gene ID
345
Gene Name
apolipoprotein C3
Gene Aliases
APOCIII, HALP2
UniGene
Hs.73849
Gene Chromosome Location
Chr.11: 116829908-116833071 on Build GRCh38
Transcripts
NM_000040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386465 c.253T>A p.W85R Chr.11: 116832837-116832837 on Build GRCh38
6269806 c.185G>T p.W62L Chr.11: 116832769-116832769 on Build GRCh38
4402746 c.213A>C p.K71N Chr.11: 116832797-116832797 on Build GRCh38
6305171 c.269A>G p.E90G Chr.11: 116832853-116832853 on Build GRCh38
4662528 c.285A>G p.S95S Chr.11: 116832869-116832869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541512801 Chr.11: 116833071-116833071 on Build GRCh38
rs113565160 Chr.11: 116833068-116833068 on Build GRCh38
rs558936201 Chr.11: 116832715-116832715 on Build GRCh38
rs980623602 Chr.11: 116832888-116832888 on Build GRCh38
rs201360025 Chr.11: 116832919-116832919 on Build GRCh38
rs1010056473 Chr.11: 116832696-116832696 on Build GRCh38
rs763333861 Chr.11: 116832936-116832936 on Build GRCh38
rs561318344 Chr.11: 116832707-116832707 on Build GRCh38
rs986916861 Chr.11: 116832858-116832858 on Build GRCh38
rs990216591 Chr.11: 116833120-116833120 on Build GRCh38
rs751325346 Chr.11: 116832809-116832809 on Build GRCh38
rs778463167 Chr.11: 116832800-116832800 on Build GRCh38
rs11540884 Chr.11: 116833033-116833033 on Build GRCh38
rs748747574 Chr.11: 116832894-116832894 on Build GRCh38
rs750185333 Chr.11: 116832863-116832863 on Build GRCh38
rs764210608 Chr.11: 116832770-116832770 on Build GRCh38
rs770499173 Chr.11: 116832838-116832838 on Build GRCh38
rs111429645 Chr.11: 116832687-116832687 on Build GRCh38
rs577085953 Chr.11: 116832980-116832980 on Build GRCh38
rs539003433 Chr.11: 116832983-116832983 on Build GRCh38
rs779886571 Chr.11: 116832801-116832801 on Build GRCh38
rs112889374 Chr.11: 116832821-116832821 on Build GRCh38
rs368892201 Chr.11: 116832755-116832755 on Build GRCh38
rs907124040 Chr.11: 116832942-116832942 on Build GRCh38
rs754898637 Chr.11: 116832813-116832813 on Build GRCh38
rs748953052 Chr.11: 116832834-116832834 on Build GRCh38
rs758179886 Chr.11: 116832902-116832903 on Build GRCh38
rs373110458 Chr.11: 116832721-116832721 on Build GRCh38
rs752729869 Chr.11: 116832757-116832757 on Build GRCh38
rs980146319 Chr.11: 116833057-116833057 on Build GRCh38
rs776707156 Chr.11: 116832932-116832932 on Build GRCh38
rs113617501 Chr.11: 116832734-116832734 on Build GRCh38
rs752681827 Chr.11: 116833016-116833016 on Build GRCh38
rs199963291 Chr.11: 116832866-116832866 on Build GRCh38
rs971827928 Chr.11: 116833046-116833046 on Build GRCh38
rs1024296488 Chr.11: 116833035-116833035 on Build GRCh38
rs900866832 Chr.11: 116832953-116832953 on Build GRCh38
rs765406427 Chr.11: 116832934-116832934 on Build GRCh38
rs548741530 Chr.11: 116832892-116832892 on Build GRCh38
rs1032811450 Chr.11: 116833002-116833002 on Build GRCh38
rs772397071 Chr.11: 116832918-116832918 on Build GRCh38
rs144573427 Chr.11: 116832872-116832872 on Build GRCh38
rs5128 Chr.11: 116832924-116832924 on Build GRCh38
rs121918382 Chr.11: 116832816-116832816 on Build GRCh38
rs769165532 Chr.11: 116832930-116832930 on Build GRCh38
rs1034820569 Chr.11: 116833075-116833075 on Build GRCh38
rs561855524 Chr.11: 116833079-116833079 on Build GRCh38
rs760769978 Chr.11: 116832764-116832764 on Build GRCh38
rs201402310 Chr.11: 116832905-116832905 on Build GRCh38
rs756891658 Chr.11: 116832902-116832902 on Build GRCh38
rs772255301 Chr.11: 116832779-116832779 on Build GRCh38
rs927467188 Chr.11: 116832906-116832906 on Build GRCh38
rs908080204 Chr.11: 116833125-116833125 on Build GRCh38
rs760028617 Chr.11: 116832740-116832740 on Build GRCh38
rs759846028 Chr.11: 116832845-116832845 on Build GRCh38
rs568474622 Chr.11: 116832688-116832688 on Build GRCh38
rs1019701320 Chr.11: 116833042-116833042 on Build GRCh38
rs5129 Chr.11: 116832748-116832748 on Build GRCh38
rs866791700 Chr.11: 116832720-116832720 on Build GRCh38
rs995893452 Chr.11: 116833018-116833019 on Build GRCh38
rs930776786 Chr.11: 116832908-116832908 on Build GRCh38
rs745586339 Chr.11: 116832910-116832910 on Build GRCh38
rs761832101 Chr.11: 116832933-116832933 on Build GRCh38
rs772522961 Chr.11: 116832848-116832848 on Build GRCh38
rs761215628 Chr.11: 116832854-116832854 on Build GRCh38
rs757490802 Chr.11: 116832790-116832790 on Build GRCh38
rs955965915 Chr.11: 116832829-116832829 on Build GRCh38
rs121918381 Chr.11: 116832864-116832864 on Build GRCh38
rs4225 Chr.11: 116832955-116832955 on Build GRCh38
rs781170854 Chr.11: 116832873-116832873 on Build GRCh38
rs964270062 Chr.11: 116833074-116833074 on Build GRCh38
rs774709008 Chr.11: 116832839-116832839 on Build GRCh38
rs764157867 Chr.11: 116832855-116832855 on Build GRCh38
rs762551524 Chr.11: 116832730-116832730 on Build GRCh38
rs754046943 Chr.11: 116832729-116832729 on Build GRCh38
rs541177581 Chr.11: 116832694-116832694 on Build GRCh38
rs530355846 Chr.11: 116832731-116832731 on Build GRCh38
rs574912557 Chr.11: 116833083-116833083 on Build GRCh38
rs999953122 Chr.11: 116832996-116832996 on Build GRCh38
rs12721084 Chr.11: 116832682-116832682 on Build GRCh38
rs149707394 Chr.11: 116832777-116832777 on Build GRCh38
rs200945195 Chr.11: 116832818-116832818 on Build GRCh38
rs187628630 Chr.11: 116833023-116833023 on Build GRCh38
rs752676984 Chr.11: 116832879-116832879 on Build GRCh38
rs756176987 Chr.11: 116832883-116832883 on Build GRCh38
rs570839541 Chr.11: 116833003-116833003 on Build GRCh38
rs984916706 Chr.11: 116833117-116833117 on Build GRCh38
rs773273781 Chr.11: 116832935-116832935 on Build GRCh38
rs753924368 Chr.11: 116832859-116832859 on Build GRCh38
rs775956260 Chr.11: 116832849-116832849 on Build GRCh38

View Full Product Details