Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5468537-5469083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD274
Entrez Gene ID
29126
Gene Name
CD274 molecule
Gene Aliases
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
UniGene
Hs.521989
Gene Chromosome Location
Chr.9: 5450503-5470567 on Build GRCh38
Transcripts
NM_001267706.1 NM_014143.3 NR_052005.1

SNP Detail

SNP ID Chromosome Location
rs776586431 Chr.9: 5468731-5468731 on Build GRCh38
rs186914186 Chr.9: 5468732-5468732 on Build GRCh38
rs1038325969 Chr.9: 5468765-5468765 on Build GRCh38
rs991609903 Chr.9: 5468970-5468970 on Build GRCh38
rs563372752 Chr.9: 5468887-5468887 on Build GRCh38
rs990401843 Chr.9: 5468654-5468654 on Build GRCh38
rs1011769981 Chr.9: 5468898-5468898 on Build GRCh38
rs776021510 Chr.9: 5468563-5468563 on Build GRCh38
rs1029283375 Chr.9: 5468637-5468637 on Build GRCh38
rs879588589 Chr.9: 5468886-5468886 on Build GRCh38
rs929892629 Chr.9: 5469013-5469013 on Build GRCh38
rs977837189 Chr.9: 5468696-5468696 on Build GRCh38
rs192793526 Chr.9: 5468649-5468649 on Build GRCh38
rs541213585 Chr.9: 5468786-5468786 on Build GRCh38
rs1005021854 Chr.9: 5468576-5468576 on Build GRCh38
rs184996177 Chr.9: 5468926-5468926 on Build GRCh38
rs994312106 Chr.9: 5468573-5468573 on Build GRCh38
rs1046844111 Chr.9: 5468805-5468805 on Build GRCh38
rs1008313117 Chr.9: 5468622-5468622 on Build GRCh38
rs967433470 Chr.9: 5468671-5468671 on Build GRCh38
rs1039456785 Chr.9: 5469034-5469034 on Build GRCh38
rs765952534 Chr.9: 5468758-5468758 on Build GRCh38
rs974332314 Chr.9: 5468992-5468992 on Build GRCh38
rs1047175251 Chr.9: 5468815-5468815 on Build GRCh38
rs576576820 Chr.9: 5468568-5468568 on Build GRCh38
rs945802545 Chr.9: 5468687-5468687 on Build GRCh38
rs912255975 Chr.9: 5469028-5469028 on Build GRCh38
rs928712878 Chr.9: 5468724-5468724 on Build GRCh38
rs561827341 Chr.9: 5468783-5468783 on Build GRCh38
rs957181713 Chr.9: 5469042-5469042 on Build GRCh38
rs184360875 Chr.9: 5468708-5468708 on Build GRCh38
rs41280723 Chr.9: 5468647-5468647 on Build GRCh38
rs919817968 Chr.9: 5469009-5469009 on Build GRCh38
rs899604155 Chr.9: 5469046-5469046 on Build GRCh38
rs973660052 Chr.9: 5468627-5468627 on Build GRCh38
rs533075809 Chr.9: 5468863-5468863 on Build GRCh38
rs754878431 Chr.9: 5468925-5468925 on Build GRCh38
rs763415855 Chr.9: 5468632-5468632 on Build GRCh38
rs969183623 Chr.9: 5468988-5468988 on Build GRCh38
rs1036106210 Chr.9: 5468933-5468933 on Build GRCh38
rs943773099 Chr.9: 5469033-5469033 on Build GRCh38
rs890847662 Chr.9: 5468823-5468823 on Build GRCh38
rs572241274 Chr.9: 5468809-5468809 on Build GRCh38
rs1021847474 Chr.9: 5468919-5468919 on Build GRCh38
rs578250681 Chr.9: 5468615-5468615 on Build GRCh38
rs925784261 Chr.9: 5468699-5468699 on Build GRCh38
rs919853270 Chr.9: 5468775-5468775 on Build GRCh38
rs565795807 Chr.9: 5468688-5468688 on Build GRCh38
rs1029231124 Chr.9: 5468942-5468942 on Build GRCh38
rs995044635 Chr.9: 5468897-5468897 on Build GRCh38
rs1007967502 Chr.9: 5468836-5468836 on Build GRCh38
rs141907112 Chr.9: 5468616-5468616 on Build GRCh38
rs1055634341 Chr.9: 5468735-5468735 on Build GRCh38
rs1046719528 Chr.9: 5469022-5469022 on Build GRCh38
rs559244638 Chr.9: 5468848-5468848 on Build GRCh38
rs1022025658 Chr.9: 5468667-5468667 on Build GRCh38
rs551235993 Chr.9: 5468880-5468880 on Build GRCh38
rs192397580 Chr.9: 5468901-5468901 on Build GRCh38
rs778829951 Chr.9: 5468932-5468932 on Build GRCh38
rs916397649 Chr.9: 5468981-5468981 on Build GRCh38
rs894418553 Chr.9: 5468759-5468759 on Build GRCh38
rs541020125 Chr.9: 5468948-5468948 on Build GRCh38
rs367673615 Chr.9: 5468835-5468835 on Build GRCh38
rs960400081 Chr.9: 5468730-5468730 on Build GRCh38

View Full Product Details