Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 140784142-140784652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR1
Entrez Gene ID
1038
Gene Name
cerebellar degeneration related protein 1
Gene Aliases
CDR, CDR34, CDR62A
UniGene
Hs.446675
Gene Chromosome Location
Chr.X: 140783260-140784558 on Build GRCh38
Transcripts
NM_004065.2

SNP Detail

SNP ID Chromosome Location
rs998781183 Chr.X: 140784250-140784250 on Build GRCh38
rs375357909 Chr.X: 140784417-140784417 on Build GRCh38
rs757222776 Chr.X: 140784340-140784340 on Build GRCh38
rs149811112 Chr.X: 140784204-140784204 on Build GRCh38
rs746954564 Chr.X: 140784526-140784526 on Build GRCh38
rs185306721 Chr.X: 140784511-140784511 on Build GRCh38
rs756810676 Chr.X: 140784484-140784484 on Build GRCh38
rs777604320 Chr.X: 140784331-140784331 on Build GRCh38
rs190798158 Chr.X: 140784421-140784421 on Build GRCh38
rs753679200 Chr.X: 140784343-140784344 on Build GRCh38
rs147745826 Chr.X: 140784330-140784330 on Build GRCh38
rs769149226 Chr.X: 140784246-140784246 on Build GRCh38
rs982405063 Chr.X: 140784399-140784399 on Build GRCh38
rs750058453 Chr.X: 140784493-140784493 on Build GRCh38
rs766894797 Chr.X: 140784277-140784312 on Build GRCh38
rs35255616 Chr.X: 140784504-140784505 on Build GRCh38
rs773496872 Chr.X: 140784272-140784272 on Build GRCh38
rs781281129 Chr.X: 140784344-140784344 on Build GRCh38
rs866031062 Chr.X: 140784185-140784185 on Build GRCh38
rs867941356 Chr.X: 140784220-140784220 on Build GRCh38
rs773964280 Chr.X: 140784184-140784184 on Build GRCh38
rs767892653 Chr.X: 140784359-140784359 on Build GRCh38
rs771043866 Chr.X: 140784178-140784178 on Build GRCh38
rs774647754 Chr.X: 140784235-140784235 on Build GRCh38
rs749411418 Chr.X: 140784199-140784199 on Build GRCh38
rs775697416 Chr.X: 140784203-140784204 on Build GRCh38
rs771367293 Chr.X: 140784385-140784385 on Build GRCh38
rs928686095 Chr.X: 140784624-140784624 on Build GRCh38
rs142598055 Chr.X: 140784348-140784348 on Build GRCh38
rs766899894 Chr.X: 140784285-140784285 on Build GRCh38
rs955032178 Chr.X: 140784381-140784381 on Build GRCh38
rs766094686 Chr.X: 140784312-140784313 on Build GRCh38
rs762325948 Chr.X: 140784259-140784294 on Build GRCh38
rs761029849 Chr.X: 140784369-140784369 on Build GRCh38
rs371729646 Chr.X: 140784413-140784413 on Build GRCh38
rs777846119 Chr.X: 140784546-140784546 on Build GRCh38
rs773584850 Chr.X: 140784368-140784368 on Build GRCh38
rs761193821 Chr.X: 140784276-140784276 on Build GRCh38
rs184339915 Chr.X: 140784420-140784420 on Build GRCh38
rs137994172 Chr.X: 140784294-140784294 on Build GRCh38
rs893363937 Chr.X: 140784311-140784311 on Build GRCh38
rs1046242172 Chr.X: 140784547-140784547 on Build GRCh38
rs754846657 Chr.X: 140784527-140784527 on Build GRCh38
rs1043018968 Chr.X: 140784491-140784491 on Build GRCh38
rs900796448 Chr.X: 140784520-140784520 on Build GRCh38
rs934683061 Chr.X: 140784628-140784628 on Build GRCh38
rs376600534 Chr.X: 140784180-140784180 on Build GRCh38
rs767892317 Chr.X: 140784252-140784252 on Build GRCh38
rs772205643 Chr.X: 140784365-140784365 on Build GRCh38
rs41299075 Chr.X: 140784312-140784312 on Build GRCh38
rs765045286 Chr.X: 140784492-140784492 on Build GRCh38
rs774900906 Chr.X: 140784259-140784276 on Build GRCh38
rs763350881 Chr.X: 140784240-140784257 on Build GRCh38
rs749968706 Chr.X: 140784290-140784291 on Build GRCh38
rs867837148 Chr.X: 140784273-140784273 on Build GRCh38
rs778862857 Chr.X: 140784397-140784398 on Build GRCh38
rs942394918 Chr.X: 140784510-140784510 on Build GRCh38
rs751335635 Chr.X: 140784339-140784339 on Build GRCh38
rs761192364 Chr.X: 140784195-140784195 on Build GRCh38
rs758180033 Chr.X: 140784511-140784528 on Build GRCh38
rs768844013 Chr.X: 140784350-140784350 on Build GRCh38
rs754878176 Chr.X: 140784345-140784346 on Build GRCh38
rs143559714 Chr.X: 140784205-140784205 on Build GRCh38
rs752394087 Chr.X: 140784325-140784325 on Build GRCh38
rs753275434 Chr.X: 140784295-140784295 on Build GRCh38
rs760219759 Chr.X: 140784295-140784312 on Build GRCh38
rs193025936 Chr.X: 140784426-140784426 on Build GRCh38
rs367868045 Chr.X: 140784247-140784247 on Build GRCh38
rs975564173 Chr.X: 140784349-140784349 on Build GRCh38

View Full Product Details