Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38083724-38084255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
UniGene
Hs.714499
Gene Chromosome Location
Chr.3: 38038595-38122741 on Build GRCh38
Transcripts
NM_001321153.1 NM_007335.3 NM_007337.3 XM_006713438.3 XM_006713439.3 XM_006713440.3 XM_011534317.2 XM_011534318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760063 c.1230G>A p.P410P Chr.3: 38084214-38084214 on Build GRCh38
6644600 c.1220G>A p.R407Q Chr.3: 38084204-38084204 on Build GRCh38
2985806 c.1234T>C p.S412P Chr.3: 38084218-38084218 on Build GRCh38
319903 c.1255G>A p.G419R Chr.3: 38084239-38084239 on Build GRCh38
6644634 c.1229C>T p.P410L Chr.3: 38084213-38084213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563110661 Chr.3: 38083873-38083873 on Build GRCh38
rs527714283 Chr.3: 38083912-38083912 on Build GRCh38
rs757427837 Chr.3: 38083895-38083895 on Build GRCh38
rs138147983 Chr.3: 38084071-38084071 on Build GRCh38
rs778396377 Chr.3: 38084180-38084180 on Build GRCh38
rs375487035 Chr.3: 38084078-38084082 on Build GRCh38
rs374561944 Chr.3: 38084110-38084110 on Build GRCh38
rs556331342 Chr.3: 38083832-38083832 on Build GRCh38
rs575923087 Chr.3: 38083841-38083841 on Build GRCh38
rs11312889 Chr.3: 38083801-38083801 on Build GRCh38
rs199507233 Chr.3: 38084195-38084195 on Build GRCh38
rs1001002115 Chr.3: 38084021-38084021 on Build GRCh38
rs902677255 Chr.3: 38084155-38084155 on Build GRCh38
rs77579651 Chr.3: 38084009-38084009 on Build GRCh38
rs954903820 Chr.3: 38084077-38084081 on Build GRCh38
rs767242024 Chr.3: 38084213-38084213 on Build GRCh38
rs986297017 Chr.3: 38083875-38083875 on Build GRCh38
rs1013357392 Chr.3: 38083753-38083753 on Build GRCh38
rs762811545 Chr.3: 38084201-38084201 on Build GRCh38
rs185036855 Chr.3: 38084067-38084067 on Build GRCh38
rs775694739 Chr.3: 38084122-38084122 on Build GRCh38
rs771423794 Chr.3: 38084186-38084186 on Build GRCh38
rs373114002 Chr.3: 38084226-38084226 on Build GRCh38
rs545707353 Chr.3: 38083992-38083992 on Build GRCh38
rs923505688 Chr.3: 38083808-38083808 on Build GRCh38
rs150427625 Chr.3: 38084037-38084037 on Build GRCh38
rs62240732 Chr.3: 38083754-38083754 on Build GRCh38
rs755277183 Chr.3: 38084178-38084180 on Build GRCh38
rs754728579 Chr.3: 38084208-38084208 on Build GRCh38
rs988416976 Chr.3: 38083804-38083804 on Build GRCh38
rs7632139 Chr.3: 38084100-38084100 on Build GRCh38
rs755549998 Chr.3: 38084179-38084179 on Build GRCh38
rs543421026 Chr.3: 38083766-38083766 on Build GRCh38
rs533472222 Chr.3: 38083962-38083962 on Build GRCh38
rs377205155 Chr.3: 38084203-38084203 on Build GRCh38
rs190980956 Chr.3: 38084006-38084006 on Build GRCh38
rs749767723 Chr.3: 38084185-38084185 on Build GRCh38
rs552051755 Chr.3: 38084232-38084232 on Build GRCh38
rs138178381 Chr.3: 38084169-38084169 on Build GRCh38
rs747764504 Chr.3: 38084177-38084177 on Build GRCh38
rs564398847 Chr.3: 38083946-38083946 on Build GRCh38
rs1011458819 Chr.3: 38083997-38083997 on Build GRCh38
rs773163930 Chr.3: 38084198-38084198 on Build GRCh38
rs777168831 Chr.3: 38083820-38083820 on Build GRCh38
rs1050577080 Chr.3: 38083953-38083953 on Build GRCh38
rs774847885 Chr.3: 38083761-38083764 on Build GRCh38
rs547371627 Chr.3: 38084041-38084041 on Build GRCh38
rs143610524 Chr.3: 38084168-38084168 on Build GRCh38
rs1023847454 Chr.3: 38083965-38083965 on Build GRCh38
rs1012625755 Chr.3: 38084193-38084193 on Build GRCh38
rs146841215 Chr.3: 38083897-38083897 on Build GRCh38
rs760665492 Chr.3: 38084129-38084129 on Build GRCh38
rs146022078 Chr.3: 38084233-38084233 on Build GRCh38
rs931911233 Chr.3: 38084216-38084216 on Build GRCh38
rs991299093 Chr.3: 38083806-38083806 on Build GRCh38
rs140518201 Chr.3: 38084035-38084035 on Build GRCh38
rs143707803 Chr.3: 38084172-38084172 on Build GRCh38
rs889965022 Chr.3: 38083974-38083974 on Build GRCh38
rs760348924 Chr.3: 38084125-38084125 on Build GRCh38
rs776029307 Chr.3: 38084016-38084016 on Build GRCh38
rs547633330 Chr.3: 38084081-38084081 on Build GRCh38
rs371956547 Chr.3: 38084113-38084113 on Build GRCh38
rs368059259 Chr.3: 38084133-38084133 on Build GRCh38
rs910641244 Chr.3: 38083890-38083890 on Build GRCh38
rs529588892 Chr.3: 38084024-38084024 on Build GRCh38
rs907704789 Chr.3: 38084083-38084083 on Build GRCh38
rs942076156 Chr.3: 38083891-38083891 on Build GRCh38
rs78482020 Chr.3: 38084094-38084094 on Build GRCh38
rs947153820 Chr.3: 38084223-38084223 on Build GRCh38
rs956390054 Chr.3: 38084022-38084022 on Build GRCh38
rs753346581 Chr.3: 38083821-38083821 on Build GRCh38
rs777349201 Chr.3: 38084222-38084222 on Build GRCh38
rs201078592 Chr.3: 38084220-38084220 on Build GRCh38
rs774794193 Chr.3: 38084187-38084187 on Build GRCh38
rs7652276 Chr.3: 38084214-38084214 on Build GRCh38
rs564619542 Chr.3: 38083872-38083872 on Build GRCh38
rs149127125 Chr.3: 38084166-38084166 on Build GRCh38
rs746219378 Chr.3: 38084188-38084188 on Build GRCh38
rs780696598 Chr.3: 38084173-38084173 on Build GRCh38
rs375938072 Chr.3: 38084111-38084111 on Build GRCh38
rs1024100261 Chr.3: 38084054-38084054 on Build GRCh38
rs371636962 Chr.3: 38083909-38083909 on Build GRCh38
rs369726203 Chr.3: 38084204-38084204 on Build GRCh38
rs182227149 Chr.3: 38084061-38084061 on Build GRCh38
rs767506017 Chr.3: 38084119-38084119 on Build GRCh38
rs960961962 Chr.3: 38084043-38084043 on Build GRCh38
rs770393404 Chr.3: 38084107-38084107 on Build GRCh38
rs960801649 Chr.3: 38083803-38083803 on Build GRCh38
rs199530367 Chr.3: 38083812-38083812 on Build GRCh38
rs199770928 Chr.3: 38084194-38084194 on Build GRCh38
rs978098243 Chr.3: 38083801-38083801 on Build GRCh38
rs927966679 Chr.3: 38083802-38083802 on Build GRCh38
rs969539318 Chr.3: 38083757-38083757 on Build GRCh38
rs749620889 Chr.3: 38084049-38084049 on Build GRCh38
rs760922898 Chr.3: 38083815-38083818 on Build GRCh38
rs186643938 Chr.3: 38083926-38083926 on Build GRCh38
rs756342989 Chr.3: 38084101-38084101 on Build GRCh38
rs984852924 Chr.3: 38084202-38084202 on Build GRCh38
rs375361669 Chr.3: 38084115-38084115 on Build GRCh38
rs181521957 Chr.3: 38083752-38083752 on Build GRCh38
rs1046995037 Chr.3: 38083924-38083924 on Build GRCh38
rs78079267 Chr.3: 38084140-38084140 on Build GRCh38
rs549729193 Chr.3: 38084034-38084034 on Build GRCh38
rs529472287 Chr.3: 38084191-38084191 on Build GRCh38
rs772402423 Chr.3: 38084190-38084190 on Build GRCh38

View Full Product Details