Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140671092-140671587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365802 c.872A>T p.Q291L Chr.5: 140671483-140671483 on Build GRCh38
5076432 c.812_813delCT p.P271fs*47 Chr.5: 140671542-140671543 on Build GRCh38
4005853 c.879G>A p.V293V Chr.5: 140671476-140671476 on Build GRCh38
5867444 c.1027G>A p.A343T Chr.5: 140671328-140671328 on Build GRCh38
448706 c.1000G>A p.E334K Chr.5: 140671355-140671355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542752474 Chr.5: 140671491-140671491 on Build GRCh38
rs561649345 Chr.5: 140671172-140671172 on Build GRCh38
rs558302892 Chr.5: 140671198-140671198 on Build GRCh38
rs201004091 Chr.5: 140671283-140671283 on Build GRCh38
rs141183092 Chr.5: 140671335-140671335 on Build GRCh38
rs775582051 Chr.5: 140671364-140671364 on Build GRCh38
rs760841858 Chr.5: 140671375-140671375 on Build GRCh38
rs915542927 Chr.5: 140671187-140671187 on Build GRCh38
rs779330167 Chr.5: 140671321-140671321 on Build GRCh38
rs182959393 Chr.5: 140671143-140671143 on Build GRCh38
rs966933092 Chr.5: 140671445-140671445 on Build GRCh38
rs781060102 Chr.5: 140671254-140671254 on Build GRCh38
rs534338256 Chr.5: 140671250-140671250 on Build GRCh38
rs567842974 Chr.5: 140671192-140671194 on Build GRCh38
rs938252548 Chr.5: 140671154-140671154 on Build GRCh38
rs528686052 Chr.5: 140671138-140671138 on Build GRCh38
rs747135715 Chr.5: 140671495-140671495 on Build GRCh38
rs560601583 Chr.5: 140671501-140671501 on Build GRCh38
rs1051579103 Chr.5: 140671296-140671296 on Build GRCh38
rs202148033 Chr.5: 140671433-140671433 on Build GRCh38
rs768864950 Chr.5: 140671499-140671499 on Build GRCh38
rs746550288 Chr.5: 140671243-140671243 on Build GRCh38
rs780853297 Chr.5: 140671330-140671330 on Build GRCh38
rs752862304 Chr.5: 140671312-140671312 on Build GRCh38
rs749094781 Chr.5: 140671457-140671457 on Build GRCh38
rs77880328 Chr.5: 140671355-140671355 on Build GRCh38
rs760828500 Chr.5: 140671251-140671251 on Build GRCh38
rs201638404 Chr.5: 140671309-140671309 on Build GRCh38
rs755793060 Chr.5: 140671449-140671449 on Build GRCh38
rs562782882 Chr.5: 140671343-140671343 on Build GRCh38
rs377680919 Chr.5: 140671563-140671563 on Build GRCh38
rs759003755 Chr.5: 140671418-140671418 on Build GRCh38
rs749074334 Chr.5: 140671345-140671345 on Build GRCh38
rs374411522 Chr.5: 140671363-140671363 on Build GRCh38
rs774536148 Chr.5: 140671446-140671465 on Build GRCh38
rs148639416 Chr.5: 140671322-140671322 on Build GRCh38
rs770818185 Chr.5: 140671307-140671307 on Build GRCh38
rs982410146 Chr.5: 140671141-140671141 on Build GRCh38
rs778776058 Chr.5: 140671479-140671479 on Build GRCh38
rs769031146 Chr.5: 140671387-140671388 on Build GRCh38
rs772019475 Chr.5: 140671485-140671485 on Build GRCh38
rs145312136 Chr.5: 140671357-140671357 on Build GRCh38
rs747815082 Chr.5: 140671259-140671259 on Build GRCh38
rs756941934 Chr.5: 140671461-140671461 on Build GRCh38
rs929116535 Chr.5: 140671219-140671219 on Build GRCh38
rs768325712 Chr.5: 140671253-140671253 on Build GRCh38
rs200389551 Chr.5: 140671376-140671376 on Build GRCh38
rs764406836 Chr.5: 140671313-140671313 on Build GRCh38
rs868843075 Chr.5: 140671161-140671161 on Build GRCh38
rs536894233 Chr.5: 140671318-140671319 on Build GRCh38
rs777631380 Chr.5: 140671452-140671452 on Build GRCh38
rs62384220 Chr.5: 140671341-140671341 on Build GRCh38
rs767711054 Chr.5: 140671310-140671310 on Build GRCh38
rs749518753 Chr.5: 140671336-140671337 on Build GRCh38
rs1021033003 Chr.5: 140671430-140671430 on Build GRCh38
rs773644539 Chr.5: 140671561-140671561 on Build GRCh38
rs757761382 Chr.5: 140671320-140671320 on Build GRCh38
rs772076500 Chr.5: 140671358-140671358 on Build GRCh38
rs539661248 Chr.5: 140671195-140671195 on Build GRCh38
rs201692447 Chr.5: 140671530-140671530 on Build GRCh38
rs777024506 Chr.5: 140671389-140671389 on Build GRCh38
rs111396059 Chr.5: 140671153-140671153 on Build GRCh38
rs555889656 Chr.5: 140671263-140671263 on Build GRCh38
rs754224591 Chr.5: 140671319-140671319 on Build GRCh38
rs967278387 Chr.5: 140671128-140671128 on Build GRCh38
rs753038745 Chr.5: 140671226-140671226 on Build GRCh38
rs773172471 Chr.5: 140671297-140671297 on Build GRCh38
rs765762813 Chr.5: 140671413-140671413 on Build GRCh38
rs765476369 Chr.5: 140671541-140671541 on Build GRCh38
rs897685205 Chr.5: 140671208-140671208 on Build GRCh38
rs550863751 Chr.5: 140671130-140671130 on Build GRCh38
rs146252353 Chr.5: 140671424-140671424 on Build GRCh38
rs941816713 Chr.5: 140671188-140671188 on Build GRCh38
rs1017627641 Chr.5: 140671354-140671354 on Build GRCh38
rs754385447 Chr.5: 140671329-140671329 on Build GRCh38
rs762898641 Chr.5: 140671306-140671306 on Build GRCh38
rs747762188 Chr.5: 140671340-140671340 on Build GRCh38
rs200835062 Chr.5: 140671462-140671462 on Build GRCh38
rs770894107 Chr.5: 140671349-140671349 on Build GRCh38
rs750826384 Chr.5: 140671415-140671415 on Build GRCh38
rs762152927 Chr.5: 140671406-140671406 on Build GRCh38
rs573335362 Chr.5: 140671201-140671201 on Build GRCh38

View Full Product Details