Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37522565-37523071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
UniGene
Hs.709527
Gene Chromosome Location
Chr.9: 37509920-37576459 on Build GRCh38
Transcripts
NM_012166.2 XM_005251439.4 XM_006716754.3 XM_017014618.1 XM_017014619.1

SNP Detail

SNP ID Chromosome Location
rs957003398 Chr.9: 37522726-37522726 on Build GRCh38
rs980669285 Chr.9: 37522861-37522861 on Build GRCh38
rs754995783 Chr.9: 37522862-37522862 on Build GRCh38
rs1036921528 Chr.9: 37522805-37522805 on Build GRCh38
rs1028513726 Chr.9: 37523010-37523010 on Build GRCh38
rs758186106 Chr.9: 37522826-37522826 on Build GRCh38
rs370454432 Chr.9: 37522901-37522901 on Build GRCh38
rs901339104 Chr.9: 37522646-37522646 on Build GRCh38
rs961926629 Chr.9: 37522795-37522795 on Build GRCh38
rs765172172 Chr.9: 37522793-37522793 on Build GRCh38
rs78562944 Chr.9: 37522596-37522596 on Build GRCh38
rs764790902 Chr.9: 37522985-37522985 on Build GRCh38
rs374397106 Chr.9: 37522836-37522836 on Build GRCh38
rs756741985 Chr.9: 37522792-37522792 on Build GRCh38
rs779913467 Chr.9: 37522837-37522837 on Build GRCh38
rs781225957 Chr.9: 37522864-37522864 on Build GRCh38
rs761379472 Chr.9: 37522965-37522965 on Build GRCh38
rs562142793 Chr.9: 37522990-37522990 on Build GRCh38
rs953194962 Chr.9: 37522741-37522741 on Build GRCh38
rs185558370 Chr.9: 37522932-37522932 on Build GRCh38
rs989920846 Chr.9: 37522727-37522727 on Build GRCh38
rs374133948 Chr.9: 37522918-37522918 on Build GRCh38
rs527891818 Chr.9: 37523013-37523013 on Build GRCh38
rs377542171 Chr.9: 37522774-37522774 on Build GRCh38
rs939939138 Chr.9: 37522799-37522799 on Build GRCh38
rs563845807 Chr.9: 37522635-37522635 on Build GRCh38
rs921012396 Chr.9: 37522730-37522730 on Build GRCh38
rs374192200 Chr.9: 37522859-37522859 on Build GRCh38
rs984437030 Chr.9: 37522755-37522755 on Build GRCh38
rs750744001 Chr.9: 37522913-37522914 on Build GRCh38
rs758076207 Chr.9: 37522997-37522997 on Build GRCh38
rs751384570 Chr.9: 37523003-37523003 on Build GRCh38
rs773369800 Chr.9: 37522915-37522915 on Build GRCh38
rs570973613 Chr.9: 37523037-37523037 on Build GRCh38
rs867339197 Chr.9: 37522963-37522963 on Build GRCh38
rs765906116 Chr.9: 37523002-37523002 on Build GRCh38
rs763052327 Chr.9: 37522865-37522865 on Build GRCh38
rs966919358 Chr.9: 37522655-37522655 on Build GRCh38
rs539580900 Chr.9: 37522817-37522817 on Build GRCh38
rs748286174 Chr.9: 37522877-37522877 on Build GRCh38
rs559306543 Chr.9: 37522605-37522605 on Build GRCh38
rs372703709 Chr.9: 37522950-37522950 on Build GRCh38
rs12343402 Chr.9: 37522815-37522815 on Build GRCh38
rs768065121 Chr.9: 37522935-37522935 on Build GRCh38
rs761323274 Chr.9: 37522682-37522682 on Build GRCh38
rs775949744 Chr.9: 37522936-37522936 on Build GRCh38
rs10973394 Chr.9: 37522675-37522675 on Build GRCh38
rs888854240 Chr.9: 37523015-37523015 on Build GRCh38
rs754281940 Chr.9: 37522640-37522640 on Build GRCh38
rs193296204 Chr.9: 37522668-37522668 on Build GRCh38
rs532730550 Chr.9: 37522647-37522647 on Build GRCh38
rs892487122 Chr.9: 37522934-37522934 on Build GRCh38
rs572067689 Chr.9: 37522641-37522641 on Build GRCh38
rs547870811 Chr.9: 37523017-37523017 on Build GRCh38
rs770997936 Chr.9: 37522919-37522919 on Build GRCh38
rs766431893 Chr.9: 37522987-37522987 on Build GRCh38
rs548906150 Chr.9: 37522933-37522933 on Build GRCh38
rs971978651 Chr.9: 37522806-37522806 on Build GRCh38
rs879330496 Chr.9: 37522899-37522899 on Build GRCh38
rs1015913478 Chr.9: 37523021-37523021 on Build GRCh38
rs756316067 Chr.9: 37523026-37523026 on Build GRCh38
rs375802633 Chr.9: 37523006-37523006 on Build GRCh38
rs747950600 Chr.9: 37522627-37522627 on Build GRCh38
rs1014167218 Chr.9: 37522601-37522601 on Build GRCh38

View Full Product Details